Sigma-Aldrich
Analytical / Chromatography > Analytical Reagents > Chromatography > Gas Chromatography > GC Solvents > ENVISOLV®
GC Solvents
ENVISOLV®
Solvents for residue analysis of dioxins, furans and PCB’s
Concept:
ENVISOLV® solvents from Sigma-Aldrich are GC-MS tested. The range of ENVISOLV® products contain less than 5 pg/l (5 ppq) of the 17 relevant dibenzodioxins and dibenzofurans listed below (see Table 1).
Table 1
Dibenzodioxins
Dibenzofurans
2,3,7,8-tetra-CDD 2,3,7,8-tetra-CDF
1,2,3,7,8-penta-CDD 2,3,4,7,8-penta-CDF
1,2,3,4,7,8-hexa-CDD 1,2,3,7,8/1,2,3,4,8-penta-CDF
1,2,3,6,7,8-hexa-CDD 1,2,3,4,7,8/1,2,3,4,7,9-hexa-CDF
1,2,3,7,8,9-hexa-CDD 1,2,3,6,7,8-hexa-CDF
1,2,3,4,6,7,8-hepta-CDD 1,2,3,7,8,9-hexa-CDF
Octa-CDD 2,3,4,6,7,8-hexa-CDF
- 11,2,3,4,6,7,8-hepta-CDF
- 1,2,3,4,7,8,9-hepta-CDF
- Octa-CDF
Table 2. Specification / Product Range
Acetone
31062
2.5 l / 7 l
Dichloromethane
34411
2.5 l / 7 l
Hexane
34412
2.5 l / 7 l
Toluene
34413
2.5 l / 7 l
assay (GC)
min. 99.8 %
min. 99.8 %
min. 95 %
min. 99.7 %
non-volatile matter
max. 0.0005 %
max. 0.0005 %
max. 0.0005 %
max. 0.0005 %
water (Karl Fischer)
max. 0.2 %
max. 0.02 %
max. 0.01 %
max. 0.02 %
suitability for GC-MS of dioxines, furanes, PCB’s
complying
complying
complying
complying
Back to Top
Back to GC Solvents Home
Back to GC Home