Fluka’s GC solvents have been developed for residue analysis in application fields such as environmental analysis and food and beverage control. All solvents are manufactured and bottled under oxygen-free conditions and sealed with a PTFE-lined cap to prevent product contamination and degradation.
Fluka’s GC solvents are free of impurities which would show greater signals than 5 ng/l lindane, in the GC/ECD retention time range of lindane to DDT.
Fluka’s GC solvents for trace analysis are controlled for high volatile halogenated hydrocarbons: With GC/ECD in the corresponding retention volume ranges (methylene chloride-pentachloroethane) are no impurities present with a signal greater than 10 μg/l in each range.
Fluka’s GC solvents, Purge & Trap Grade are suitable for GC/MS analysis of volatile organics in water and oil sediment samples according to the EPA purge & trap methods 601, 624 and 8240.
Product Range
| Catalog No. |
Product Name |
Purity |
Package Size |
| 00562 |
Acetone |
>99.8% (GC) |
1 l, 2.5 l |
| 00689 |
Acetonitrile |
>99.9% (GC) |
1 l, 2.5 l |
| 20246 |
tert-Butyl methyl ether |
>99.8% (GC) |
1 l, 2.5 l |
| 25669 |
Chloroform |
>99.8% (GC), stab. with ~1% ethanol |
1 l, 2.5 l |
| 28919 |
Cyclohexane |
>99.8% (GC) |
1 l, 2.5 l |
| 66741 |
Dichloromethane |
>99.8% (GC), stab. with ~0.002% amylene |
1 l, 2.5 l |
| 31671 |
Diethyl ether |
>99.8% (GC), stab. with ~2% ethanol |
1 l, 2.5 l |
| 40243 |
N,N-Dimethylformamide |
>99.8% (GC) |
1 l, 2.5 l |
| 02851 |
Ethanol |
>99.8% (v/v) |
1 l, 2.5 l |
| 45758 |
Ethyl acetate |
>99.8% (GC) |
1 l, 2.5 l |
| 52767 |
Hexane |
>99.0% (GC) |
1 l, 2.5 l |
| 52756 |
Hexane |
>95% (GC), for trace analysis |
1 l, 2.5 l |
| 52749 |
Hexane |
>95% (GC) |
1 l, 2.5 l |
| 59025 |
Isooctane |
>99.5% (GC) |
1 l, 2.5 l |
| 65554 |
Methanol |
>99.8% (GC), for trace analysis |
1 l |
| 65539 |
Methanol |
>99.8% (GC) |
1 l, 2.5 l |
| 65553 |
Methanol |
>99.8% (GC), Purge & Trap Grade |
1 l |
| 76869 |
Pentane |
>99.0% (GC) |
1 l, 2.5 l |
| 76866 |
Pentane |
>99.0% (GC), for trace analysis |
1 l, 2.5 l |
| 00689 |
Pentane |
>99.0% (GC) |
1 l, 2.5 l |
| 77397 |
Petroleum ether |
bp 40-60°C |
1 l |
| 59301 |
2-Propanol |
>99.8% (GC) |
1 l, 2.5 l |
| 89676 |
Toluene |
>99.8% (GC) |
1 l, 2.5 l |
Back to Top
Back to GC Solvents Home
Back to GC Home |