Sigma-Aldrich
Analytical / Chromatography > Analytical Reagents > Chromatography > Gas Chromatography > GC Solvents > Fluka GC Solvents
GC Solvents
Fluka GC Solvents
Fluka’s GC solvents have been developed for residue analysis in application fields such as environmental analysis and food and beverage control. All solvents are manufactured and bottled under oxygen-free conditions and sealed with a PTFE-lined cap to prevent product contamination and degradation.
Fluka’s GC solvents are free of impurities which would show greater signals than 5 ng/l lindane, in the GC/ECD retention time range of lindane to DDT.
Fluka’s GC solvents for trace analysis are controlled for high volatile halogenated hydrocarbons: With GC/ECD in the corresponding retention volume ranges (methylene chloride-pentachloroethane) are no impurities present with a signal greater than 10 μg/l in each range.
Fluka’s GC solvents, Purge & Trap Grade are suitable for GC/MS analysis of volatile organics in water and oil sediment samples according to the EPA purge & trap methods 601, 624 and 8240.
Product Range
Catalog No.
Product Name
Purity
Package Size
00562
Acetone >99.8% (GC)
1 l, 2.5 l
00689
Acetonitrile >99.9% (GC)
1 l, 2.5 l
20246
tert-Butyl methyl ether >99.8% (GC)
1 l, 2.5 l
25669
Chloroform >99.8% (GC), stab. with ~1% ethanol
1 l, 2.5 l
28919
Cyclohexane >99.8% (GC)
1 l, 2.5 l
66741
Dichloromethane >99.8% (GC), stab. with ~0.002% amylene
1 l, 2.5 l
31671
Diethyl ether >99.8% (GC), stab. with ~2% ethanol
1 l, 2.5 l
40243
N,N-Dimethylformamide >99.8% (GC)
1 l, 2.5 l
02851
Ethanol >99.8% (v/v)
1 l, 2.5 l
45758
Ethyl acetate >99.8% (GC)
1 l, 2.5 l
52767
Hexane >99.0% (GC)
1 l, 2.5 l
52756
Hexane >95% (GC), for trace analysis
1 l, 2.5 l
52749
Hexane >95% (GC)
1 l, 2.5 l
59025
Isooctane >99.5% (GC)
1 l, 2.5 l
65554
Methanol >99.8% (GC), for trace analysis
1 l
65539
Methanol >99.8% (GC)
1 l, 2.5 l
65553
Methanol >99.8% (GC), Purge & Trap Grade
1 l
76869
Pentane >99.0% (GC)
1 l, 2.5 l
76866
Pentane >99.0% (GC), for trace analysis
1 l, 2.5 l
00689
Pentane >99.0% (GC)
1 l, 2.5 l
77397
Petroleum ether bp 40-60°C
1 l
59301
2-Propanol >99.8% (GC)
1 l, 2.5 l
89676
Toluene >99.8% (GC)
1 l, 2.5 l
Back to Top
Back to GC Solvents Home
Back to GC Home