Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Gas Chromatography
GC Standards
Gas Chromatography (GC) owes its enormous growth in part to its speed, simplicity, relatively low cost, and wide applicability as a separation tool. Gas Chromatography (GC) is widely used for determining the presence or absence of components.
The simplest method of identification of a chromatographic peak is comparison of its retention time with that of an authentic sample of the suspected compound. The most reliable way to do this is by co-chromatography, in which the authentic sample is added to the unknown.
The most straightforward method for quantitative chromatographic analyses involves the preparation of a series of standard solutions that approximate the composition of the unknown. Chromatograms for the standards are then obtained, and peak areas are plotted as a function of concentration.
GC standards are mainly used in:
  • Pharmaceutical Industry
  • Environmental Laboratories
  • Flavor & Fragrance Companies
  • Petrol Industry
For all purposes, high purity standards are required to exclude impurity peaks which come from the standard. Fluka’s GC product range includes more than 400 standards belonging to various organic structure classes. Please take a look at the sections of your interest by just clicking on them:
Table 1. Alkane Standards
Catalog No.
Product Name
CAS No.
Description
Package Size
30540
Decane
124-18-5
puriss. p.a. standard for GC, >99.8% (GC)
5 ml, 25 ml
32948
4,4'-Diaminodiphenylmethane
101-77-9
puriss. p.a. standard for GC, >99.0% (GC)
1 g, 5 g
39730
2,2-Dimethylbutane
75-83-2
puriss. p.a. standard for GC, >99.5% (GC)
5 ml, 10 ml
39760
2,3-Dimethylbutane
79-29-8
puriss. p.a. standard for GC, >99.5% (GC)
5 ml, 10 ml
40512
3,4-Dimethylhexane
583-48-2
puriss. p.a. standard for GC, >99.5% (GC)
5 ml, 10 ml, 100 ml
41085
2,3-Dimethylpentane
565-59-3
puriss. p.a. standard for GC, >99.5% (GC)
1 ml, 5 ml
43942
Docosane
629-97-0
puriss. p.a. standard for GC, >99.5% (GC)
1 g, 5 g
44010
Dodecane
112-40-3
puriss. p.a. standard for GC, >99.8% (GC)
5 ml, 25 ml
44249
Dopentacontane
7719-79-1
p.a. standard for GC, ~95% (GC)
250 mg, 1 g
44250
Dotetracontane
7098-20-6
puriss. p.a. standard for GC, >99.5% (GC)
100 mg, 500 mg
44253
Dotriacontane
544-85-4
puriss. p.a. standard for GC, >99.5% (GC)
250 mg, 1 g
44818
Eicosane
112-95-8
puriss. p.a. standard for GC, >99.8% (GC)
1 g, 5 g
51523
Heneicosane
629-94-7
puriss. p.a. standard for GC, >99.5% (GC)
1 g, 5 g
51531
Hentetracontane
7194-87-8
puriss. p.a. standard for GC, >99.0% (GC)
250 mg, 1 g
51529
Hentriacontane
630-04-6
puriss. p.a. standard for GC, >99.5% (GC)
250 mg, 1 g
51559
Heptacosane
593-49-7
puriss. p.a. standard for GC, >99.5% (GC)
250 mg, 1 g
51578
Heptadecane
629-78-7
puriss. p.a. standard for GC, >99.8% (GC)
1 ml, 5 ml
51730
Heptane
142-82-5
puriss. p.a. standard for GC, >99.8% (GC)
5 ml, 10 ml, 50 ml
51848
Heptatriacontane
7194-84-5
puriss. p.a. standard for GC, >99.5% (GC)
250 mg, 1 g
52183
Hexacosane
630-01-3
puriss. p.a. standard for GC, >99.5% (GC)
250 mg, 1 g
52209
Hexadecane
544-76-3
puriss. p.a. standard for GC, >99.8% (GC)
5 ml, 25 ml
52750
Hexane
110-54-3
puriss. p.a. standard for GC, >99.7% (GC)
5 ml, 10 ml, 50 ml
52908
Hexapentacontane
7719-82-6
purum p.a. standard for GC, >97.0% (GC)
100 mg, 500 mg
52913
Hexatetracontane
7098-24-0
puriss. p.a. standard for GC, >99.0% (GC)
250 mg, 1 g
52919
Hexatriacontane
630-06-8
puriss. p.a. standard for GC, >99.5% (GC)
250 mg, 1 g
59030
Isooctane
540-84-1
puriss. p.a. standard for GC, >99.7% (GC)
5 ml, 10 ml, 50 ml
59060
2-Methylbutane
78-78-4
puriss. p.a. standard for GC, >99.7% (GC)
5 ml, 10 ml, 50 ml
68310
2-Methylpentane
107-83-5
puriss. p.a. standard for GC, >99.5% (GC)
5 ml, 10 ml, 50 ml
68320
3-Methylpentane
96-14-0
puriss. p.a. standard for GC, >99.5% (GC)
5 ml, 10 ml, 50 ml
74156
Nonacosane
630-03-5
puriss. p.a. standard for GC, >99.5% (GC)
250 mg, 1 g
74158
Nonadecane
629-92-5
puriss. p.a. standard for GC, >99.8% (GC)
5 g
74250
Nonane
111-84-2
puriss. p.a. standard for GC, >99.8% (GC)
5 ml
74321
Nonatriacontane
7194-86-7
puriss. p.a. standard for GC, >99.5% (GC)
250 mg, 1 g
74684
Octacosane
630-02-4
puriss. p.a. standard for GC, >99.5% (GC)
250 mg, 1 g
74691
Octadecane
593-45-3
puriss. p.a. standard for GC, >99.8% (GC)
1 g, 5 g
74820
Octane
111-65-9
puriss. p.a. standard for GC, >99.8% (GC)
5 ml, 10 ml, 50 ml
74892
Octatetracontane
7098-26-2
puriss. p.a. standard for GC, ~99% (GC)
100 mg
76493
Pentacosane
629-99-2
puriss. p.a. standard for GC, >99.5% (GC)
250 mg, 1 g
76509
Pentadecane
629-62-9
puriss. p.a. standard for GC, >99.8% (GC)
5 ml, 25 ml
76870
Pentane
109-66-0
puriss. p.a. standard for GC, >99.8% (GC)
5 ml, 10 ml, 50 ml
76968
Pentatriacontane
630-07-9
puriss. p.a. standard for GC, >99.5% (GC)
250 mg, 1 g
78366
1-Phenyldecane
104-72-3
puriss. p.a. standard for GC, >99.5% (GC)
500 mg
78638
1-Phenylheptane
1078-71-3
puriss. p.a. standard for GC, >99.8% (GC)
1 ml, 5 ml
80165
Phytane
638-36-8
puriss. p.a. standard for GC, ~99% (GC)
50 mg
87086
Tetracontane
4181-95-7
puriss. p.a. standard for GC, >99.5% (GC)
250 mg, 1 g
87089
Tetracosane
646-31-1
puriss. p.a. standard for GC, >99.5% (GC)
1 g, 5 g
87139
Tetradecane olefine free
629-59-4
puriss. p.a. standard for GC, >99.5% (GC)
5 ml, 25 ml
87992
Tetrapentacontane
5856-66-6
puriss. p.a. standard for GC, >99.0% (GC)
100 mg, 500 mg
88144
Tetratetracontane
7098-22-8
puriss. p.a. standard for GC, ~99% (GC)
250 mg, 1 g
88152
Tetratriacontane
14167-59-0
puriss. p.a. standard for GC, >99.5% (GC)
1 g
88152
Tetratriacontane
14167-59-0
puriss. p.a. standard for GC, >99.5% (GC)
250 mg
90270
Triacontane
638-68-6
puriss. p.a. standard for GC, >99.5% (GC)
250 mg, 1 g
91447
Tricosane
638-67-5
puriss. p.a. standard for GC, >99.5% (GC)
1 g, 5 g
91490
Tridecane
629-50-5
puriss. p.a. standard for GC, >99.5% (GC)
5 ml, 10 ml, 50 ml
93435
Tritriacontane
630-05-7
puriss. p.a. standard for GC, >99.5% (GC)
250 mg, 1 g
94000
Undecane
1120-21-4
puriss. p.a. standard for GC, >99.8% (GC)
5 ml, 25 ml
Back to Top
Table 2. Alkene standards
Catalog No.
Product Name
CAS No.
Description
Package Size
30649
1-Decene
872-05-9
puriss. p.a. standard for GC, >99.5% (GC)
5 ml, 10 ml, 50 ml
39833
3,3-Dimethyl-1-butene
558-37-2
puriss. p.a. standard for GC, >99.5% (GC)
1 ml, 5 ml
44146
1-Dodecene
112-41-4
puriss. p.a. standard for GC, >99.8% (GC)
1 ml, 5 ml
51856
1-Heptene
592-76-7
puriss. p.a. standard for GC, >99.5% (GC)
1 ml, 5 ml
52276
1-Hexadecene
629-73-2
puriss. p.a. standard for GC, >99.8% (GC)
1 ml, 5 ml
52930
1-Hexene
592-41-6
puriss. p.a. standard for GC, >99.8% (GC)
5 ml, 10 ml, 50 ml
59240
Isoprene , stab.
78-79-5
puriss. p.a. standard for GC, >99.5% (GC), stab. with ~0.02% 4-tert-butylcatechol
1 ml, 5 ml, 10 ml
66030
2-Methyl-1-butene
563-46-2
puriss. p.a. standard for GC, >99.5% (GC)
5 ml, 10 ml, 50 ml
66050
2-Methyl-2-butene
513-35-9
puriss. p.a. standard for GC, >99.5% (GC)
5 ml, 10 ml, 50 ml
66070
3-Methyl-1-butene
563-45-1
puriss. p.a. standard for GC, >99.0% (GC)
Vial of 10 ml
68450
2-Methyl-1-pentene
763-29-1
puriss. p.a. standard for GC, >99.5% (GC)
5 ml, 25 ml
68510
4-Methyl-1-pentene
691-37-2
puriss. p.a. standard for GC, >99.5% (GC)
5 ml, 25 ml
74230
1-Nonadecene
18435-45-5
puriss. p.a. standard for GC, >99.0% (GC)
1 g, 5 g
74323
1-Nonene
124-11-8
puriss. p.a. standard for GC, >99.5% (GC)
1 ml, 5 ml
74738
1-Octadecene
112-88-9
puriss. p.a. standard for GC, >99.8% (GC)
1 ml, 5 ml
74900
1-Octene
111-66-0
puriss. p.a. standard for GC, >99.8% (GC)
5 ml, 10 ml, 50 ml
76570
1-Pentadecene
13360-61-7
puriss. p.a. standard for GC, >99.5% (GC)
1 ml, 5 ml
76969
1-Pentene
109-67-1
puriss. p.a. standard for GC, >99.5% (GC)
1 ml, 5 ml, 50 ml
87187
1-Tetradecene
1120-36-1
puriss. p.a. standard for GC, >99.8% (GC)
1 ml, 5 ml, 25 ml
91578
1-Tridecene
2437-56-1
puriss. p.a. standard for GC, >99.5% (GC)
1 ml, 5 ml
92670
2,4,4-Trimethyl-1-pentene
107-39-1
puriss. p.a. standard for GC, >99.5% (GC)
5 ml, 25 ml
92678
2,4,4-Trimethyl-2-pentene
107-40-4
puriss. p.a. standard for GC, >99.5% (GC)
1 ml, 5 ml
94138
1-Undecene
821-95-4
puriss. p.a. standard for GC, >99.5% (GC)
1 ml, 5 ml
Back to Top
Table 3. Aromatic Hydrocarbons
Catalog No.
Product Name
CAS No.
Description
Package Size
12540
Benzene
71-43-2
puriss. p.a. standard for GC, >99.9% (GC)
5 ml, 10 ml, 50 ml
19620
sec-Butylbenzene
135-98-8
puriss. p.a. standard for GC, >99.5% (GC)
5 ml, 10 ml
19640
tert-Butylbenzene
98-06-6
puriss. p.a. standard for GC, >99.5% (GC)
5 ml, 10 ml
19600
Butylbenzene
104-51-8
puriss. p.a. standard for GC, >99.8% (GC)
5 ml
28220
Cumene
98-82-8
puriss. p.a. standard for GC, >99.5% (GC)
5 ml, 25 ml
30037
m-Cymene
535-77-3
puriss. p.a. standard for GC, >99.5% (GC)
250 mg, 1 g
30039
p-Cymene
99-87-6
puriss. p.a. standard for GC, >99.5% (GC)
1 ml, 5 ml
33359
2,4-Diaminotoluene
95-80-7
puriss. p.a. standard for GC, >99.0% (GC)
1 g, 5 g
32018
1,4-Diethylbenzene
105-05-5
puriss. p.a. standard for GC, >99. 5% (GC)
1 ml, 5 ml
38353
1,4-Diisopropylbenzene
100-18-5
puriss. p.a. standard for GC, >99.5% (GC)
1 ml, 5 ml
44178
Dodecylbenzene
123-01-3
puriss. p.a. standard for GC, >99.5% (GC)
500 mg
03079
Ethylbenzene
100-41-4
puriss. p.a. standard for GC, >99.5% (GC)
5 ml, 10 ml
51668
Heptadecylbenzene
14752-75-1
puriss. p.a. standard for GC, >99.5% (GC)
500 mg
52298
Hexadecylbenzene
1459-09-2
puriss. p.a. standard for GC, >99.5% (GC)
500 mg
53158
Hexylbenzene
1077-16-3
puriss. p.a. standard for GC, >99.8% (GC)
1 ml, 5 ml
58498
Isobutylbenzene
538-93-2
puriss. p.a. standard for GC, >99.5% (GC)
1 ml, 5 ml
63908
Mesitylene
108-67-8
puriss. p.a. standard for GC, >99.8% (GC)
1 ml, 5 ml
66687
4,4'-Methylene-bis(2-methylaniline)
838-88-0
puriss. p.a. standard for GC, >99.0% (GC)
1 g, 5 g
69168
alpha-Methylstyrene monomer
98-83-9
puriss. p.a. standard for GC, >99.5% (GC), stab. with ~0.0025% 4-tert-butylcatechol
1 ml, 5 ml
74238
Nonadecylbenzene
29136-19-4
puriss. p.a. standard for GC, >99.5% (GC)
500 mg
74408
Nonylbenzene
1081-77-2
puriss. p.a. standard for GC, >99.8% (GC)
1 ml, 5 ml
74753
Octadecylbenzene
4445-07-2
puriss. p.a. standard for GC, >99.5% (GC)
500 mg
75008
Octylbenzene
2189-60-8
puriss. p.a. standard for GC, >99.8% (GC)
1 ml, 5 ml
76588
Pentadecylbenzene
2131-18-2
puriss. p.a. standard for GC, >99.5% (GC)
500 mg
77078
Pentylbenzene
538-68-1
puriss. p.a. standard for GC, >99.8% (GC)
1 ml, 5 ml
82118
Propylbenzene
103-65-1
puriss. p.a. standard for GC, >99.8% (GC)
5 ml, 10 ml, 50 ml
82540
Pseudocumene
95-63-6
puriss. p.a. standard for GC, >99.5% (GC)
5 ml, 10 ml, 50 ml
87204
Tetradecylbenzene
1459-10-5
puriss. p.a. standard for GC, >99.5% (GC)
500 mg
89680
Toluene
108-88-3
puriss. p.a. standard for GC, >99.9% (GC)
5 ml, 25 ml
91598
Tridecylbenzene
123-02-4
puriss. p.a. standard for GC, >99.5% (GC)
500 mg
95660
o-Xylene
95-47-6
puriss. p.a. standard for GC, >99.5% (GC)
5 ml, 10 ml, 50 ml
95670
m-Xylene
108-38-3
puriss. p.a. standard for GC, >99.5% (GC)
5 ml, 10 ml, 50 ml
95680
p-Xylene
106-42-3
puriss. p.a. standard for GC, >99.5% (GC)
5 ml, 10 ml, 50 ml
Back to Top
Table 4. Cyclic Hydrocarbons
Catalog No.
Product Name
CAS No.
Description
Package Size
28908
1,4-Cyclohexadiene
628-41-1
puriss. p.a. standard for GC, >99.0% (GC), stab. with ~0.2% 2,6-di-tert-butyl-p-cresol
1 ml, 5 ml
28920
Cyclohexane
110-82-7
puriss. p.a. standard for GC, >99.7% (GC)
5 ml, 10 ml, 50 ml
02482
Cyclohexanone
108-94-1
puriss. p.a. standard for GC, >99.9% (GC)
1 ml
29598
Cyclooctane
292-64-8
puriss. p.a. standard for GC, >99.5% (GC)
1 ml, 5 ml
29648
Cyclooctene
931-88-4
puriss. p.a. standard for GC, >99.5% (GC)
1 ml, 5 ml
29680
Cyclopentane
287-92-3
puriss. p.a. standard for GC, >99.5% (GC)
5 ml, 10 ml, 50 ml
66294
Methylcyclohexane
108-87-2
puriss. p.a. standard for GC, >99.5% (GC)
1 ml, 5 ml
66490
Methylcyclopentane
96-37-7
puriss. p.a. standard for GC, >99.5% (GC)
5 ml, 10 ml, 50 ml
94950
4-Vinyl-1-cyclohexene
100-40-3
puriss. p.a. standard for GC, >99.5% (GC)
5 ml, 25 ml
Back to Top
Table 5. Fatty acid standards
Catalog No.
Product Name
CAS No.
Description
Package Size
11909
Behenic acid
112-85-6
puriss. p.a. standard for GC; >99.0% (GC)
5 g
21409
Capric acid
334-48-5
puriss. p.a. standard for GC; >99.5% (GC)
5 g
21529
Caproic acid
142-62-1
puriss. p.a. standard for GC; >99.5% (GC)
5 ml
21639
Caprylic acid
124-07-2
puriss. p.a. standard for GC; >99.5% (GC)
5 ml
43962
all-cis-7,10,13,16,19-Docosapentaenoicacid
24880-45-3
puriss. p.a. standard for GC; >99.0% (GC)
100 mg, 500 mg
44864
cis-5,8,11,14,17-Eicosapentaenoic acid
10417-94-4
puriss. p.a. standard for GC; ~99% (GC)
100 mg, 500 mg
44874
cis,cis,cis-11,14,17-Eicosatrienoic acid
17046-59-2
puriss. p.a. standard for GC; ~99% (GC)
100 mg, 500 mg
44878
11-Eicosenoic acid
5561-99-9
puriss. p.a. standard for GC; >99.0% (GC)
100 mg, 500 mg
45089
Elaidic acid
112-79-8
puriss. p.a. standard for GC; >99.0% (GC)
1 g, 5 g
45629
Erucic acid
112-86-7
puriss. p.a. standard for GC; >99.5% (GC)
250 mg, 1 g, 5 g
61609
Lauric acid
143-07-7
puriss. p.a. standard for GC; >99.5% (GC)
5 g
62230
Linoleic acid
60-33-3
puriss. p.a. standard for GC; ~99% (GC)
5 ml, 25 ml
62160
Linolenic acid
463-40-1
puriss. p.a. standard for GC; ~99% (GC)
1 ml, 5 ml
62174
gamma-Linolenic acid
506-26-3
puriss. p.a. standard for GC; >99.0% (GC)
100 mg, 500 mg
70079
Myristic acid
544-63-8
puriss. p.a. standard for GC; >99.5% (GC)
5 g
74746
11-Octadecenoic acid
506-17-2
puriss. p.a. standard for GC; ~99% (GC)
100 mg, 500 mg
75090
Oleic acid , natural
112-80-1
puriss. p.a. standard for GC; natural; ~99% (GC)
5 ml, 25 ml
76119
Palmitic acid
57-10-3
puriss. p.a. standard for GC; >99.0% (GC)
5 g
76169
Palmitoleic acid
373-49-9
puriss. p.a. standard for GC; >99.0% (GC)
1 g
85679
Stearic acid
57-11-4
puriss. p.a. standard for GC; >99.5%
5 g
87117
15-Tetracosenoic acid
506-37-6
puriss. p.a. standard for GC; >99.0% (GC)
100 mg, 500 mg
Back to Top
Table 6. Halogenated Compounds
Catalog No.
Product Name
CAS No.
Description
Package Size
02671
Carbon tetrachloride
56-23-5
puriss. p.a. standard for GC; >99.7% (GC)
1 ml, 5 ml
02487
Chloroform
67-66-3
puriss. p.a. standard for GC, >99.9% (GC), stab.
1 ml, 5 ml
24771
1-Chlorohexane
544-10-5
puriss. p.a. standard for GC, >99.5% (GC)
1 ml
25078
4-Chloro-2-methylaniline
95-69-2
puriss. p.a. standard for GC, >99.5% (GC)
1 g
25769
1-Chloropentane
543-59-9
puriss. p.a. standard for GC, >99.5% (GC)
1 ml
02488
1-Chloropropane
540-54-5
puriss. p.a. standard for GC, >99.9% (GC)
1 ml, 5 ml
02489
2-Chloropropane
75-29-6
puriss. p.a. standard for GC, >99. 5% (GC)
1 ml, 5 ml
35037
1,1-Dichloroacetone
513-88-2
puriss. p.a. standard for GC, >99.0% (GC)
1 ml, 5 ml
36085
Dichloroacetonitrile
3018-12-0
puriss. p.a. standard for GC, >99.5% (GC)
1 ml, 5 ml
02562
1,2-Dichloroethane
107-06-2
puriss. p.a. standard for GC, >99.9% (GC)
1 ml, 5 ml
02574
1,1-Dichloroethene
75-35-4
puriss. p.a. standard for GC, >99.9% (GC), stab.
1 ml, 5 ml
02575
Dichloromethane
75-09-2
puriss. p.a. standard for GC, >99.9% (GC), stab.
1 ml, 5 ml
02577
1,2-Dichloropropane
78-87-5
puriss. p.a. standard for GC, >99.7% (GC)
1 ml
02578
(+/-)-Epichlorohydrin
106-89-8
puriss. p.a. standard for GC, >99.9% (GC)
1 ml
66681
4,4'-Methylene-bis(2-chloroaniline)
101-14-4
puriss. p.a. standard for GC, ~99% (GC)
250 mg, 1 g
02664
1,1,1,2-Tetrachloroethane
630-20-6
puriss. p.a. standard for GC, >99.8% (GC)
1 ml
02579
1,1,2,2-Tetrachloroethane
79-34-5
puriss. p.a. standard for GC, >99. 8% (GC)
1 ml, 5 ml
02666
Tetrachloroethylene
127-18-4
puriss. p.a. standard for GC, >99.9% (GC)
1 ml, 5 ml
02669
1,1,1-Trichloroethane
71-55-6
puriss. p.a. standard for GC, >99.9% (GC)
1 ml, 5 ml
02668
1,1,2-Trichloroethane
79-00-5
puriss. p.a. standard for GC, >99.9% (GC)
1 ml, 5 ml
02667
Trichloroethylene
79-01-6
puriss. p.a. standard for GC, >99.9% (GC)
1 ml, 5 ml
02657
1,2,3-Trichloropropane
96-18-4
puriss. p.a. standard for GC, >99.9% (GC)
1 ml
Back to Top
Table 7. Ketones
Catalog No.
Product Name
CAS No.
Description
Package Size
06629
3-Aminoacetophenone
99-03-6
puriss. p.a. standard for GC, >99.5% (GC)
1 g
06638
4-Aminoacetophenone
99-92-3
puriss. p.a. standard for GC, >99.5% (GC)
1 g
02469
2-Butanone
78-93-3
puriss. p.a. standard for GC, >99.9% (GC)
1 ml, 5 ml
02482
Cyclohexanone
108-94-1
puriss. p.a. standard for GC, >99.9% (GC)
1 ml
68228
2-Decanone
693-54-9
puriss. p.a. standard for GC, >99.5% (GC)
1 ml, 5 ml
02476
2-Heptanone
110-43-0
puriss. p.a. standard for GC, >99.9% (GC)
1 ml
02477
3-Heptanone
106-35-4
puriss. p.a. standard for GC, >99.5% (GC)
1 ml, 5 ml
02473
2-Hexanone
591-78-6
puriss. p.a. standard for GC, >99.5% (GC)
1 ml, 5 ml
02474
4-Methyl-2-pentanone
108-10-1
puriss. p.a. standard for GC, >99.7% (GC)
1 ml, 5 ml
02479
2-Octanone
111-13-7
puriss. p.a. standard for GC, >99.5% (GC)
1 ml, 5 ml
02471
3-Pentanone
96-22-0
puriss. p.a. standard for GC, >99.5% (GC)
1 ml
Back to Top
Table 8. Lactone standards
Catalog No.
Product Name
CAS No.
Description
Package Size
00306
(+/-)-5-Decanolide
705-86-2
purum p.a. standard for GC, >98.0% (GC)
1ml, 5 ml
00501
(+/-)-4-Heptanolide
105-21-5
purum p.a. standard for GC; >98.0% (GC)
1ml, 5 ml
00552
(+/-)-5-Dodecanolide
713-95-1
purum p.a. standard for GC; >98.0% (GC)
1ml, 5 ml
00553
(+/-)-4-Dodecanolide
2305-05-7
purum p.a. standard for GC; >98.0% (GC)
1ml, 5 ml
Back to Top
Table 9. Terpene stadnards
Catalog No.
Product Name</font>
CAS No.
Description
Package Size
05818
4-Allylanisole
140-67-0
puriss. p.a. terpene standard, >99.0% (GC)
1 ml, 5 ml
10368
Anethole
104-46-1
puriss. p.a. terpene standard, >99.5% (GC)
1 ml, 5 ml
11107
alpha-Asarone
2883-98-9
puriss. p.a. terpene standard, ~99% (GC)
1 g, 5 g
15598
(-)-Borneol
464-45-9
puriss. p.a. terpene standard, >99.0% (GC, sum of enantiomers)
1 g, 5 g, 25 g
45853
(+)-Bornyl acetate
20347-65-3
puriss. p.a. terpene standard, >99.0% (GC, sum of enantiomers)
1 ml, 5 ml
45855
(-)-Bornyl acetate
5655-61-8
puriss. p.a. terpene standard, >99.0% (GC, sum of enantiomers)
1 ml, 5 ml
21986
(+)-3-Carene
498-15-7
puriss. p.a. terpene standard; ~99% (GC, sum of enantiomers)
5 ml, 25 ml
22070
(+)-Carvone
2244-16-8
puriss. p.a. terpene standard, >99.0 (GC, sum of enantiomers)
1 ml, 25 ml, 100 ml
22060
(-)-Carvone
6485-40-1
puriss. p.a. terpene standard, >99.0% (GC, sum of enantiomers)
1 ml, 5 ml
27393
1,4-Cineole
470-67-7
puriss. p.a. terpene standard, ~99% (GC)
1 ml, 5 ml
27475
(+)-beta-Citronellene
2436-90-0
puriss. p.a. terpene standard, ~99% (GC, sum of enantiomers)
1 ml, 5 ml
27478
(+)-beta-Citronellol
1117-61-9
puriss. p.a. terpene standard, >99.0% (GC, sum of enantiomers)
1 ml, 5 ml
27483
(-)-beta-Citronellol
7540-51-4
puriss. p.a. terpene standard, >99.0% (GC, sum of enantiomers)
1 ml, 5 ml
00138
Dihydrojasmone
1128-08-1
purum p.a. terpene standard, >98.0% (GC)
1 ml
29210
Eucalyptol
470-82-6
puriss. p.a. terpene standard, >99.7% (GC)
1 ml
00138
(+)-Fenchol
2217-02-9
puriss. p.a. terpene standard, >99.0% (GC, sum of enantiomers)
1 g, 5 g
46208
(+)-Fenchone
4695-62-9
puriss. p.a. terpene standard, >99.5% (GC, sum of enantiomers)
1 ml, 5 ml, 25 ml
29242
(-)-Guajol
489-86-1
puriss. p.a. terpene standard, >99.0% (HPLC)
250 mg
58929
(+)-Isomenthol
23283-97-8
puriss. p.a. terpene standard, >99.0% (GC, sum of enantiomers)
1 g, 50 g, 250 g
59765
(+)-Isopulegol
104870-56-6
puriss. p.a. terpene standard, >99.0% (GC, sum of enantiomers)
1 ml, 5 ml
59770
(-)-Isopulegol
89-79-2
puriss. p.a. terpene standard, >99.0% (GC, sum of enantiomers)
1 ml, 5 ml, 25 ml
61736
(+/-)-Lavandulol acetate
50373-59-6
puriss. p.a. terpene standard; >99.0% (GC)
5 ml, 25 ml
62118
R(+)-Limonene
5989-27-5
puriss. p.a. terpene standard, >99.0% (GC, sum of enantiomers)
1 ml, 5 ml, 25 ml
62128
S(-)-Limonene
5989-54-8
puriss. p.a. terpene standard, >99.0% (GC, sum of enantiomers)
1 ml, 5 ml, 25 ml
62136
(+)-trans-Limonene-1,2-epoxide
6909-30-4
puriss. p.a. terpene standard, >99.0% (GC, sum of enantiomers)
1 ml, 5 ml
63655
(+)-p-Menth-1-ene
1195-31-9
puriss. p.a. terpene standard, ~99% (GC, sum of enantiomers)
1 ml, 5 ml
63661
(+)-Menthofuran
17957-94-7
puriss. p.a. terpene standard, >99.0% (GC, sum of enantiomers)
1 ml, 5 ml
63658
(+)-Menthol
15356-60-2
puriss. p.a. terpene standard, >99.0% (GC, sum of enantiomers)
1 g, 10 g, 50 g
63660
(-)-Menthol
2216-51-5
puriss. p.a. terpene standard, >99.0% (GC, sum of enantiomers)
1 g, 100 g, 500 g
63675
(+)-Menthone
3391-87-5
puriss. p.a. terpene standard, >99.0% (GC, sum of enantiomers)
1 ml, 5 ml
63677
(-)-Menthone
14073-97-3
puriss. p.a. terpene standard, >99.0% (GC, sum of enantiomers)
1 ml, 5 ml, 25 ml
45987
(+)-(1S)-Menthyl acetate
5157-89-1
puriss. p.a. terpene standard, >99. 0% (GC, sum of enantiomers)
1 ml, 5 ml
45985
(-)-(1R)-Menthyl acetate
2623-23-6
puriss. p.a. terpene standard, >99.0% (GC, sum of enantiomers)
1 ml, 5 ml
63682
(1R)-(-)-Menthyl chloride
16052-42-9
puriss. p.a. terpene standard, >99.0% (GC, sum of enantiomers)
1 ml, 10 ml
70154
(-)-cis-Myrtanol
51152-12-6
puriss. p.a. terpene standard, ~99% (GC, sum of enantiomers)
1 ml, 10 ml
70117
(+)-trans-Myrtanol
132203-71-5
puriss. p.a. terpene standard, ~99% (GC, sum of enantiomers)
1 ml, 5 ml
46004
(-)-trans-Myrtanyl acetate
90934-53-5
puriss. p.a. terpene standard, >99.0% (GC, sum of enantiomers)
1 ml, 5 ml
70119
(-)-Myrtenal
564-94-3
puriss. p.a. terpene standard, >99.0% (GC, sum of enantiomers)
1 ml, 5 ml
70158
(-)-Myrtenol
19894-97-4
puriss. p.a. terpene standard, ~99% (GC, sum of enantiomers)
1 ml, 5 ml
72066
Neoabietic acid, standard
471-77-2
puriss. p.a. standard, ~99% (GC)
50 mg
72134
(+)-Neomenthol
2216-52-6
puriss. p.a. terpene standard, >99.0% (GC, sum of enantiomers)
1 ml, 5 ml
72139
(-)-Neomenthol
20747-49-3
puriss. p.a. terpene standard, >99.0% (GC, sum of enantiomers)
1 ml, 5 ml
72135
(+/-)-Neomenthol
3623-51-6
puriss. p.a. terpene standard, >99.5% (GC)
5 g
46008
(+)-(1S,2S,5R)-Neomenthyl acetate
2552-91-2
puriss. p.a. terpene standard, >99.0% (GC, sum of enantiomers)
1 g, 5 g
46007
(-)-(1R,2R,5S)-Neomenthyl acetate
146502-80-9
puriss. p.a. terpene standard, >99.0% (GC, sum of enantiomers)
250 mg, 1 g, 5 g
46015
Neryl acetate
141-12-8
puriss. p.a. terpene standard, >99.0% (GC)
1 ml
77303
(+)-Perillyl alcohol
57717-97-2
puriss. p.a. terpene standard, >99.0% (GC, sum of enantiomers)
1 ml, 5 ml
80593
(1R)-(+)-cis-Pinane
4795-86-2
puriss. p.a. terpene standard, >99.0% (GC, sum of enantiomers )
1 ml, 5 ml
80595
(1S)-(-)-trans-Pinane
10281-53-5
puriss. p.a. terpene standard, ~99% (GC, sum of enantiomers)
1 ml, 5 ml
80598
(1R)-(-)-trans-Pinane-1,10-diol
53312-42-8
puriss. p.a. terpene standard; >99.0% (GC, sum of enantiomers )
250 mg, 1 g
80602
cis-Pinan-2-ol
4948-28-15
puriss. p.a. standard, >99.0% (GC)
1 g, 5 g
80605
(+)-alpha-Pinene
7785-70-8
puriss. p.a. terpene standard, >99.5% (GC, sum of enantiomers)
1 ml, 5 ml
80599
(-)-alpha-Pinene
7785-26-4
puriss. p.a. terpene standard, >99.5% (GC, sum of enantiomers)
1 ml, 5 ml
80606
(-)-alpha-Pinene
7785-26-4
puriss. p.a. terpene standard; >99.0% (GC, sum of enantiomers)
5 ml, 25 ml, 100 ml
80607
(+)-beta-Pinene
19902-08-0
puriss. p.a. terpene standard, ~99% (GC, sum of enantiomers)
1 ml, 5 ml
80609
(-)-beta-Pinene
18172-67-3
puriss. p.a. terpene standard, >99.0% (GC, sum of enantiomers)
1 ml, 5 ml
82569
(+)-Pulegone
89-82-7
puriss. p.a. terpene standard, ~99% (GC, sum of enantiomers)
1 ml, 5 ml, 25 ml
83915
(+)-Rose oxide
16409-43-1
puriss. p.a. terpene standard; >99.0% (GC) mixture of isomers (cis+trans)
1 ml, 10 ml
83917
(-)-Rose oxide
16409-43-1
puriss. p.a. terpene standard, >99.0% (GC, mixture of isomers (cis+trans))
1 ml, 10 ml
84085
(+)-Sabinene
2009-00-9
puriss. p.a. terpene standard, ~99% (GC, sum of enantiomers)
1 ml
84500
(+)-Santolina alcohol
35671-15-9
puriss. p.a. terpene standard, ~99% (GC, sum of enantiomers)
1 ml
09828
trans-Terpin
565-50-4
puriss. p.a. terpene standard, >99.0% (GC)
1 g
86476
gamma-Terpinene
99-85-4
puriss. p.a. terpene standard, ~99% (GC)
1 ml, 5 ml
86477
(+)-Terpinen-4-ol
2438-10-0
puriss. p.a. terpene standard, >99.0% (GC, sum of enantiomers)
1 ml, 5 ml
86482
(-)-alpha-Terpineol
10482-56-1
puriss. p.a. terpene standard, ~99% (GC, sum of enantiomers)
1 ml, 5 ml
94882
(-)-Verbenone
80-57-9
puriss. p.a. terpene standard, >99.0% (GC, sum of enantiomers)
1 ml, 5 ml, 25 ml
Back to Top
Back to GC Home