Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Liquid Chromatography
GPC GFC Standards
Fluka offers a broad range of GPC standards for various applications, e.g.:
  • calibration of GPC/SEC columns using the polymer your are investigating
  • investigation of polymer degradation and its mechanism
  • determination of physical properties of monodisperse polymers as a function of molecular weight and/or tacticity
Our GPC standards are provided with a certificate of analysis stating all relevant measuring parameters like the molecular weight values Mn, Mw and the polydispersity D.
Polystyrenes, Polyethylene oxides and Dextranes are also available as certified polymers according to DIN.
GPC working Standards:
Product Range
Cat.No.
Dextranes
Pack Size
00268
Dextran Standard 1'000 certified acc. to DIN
100 mg, 500 mg
00269
Dextran Standard 5'000 certified acc. to DIN
100 mg, 500 mg
00270
Dextran Standard 12'000 certified acc. to DIN
100 mg, 500 mg
00271
Dextran Standard 25'000 certified acc. to DIN
100 mg, 500 mg
00891
Dextran Standard 50'000 certified acc. to DIN
100 mg, 500 mg
00892
Dextran Standard 80'000 certified acc. to DIN
100 mg, 500 mg
00893
Dextran Standard 150'000 certified acc. to DIN
100 mg, 500 mg
00894
Dextran Standard 270'000 certified acc. to DIN
100 mg, 500 mg
00895
Dextran Standard 410'000 certified acc. to DIN
100 mg, 500 mg
00896
Dextran Standard 670'000 certified acc. to DIN
100 mg, 500 mg
31416
Dextran Standard 1'000 from Leuconostoc mesenteroides
100 mg
31417
Dextran Standard 5'000 from Leuconostoc mesenteroides
100 mg
31418
Dextran Standard 12'000 from Leuconostoc mesenteroides
100 mg
31419
Dextran Standard 25'000 from Leuconostoc mesenteroides
100 mg
31420
Dextran Standard 50'000 from Leuconostoc mesenteroides
100 mg
31421
Dextran Standard 80'000 from Leuconostoc mesenteroides
100 mg
31422
Dextran Standard 150'000 from Leuconostoc mesenteroides
100 mg
31423
Dextran Standard 270'000 from Leuconostoc mesenteroides
100 mg
31424
Dextran Standard 410'000 from Leuconostoc mesenteroides
100 mg
31425
Dextran Standard 670'000 from Leuconostoc mesenteroides
100 mg
31426
Dextran Standard 750'000 from Leuconostoc mesenteroides
100 mg
31427
Dextran Standard 1'800'000 from Leuconostoc mesenteroides
100 mg
31428
Dextran Standard 4'900'000
100 mg
31430
Dextran Standard Set M(p) 1'000-400'000, 10x500 mg
Set with 10 samples
Cat.No.
Polyacrylic Acids
Pack Size
81116
Polyacrylic acid Sodium salt Standard 995
250 mg
81117
Polyacrylic acid Sodium salt Standard 1'250
250 mg
81118
Polyacrylic acid Sodium salt Standard 1'770
250 mg
81119
Polyacrylic acid Sodium salt Standard 2'925
250 mg
81120
Polyacrylic acid Sodium salt Standard 4'100
250 mg
81121
Polyacrylic acid Sodium salt Standard 4'500
250 mg
81122
Polyacrylic acid Sodium salt Standard 7'500
250 mg
81123
Polyacrylic acid Sodium salt Standard 16'000
250 mg
81124
Polyacrylic acid Sodium salt Standard 28'000
250 mg
81126
Polyacrylic acid Sodium salt Standard 35'000
250 mg
81129
Polyacrylic acid Sodium salt Standard 62'900
250 mg
81131
Polyacrylic acid Sodium salt Standard 90'500
250 mg
81133
Polyacrylic acid Sodium salt Standard 130'500
250 mg
81135
Polyacrylic acid Sodium salt Standard 193'800
250 mg
81137
Polyacrylic acid Sodium salt Standard 272'900
250 mg
81139
Polyacrylic acid Sodium salt Standard 333'000
250 mg
81141
Polyacrylic acid Sodium salt Standard 467'000
250 mg
81143
Polyacrylic acid Sodium salt Standard 585'400
250 mg
81145
Polyacrylic acid Sodium salt Standard 782'200
250 mg
81147
Polyacrylic acid Sodium salt Standard 1'100'000
250 mg
Cat.No.
Polybutadienes
Pack Size
81261
1,4-Polybutadiene Standard 640, stab.
500 mg
81262
1,4-Polybutadiene Standard 2'000, stab.
500 mg
81263
1,4-Polybutadiene Standard 5'000, stab.
500 mg
81264
1,4-Polybutadiene Standard 14'000, stab.
500 mg
81254
1,4-Polybutadiene Standard 20'000, stab.
500 mg
81256
1,4-Polybutadiene Standard 30'000, stab.
500 mg
81265
1,4-Polybutadiene Standard 36'000, stab.
500 mg
81266
1,4-Polybutadiene Standard 66'000, stab.
500 mg
81267
1,4-Polybutadiene Standard 160'000,stab.
500 mg
81257
1,4-Polybutadiene Standard 200'000,stab.
500 mg
81258
1,4-Polybutadiene Standard 300'000,stab.
500 mg
81259
1,4-Polybutadiene Standard 800'000,stab.
500 mg
Cat.No.
Poly(tert-butyl methacrylates)
Pack Size
81469
Poly(tert-butyl methacrylate) Standard 2'500
250 mg
81476
Poly(tert-butyl methacrylate) Standard 5'000
250 mg
81477
Poly(tert-butyl methacrylate) Standard 10'000
250 mg
81478
Poly(tert-butyl methacrylate) Standard 20'000
250 mg
81479
Poly(tert-butyl methacrylate) Standard 30'000
250 mg
81481
Poly(tert-butyl methacrylate) Standard 70'000
250 mg
81482
Poly(tert-butyl methacrylate) Standard 100'000
250 mg
81483
Poly(tert-butyl methacrylate) Standard 200'000
250 mg
81484
Poly(tert-butyl methacrylate) Standard 300'000
250 mg
81485
Poly(tert-butyl methacrylate) Standard 500'000
250 mg
81486
Poly(tert-butyl methacrylate) Standard 700'000
250 mg
Cat.No.
Poly(butyl methacrylates)
Pack Size
81464
Poly(butyl methacrylate) Standard 2'000
250 mg
81456
Poly(butyl methacrylate) Standard 4'000
250 mg
81457
Poly(butyl methacrylate) Standard 10'000
250 mg
81458
Poly(butyl methacrylate) Standard 20'000
250 mg
81459
Poly(butyl methacrylate) Standard 40'000
250 mg
81471
Poly(butyl methacrylate) Standard 90'000
250 mg
81472
Poly(butyl methacrylate) Standard 200'000
250 mg
81473
Poly(butyl methacrylate) Standard 300'000
250 mg
81474
Poly(butyl methacrylate) Standard 500'000
250 mg
81475
Poly(butyl methacrylate) Standard 800'000
250 mg
Cat.No.
Polycarbonates
Pack Size
02453
Polycarbonate Standard 326
100 mg
02454
Polycarbonate Standard 1'590
100 mg
02455
Polycarbonate Standard 3'860
100 mg
02456
Polycarbonate Standard 6'800
100 mg
02461
Polycarbonate Standard 10'400
100 mg
02458
Polycarbonate Standard 20'200
100 mg
02459
Polycarbonate Standard 30'600
100 mg
02466
Polycarbonate Standard 50'000
100 mg
02467
Polycarbonate Standard 127'000
100 mg
02468
Polycarbonate Standard 195'000
100 mg
Cat.No.
Polydimethyl Siloxanes
Pack Size
81154
Polydimethylsiloxane 1'850
100 mg
81155
Polydimethylsiloxane 3'320
100 mg
81156
Polydimethylsiloxane 4'400
100 mg
81158
Polydimethylsiloxane 16'000
100 mg
81159
Polydimethylsiloxane 25'800
100 mg
81161
Polydimethylsiloxane 35'900
100 mg
81163
Polydimethylsiloxane 47'500
100 mg
81164
Polydimethylsiloxane 53'500
100 mg
81165
Polydimethylsiloxane 71'000
100 mg
81166
Polydimethylsiloxane 89'800
100 mg
81167
Polydimethylsiloxane 158'000
100 mg
81168
Polydimethylsiloxane 173'000
100 mg
81169
Polydimethylsiloxane 197'000
100 mg
81171
Polydimethylsiloxane 303'000
100 mg
81157
Polydimethylsiloxane 7'100
100 mg
Cat.No.
Polyethylenes
Pack Size
81219
Polyethylene Standard 1'000
100 mg
81223
Polyethylene Standard 2'000
100 mg
81224
Polyethylene Standard 17'000
100 mg
81226
Polyethylene Standard 21'000
100 mg
81229
Polyethylene Standard 27'000
100 mg
81232
Polyethylene Standard 30'000
100 mg
81234
Polyethylene Standard 37'000
100 mg
81236
Polyethylene Standard 60'000
100 mg
81238
Polyethylene Standard 77'000
100 mg
81241
Polyethylene Standard 85'000
100 mg
81244
Polyethylene Standard 92'000
100 mg
81246
Polyethylene Standard 116'000
100 mg
81248
Polyethylene Standard 153'000
100 mg
81250
Polyethylene Standard 169'000
100 mg
81252
Polyethylene Standard 183'000
100 mg
Cat.No.
Polyethylene Glycols
Pack Size
81282
Polyethylene glycol Standard 200
1 g
81283
Polyethylene glycol Standard 300
1 g
81284
Polyethylene glycol Standard 400
1 g
81286
Polyethylene glycol Standard 600
1 g
81288
Polyethylene glycol Standard 1'000
1 g
81289
Polyethylene glycol Standard 1'500
1 g
81290
Polyethylene glycol Standard 2'000
1 g
81291
Polyethylene glycol Standard 3'000
1 g
81292
Polyethylene glycol Standard 4'000
1 g
81293
Polyethylene glycol Standard 6'000
1 g
81294
Polyethylene glycol Standard 8'000
1 g
81296
Polyethylene glycol Standard 12'000
1 g
81297
Polyethylene glycol Standard 17'500
1 g
81298
Polyethylene glycol Standard 20'000
1 g
81299
Polyethylene glycol Standard 35'000
1 g
03461
Polyethylene glycol Standard 330 certified acc. to DIN
1 g
03462
Polyethylene glycol Standard 420 certi-fied acc. to DIN
1 g
03463
Polyethylene glycol Standard 600 certi-fied acc. to DIN
1 g
03464
Polyethylene glycol Standard 1'000 certified acc. to DIN
1 g
03465
Polyethylene glycol Standard 2'000 certified acc. to DIN
1 g
03466
Polyethylene glycol Standard 3'100 certified acc. to DIN
1 g
03467
Polyethylene glycol Standard 4'400 certified acc. to DIN
1 g
03468
Polyethylene glycol Standard 6'200 certified acc. to DIN
1 g
03469
Polyethylene glycol Standard 8'600 certified acc. to DIN
1 g
03471
Polyethylene glycol Standard 12'000 cer-tified acc. to DIN
1 g
03472
Polyethylene glycol Standard 23'000 cer-tified acc. to DIN
1 g
03473
Polyethylene glycol Standard 40'000 cer-tified acc. to DIN
1 g
81396
Polyethylene glycol Standard Set M(p)400-40'000
Set with 10 samples
02393
PEG/PEO Standard ReadyCal Set Mp 500-2'740'000
Set with 10 samples
Cat.No.
Polyethylene Oxides
Pack Size
81200
Polyethylene oxide Standard 55'000
500 mg
81201
Polyethylene oxide Standard 108'000
500 mg
81202
Polyethylene oxide Standard 116'000
500 mg
81203
Polyethylene oxide Standard 132'000
500 mg
81204
Polyethylene oxide Standard 182'000
500 mg
81206
Polyethylene oxide Standard 218'000
500 mg
81207
Polyethylene oxide Standard 237'000
500 mg
81208
Polyethylene oxide Standard 246'000
500 mg
81209
Polyethylene oxide Standard 288'000
500 mg
81211
Polyethylene oxide Standard 438'000
500 mg
81212
Polyethylene oxide Standard 463'000
500 mg
81213
Polyethylene oxide Standard 511'000
500 mg
81205
Polyethylene oxide Standard 205'000
500 mg
Cat.No.
Poly(ethyl methacrylates)
Pack Size
81585
Poly(ethyl methacrylate) Standard1'800
100 mg
81587
Poly(ethyl methacrylate) Standard4'700
100 mg
81589
Poly(ethyl methacrylate) Standard7'500
100 mg
81591
Poly(ethyl methacrylate) Standard45'000
100 mg
81593
Poly(ethyl methacrylate) Standard142'000
100 mg
81595
Poly(ethyl methacrylate) Standard165'000
100 mg
81597
Poly(ethyl methacrylate) Standard275'000
100 mg
81599
Poly(ethyl methacrylate) Standard300'000
100 mg
81601
Poly(ethyl methacrylate) Standard345'000
100 mg
81603
Poly(ethyl methacrylate) Standard500'000
100 mg
Cat.No.
Poly(isobutylenes)
Pack Size
81564
Poly(isobutylene) Standard 390,
250 mg
81565
Poly(isobutylene) Standard 680,
250 mg
81566
Poly(isobutylene) Standard 2'500
250 mg
81567
Poly(isobutylene) Standard 4'400
250 mg
81568
Poly(isobutylene) Standard 10'000
250 mg
81569
Poly(isobutylene) Standard 24'000
250 mg
81571
Poly(isobutylene) Standard 34'000
250 mg
81573
Poly(isobutylene) Standard 43'000
250 mg
81574
Poly(isobutylene) Standard 73'000
250 mg
81576
Poly(isobutylene) Standard 86'000
250 mg
81577
Poly(isobutylene) Standard 134'000
250 mg
81579
Poly(isobutylene) Standard 302'000
250 mg
81581
Poly(isobutylene) Standard 583'000
250 mg
81583
Poly(isobutylene) Standard 856'000
250 mg
Cat.No.
1,4-Polyisoprenes
Pack Size
81441
1,4-Polyisoprene Standard 700, stab.
500 mg
81442
1,4-Polyisoprene Standard 2'000, stab.
500 mg
81443
1,4-Polyisoprene Standard 5'000, stab.
500 mg
81444
1,4-Polyisoprene Standard 8'000, stab.
500 mg
81445
1,4-Polyisoprene Standard 10'000, stab.
500 mg
81446
1,4-Polyisoprene Standard 20'000, stab.
500 mg
81447
1,4-Polyisoprene Standard 30'000, stab.
250 mg
81448
1,4-Polyisoprene Standard 50'000, stab.
500 mg
81449
1,4-Polyisoprene Standard 70'000 , stab.
500 mg
81451
1,4-Polyisoprene Standard 100'000,stab.
500 mg
81452
1,4-Polyisoprene Standard 150'000, stab.
500 mg
81453
1,4-Polyisoprene Standard 200'000, stab.
500 mg
81454
1,4-Polyisoprene Standard 300'000, stab.
500 mg
81461
1,4-Polyisoprene Standard 400'000, stab.
250 mg
81463
1,4-Polyisoprene Standard 600'000, stab.
250 mg
81455
1,4-Polyisoprene Standard 800'000, stab.
500 mg
Cat.No.
Poly(methacrylic acid Sodium salts)
Pack Size
02351
Poly(methacrylic acid Sodium salt) Standard 1'270
250 mg
02353
Poly(methacrylic acid Sodium salt) Standard 4'030
250 mg
02354
Poly(methacrylic acid Sodium salt) Standard 5'880
250 mg
02355
Poly(methacrylic acid Sodium salt) Standard 7'750
250 mg
02356
Poly(methacrylic acid Sodium salt) Standard 15'200
250 mg
02357
Poly(methacrylic acid Sodium salt) Standard 31'100
250 mg
02358
Poly(methacrylic acid Sodium salt) Standard 54'400
250 mg
02359
Poly(methacrylic acid Sodium salt) Standard 75'100
250 mg
02360
Poly(methacrylic acid Sodium salt) Standard 129'000
250 mg
02361
Poly(methacrylic acid Sodium salt) Standard 236'000
250 mg
02363
Poly(methacrylic acid Sodium salt) Standard 483'000
250 mg
00971
Poly(methacrylic acid Sodium salt) Standard 790'000
250 mg
Cat.No.
Poly(methyl methacrylates)
Pack Size
81488
Poly(methyl methacrylate) Standard 600
500 mg
81489
Poly(methyl methacrylate) Standard 2'000
500 mg
81496
Poly(methyl methacrylate) Standard 5'000
500 mg
81512
Poly(methyl methacrylate) Standard 8'000
500 mg
81497
Poly(methyl methacrylate) Standard 10'000
500 mg
81498
Poly(methyl methacrylate) Standard 20'000
500 mg
81499
Poly(methyl methacrylate) Standard 30'000
500 mg
81501
Poly(methyl methacrylate) Standard 50'000
500 mg
81502
Poly(methyl methacrylate) Standard 75'000
500 mg
81503
Poly(methyl methacrylate) Standard 100'000
500 mg
81504
Poly(methyl methacrylate) Standard 150'000
500 mg
81508
Poly(methyl methacrylate) Standard 200'000
500 mg
81509
Poly(methyl methacrylate) Standard 300'000
500 mg
81510
Poly(methyl methacrylate) Standard 500'000
250 mg
81511
Poly(methyl methacrylate) Standard 800'000
250 mg
03558
Poly(methyl methacrylate) Standard 2'480'000
250 mg
81506
Poly(methyl methacrylate) StandardReadyCal Set
Set with 12 samples
Cat.No.
Poly(alpha-methylstyrenes)
Pack Size
81513
Poly(alpha-methylstyrene) Standard 2'500
250 mg
81514
Poly(alpha-methylstyrene) Standard 7'000
250 mg
81516
Poly(alpha-methylstyrene) Standard 10'000
250 mg
81517
Poly(alpha-methylstyrene) Standard 14'000
250 mg
81518
Poly(alpha-methylstyrene) Standard 20'000
250 mg
81519
Poly(alpha-methylstyrene) Standard 50'000
250 mg
81520
Poly(alpha-methylstyrene) Standard 100'000
250 mg
81522
Poly(alpha-methylstyrene) Standard 160'000
250 mg
81523
Poly(alpha-methylstyrene) Standard 300'000
250 mg
81524
Poly(alpha-methylstyrene) Standard 700'000
250 mg
Cat.No.
Propylenglycols
Pack Size
81526
Polypropylene glycol Standard 460
500 mg
81527
Polypropylene glycol Standard 810
500 mg
81528
Polypropylene glycol Standard 1'150
500 mg
81530
Polypropylene glycol Standard 2'150
500 mg
81531
Polypropylene glycol Standard 3'250
500 mg
81533
Polypropylene glycol Standard 5'300
500 mg
Cat.No.
Polystyrenes
Pack Size
81401
Polystyrene Standard 500
250 mg, 1g
81402
Polystyrene Standard 1'000
250 mg, 1g
81403
Polystyrene Standard 2'000
250 mg, 1g
81404
Polystyrene Standard 3'000
250 mg, 1g
81405
Polystyrene Standard 5'000
250 mg, 1g
81406
Polystyrene Standard 10'000
250 mg, 1g
81407
Polystyrene Standard 20'000
250 mg, 1g
81408
Polystyrene Standard 30'000
250 mg, 1g
81409
Polystyrene Standard 70'000
250 mg, 1g
81410
Polystyrene Standard 100'000
250 mg, 1g
81411
Polystyrene Standard 150'000
250 mg, 1g
81412
Polystyrene Standard 200'000
250 mg, 1g
81413
Polystyrene Standard 300'000
250 mg, 1g
81414
Polystyrene Standard 500'000
250 mg, 1g
81415
Polystyrene Standard 700'000
250 mg, 1g
81416
Polystyrene Standard 1'000'000
250 mg, 1g
81417
Polystyrene Standard 2'000'000
250 mg, 1g
81418
Polystyrene Standard 4'000'000
250 mg, 1g
81419
Polystyrene Standard 8'000'000
250 mg, 1g
00926
Polystyrene Standard 400 certified acc. to DIN
1g
00927
Polystyrene Standard 2'000 certified acc. to DIN
1g
00928
Polystyrene Standard 3'000 certified acc. to DIN
1g
00945
Polystyrene Standard 10'000 certified acc. to DIN
1g
00946
Polystyrene Standard 20'000 certified acc. to DIN
1g
00947
Polystyrene Standard 33'000 certified acc. to DIN
1g
00948
Polystyrene Standard 70'000 certified acc. to DIN
1g
00949
Polystyrene Standard 130'000 certified acc. to DIN
1g
00951
Polystyrene Standard 250'000 certified acc. to DIN
1g
00952
Polystyrene Standard 650'000 certified acc. to DIN
1g
00953
Polystyrene Standard 1'000'000 certi-fied acc. to DIN
1g
00954
Polystyrene Standard 2'200'000 certi-fied acc. to DIN
1g
81434
Polystyrene Standard ReadyCal Set M(p)400-2'000'000
Set with 12 samples
81421
Polystyrene Standard Set M(p) 400-2'000'000
Set with 12 samples
Cat.No.
Poly(styrene sulfonic acid Sodium salts)
Pack Size
81604
Poly(styrenesulfonic acid Sodium salt) Standard 210
250 mg
81605
Poly(styrenesulfonic acid Sodium salt) Standard 1'400
250 mg
81606
Poly(styrenesulfonic acid Sodium salt) Standard 4'300
250 mg
81607
Poly(styrenesulfonic acid Sodium salt) Standard 6'800
250 mg
81608
Poly(styrenesulfonic acid Sodium salt) Standard 13'000
250 mg
81609
Poly(styrenesulfonic acid Sodium salt) Standard 17'000
250 mg
81610
Poly(styrenesulfonic acid Sodium salt) Standard 32'000
250 mg
81611
Poly(styrenesulfonic acid Sodium salt) Standard 49'000
250 mg
81612
Poly(styrenesulfonic acid Sodium salt) Standard 77'000
250 mg
81614
Poly(styrenesulfonic acid Sodium salt) Standard 150'000
250 mg
81615
Poly(styrenesulfonic acid Sodium salt) Standard 350'000
250 mg
81616
Poly(styrenesulfonic acid Sodium salt) Standard 990'000
250 mg
81617
Poly(styrenesulfonic acid Sodium salt) Standard 2'600'000
250 mg, 1g
Cat.No.
Polyvinyl Alcohols
Pack Size
81368
Polyvinyl alcohol Standard 7'200
250 mg, 1g
81369
Polyvinyl alcohol Standard 35'000
250 mg, 1g
81372
Polyvinyl alcohol Standard 55'000
250 mg, 1g
81373
Polyvinyl alcohol Standard 82'000
250 mg, 1g
81374
Polyvinyl alcohol Standard 122'400
250 mg, 1g
81371
Polyvinyl alcohol Standard 137'100
250 mg, 1g
81375
Polyvinyl alcohol Standard 170'500
250 mg, 1g
81376
Polyvinyl alcohol Standard 200'000
250 mg, 1g
Cat.No.
Poly(vinyl chlorides)
Pack Size
81424
Poly(vinyl chloride) Standard 10'000
100 mg
81425
Poly(vinyl chloride) Standard 36'000
100 mg
81427
Poly(vinyl chloride) Standard 75'000
100 mg
81428
Poly(vinyl chloride) Standard 100'000
100 mg
81429
Poly(vinyl chloride) Standard 130'000
100 mg
81431
Poly(vinyl chloride) Standard 200'000
100 mg
Cat.No.
Poly(2-vinylpyridines)
Pack Size
81620
Poly(2-vinylpyridine) Standard 1'500
100 mg
81622
Poly(2-vinylpyridine) Standard 2'900
100 mg
81624
Poly(2-vinylpyridine) Standard 6'500
100 mg
81626
Poly(2-vinylpyridine) Standard 12'000
100 mg
81628
Poly(2-vinylpyridine) Standard 21'000
100 mg
81631
Poly(2-vinylpyridine) Standard 47'000
100 mg
81633
Poly(2-vinylpyridine) Standard 64'000
100 mg
81635
Poly(2-vinylpyridine) Standard 79'000
100 mg
81637
Poly(2-vinylpyridine) Standard 115'000
100 mg
81639
Poly(2-vinylpyridine) Standard 135'000
100 mg
81641
Poly(2-vinylpyridine) Standard 287'000
100 mg
81643
Poly(2-vinylpyridine) Standard 1'260'000
100 mg
Cat.No.
Pullulanes
Pack Size
82439
Pullulan Standard Set
set with 8 pullulan standards (200 mg each)
Back to Top
Back to LC Home