Sigma-Aldrich
Analytical / Chromatography > Analytical Reagents > Chromatography > Liquid Chromatography > HPLC Buffers
Analytical Reagents
HPLC Buffers
Fluka is expanding its HPLC auxiliaries and reagents portfolio steadily. Currently we offer more than 30 application tested reagents. For most of the products the following quality controll tests are performed:
  • RP gradient: to test correspondance with standard
  • Solubility: to test whether the solution is clear
  • Residue: to test whether there is no residue in the filter (millipore® 0.45 mm membrane filter)
  • UV Absorption: to test correspondance of the maximum UV absorption at a HPLC relevant wavelength with standard
  • pH: to test whether the pH of the aqueous solution corresponds
  • Cyclic Voltammogram: to test whether it corresponds
The table below lists the recently launched HPLC grade products:
Catalog No.
Product
Description
Package Size
Add to Cart
17836
Ammonium acetate puriss. p.a. for HPLC; >99.0% (NT) 50 g/250 g
17843
Ammonium formate puriss. p.a. for HPLC, >99.0% (NT) (dried material) 50 g/250 g
17837
Ammonium hydroxide solution puriss. p.a. for HPLC; ~10% (T) in water 100 ml/1 l
17842
Ammonium phosphate monobasic puriss. p.a. for HPLC; >99.0% (T) 50 g/250 g
17839
Ammonium trifluoroacetate puriss. p.a. for HPLC; >99.0% (NT) 10 g/50 g
09676
Formic acid, 50% puriss. p.a. for HPLC; 49-51% (T) 100 ml/500 ml
09751
Formic acid:Triethylamine 2M:1M concentrate puriss. p.a. for HPLC 100 ml/500 ml
09752
Formic acid:Triethylamine 2M:2M concentrate puriss. p.a. for HPLC 100 ml/500 ml
17835
Potassium phosphate dibasic anhydrous puriss. p.a. for HPLC; >99.0% (T) 50 g/250 g
17841
Sodium formate puriss. p.a. for HPLC; >99.0% (NT) 50 g/250 g
71633
Sodium phospate dibasic dihydrate puriss. p.a. for HPLC; ~99% (T) 50 g/250 g
17844
Sodium phosphate monobasic anhydrous puriss. p.a. for HPLC; >99.0% (T) 50 g/250 g
17840
Sodium trifluoroacetate puriss. p.a. for HPLC; >99.0% (T) 10 g/50 g
17924
Triethylamine puriss. p.a. for HPLC; >99.5% (GC) 10x2 ml Amp.
09746
Trifluoroacetic acid:Triethylamine 2M:1M puriss. p.a. for HPLC 100 ml/500 ml
09747
Trifluoroacetic acid:Triethylamine 2M:2M puriss. p.a. for HPLC 100 ml/500 ml
Back to top
Back to Analytical Reagents