|
Liquid Chromatography
HPLC Derivatizing Agents
|
|
|
Derivatizing of HPLC samples has become vitally important in the pharmaceutical, chemical and agricultural industries. Derivatizing of reactive analytes for HPLC shows following advantages:
- increase in analyte stability
- higher selectivity and sensitivity caused by reduced polarity that leads to better chromatographic separation
Fluka offers a selection of the most useful chiral and non-chiral derivatizing reagents for all your HPLC applications. A unique Fluka quality set of specifically selected reagents, carefully produced and rigorously analyzed in our laboratories.
Chiral derivatizing agents offers an opportunity for the separation of optical isomers. Our chiral HPLC reagents exhibits an enantiomeric ratio of at least >99.5 : 0.5 . As a rule, the actual value is distinctly higher in most cases.
Non-chiral derivatisating agents are a accurately tested by 1H-NMR, IR- and NIR-Spectroscopy.
Product Range
| Catalog No. |
Product Name |
Package Size |
| 15411 |
BOC-L-cysteine |
250 mg, 1 g |
| 21286 |
(+)-Camphanic acid chloride |
250 mg, 1 g |
| 21287 |
(-)-Camphanic acid chloride |
5 g, 25 g |
| 21360 |
(+)-Camphor-10-sulfonic acid |
100 g, 500 g |
| 21365 |
(-)-Camphor-10-sulfonic acid |
50 g, 250 g |
| 21380 |
(+)-Camphor-10-sulfonyl chloride |
10 g, 50 g |
| 21382 |
(-)-Camphor-10-sulfonyl chloride |
5 g, 25 g |
| 17925 |
(9-Carbazolyl) acetic acid |
100 mg, 500 mg |
| 17926 |
3-(9-Carbazolyl) propionic acid |
100 mg, 500 mg |
| 29285 |
R(-)-1-Cyclohexylethylamine |
10 ml, 50 ml |
| 29287 |
S(+)-1-Cyclohexylethylamine |
10 ml, 50 ml |
| 31587 |
(+)-Di-O-acetyl-L-tartaric anhydride |
50 g, 250 g |
| 17927 |
(9,10-Dihydro-9-oxo-10-acridinyl) acetic acid |
50 mg, 250 mg |
| 40764 |
(R)-2,8-Dimethyl-5,11-methanodibenzo(b,f)(1,5) diazocin |
100 mg |
| 40765 |
(S)-2,8-Dimethyl-5,11-methanodibenzo(b,f)(1,5) diazocin |
100 mg |
| 42033 |
N-(3,5-Dinitrobenzyl)-L-alpha-phenyl-glycine |
1 g, 5 g |
| 42032 |
N-(3,5-Dinitrobenzyl)-D-alpha-phenyl-glycine |
5 g, 25 g |
| 42095 |
Na-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide |
100 mg |
| 42100 |
Na-(2,4-Dinitro-5-fluorophenyl)-D-valinamide |
100 mg, 500 mg |
| 42102 |
Na-(2,4-Dinitro-5-fluorophenyl)-L-valinamide |
100 mg, 500 mg |
| 42210 |
2,4-Dinitrophenylhydrazine |
25 g, 100 g, 500 g |
| 42215 |
2,4-Dinitrophenylhydrazine phosphoric acid solution, ~0.2 M |
250 ml, 1 l |
| 43135 |
2,2-Diphenyl-1-oxa-3-oxonia-2-borata-naphthalene |
1 g, 5 g |
| 23182 |
(+)-FLEC solution, 50 mg in 10 ml acetone |
1 ea |
| 23183 |
(-)-FLEC solution, 50 mg in 10 ml acetone |
1 ea |
| 47734 |
R(-)-a-(Formyloxy)phenylacetyl chloride |
1 ml, 5 ml |
| 58689 |
N-Isobutyryl-D-cysteine |
250 mg, 1 g |
| 58698 |
N-Isobutyryl-L-cysteine |
250 mg |
| 23197 |
(+)-(1S)-Menthyl chloroformate |
5 ml, 25 ml |
| 23200 |
(-)-(1R)-Menthyl chloroformate |
10 ml, 50 ml |
| 77879 |
R(+)-a-Methylbenzyl amine |
5 ml, 25 ml |
| 77869 |
L(-)-a-Methylbenzyl amine |
5 ml, 25 ml |
| 70710 |
R(+)-1-(1-Naphthyl)ethyl amine |
1 ml |
| 70712 |
S(-)-1-(1-Naphthyl)ethyl amine |
1 ml |
| 70725 |
R(-)-1-(1-Naphthyl)ethyl isocyanate stab. |
1 ml |
| 70726 |
S(+)-1-(1-Naphthyl)ethyl isocyanate stab. |
1 ml |
| 92614 |
4-Nitro-7-piperazino-2,1,3-benzoxadiazole; used for Analysis of Isocyanates |
100 mg |
| 74152 |
(+)-Noe's Reagent |
250 mg, 1 g |
| 74153 |
(-)-Noe's Reagent |
250 mg, 1 g |
| 74865 |
R(-)-2-Octanol |
1 ml, 5 ml |
| 74863 |
S(+)-2-Octanol |
1 ml, 5 ml, 25 ml |
| 75946 |
(R)-5-Oxotetrahydrofuran-2-carboxylic acid |
1 g, 5 g |
| 75947 |
(S)-5-Oxotetrahydrofuran-2-carboxylic acid |
1 g, 5 g |
| 76744 |
R(+)-1-(Pentafluorophenyl) ethanol |
250 mg, 1 g |
| 76746 |
S(-)-1-(Pentafluorophenyl) ethanol |
250 mg, 1 g |
| 78241 |
R(-)-2-Phenylbutyric acid |
1 ml, 5 ml |
| 78244 |
S(+)-2-Phenylbutyric acid |
1 ml, 5 ml |
| 77848 |
R(+)-1-Phenylethanol |
1 ml |
| 77849 |
S(-)-1-Phenylethanol |
1 ml, 5 ml |
| 77968 |
R(+)-1-Phenylethyl isocyanate stab. |
1 ml, 5 ml |
| 77970 |
S(-)-1-Phenylethyl isocyanate stab. |
1 ml, 5 ml |
| 78922 |
R(+)-1-Phenyl-1-propanol |
1 ml, 5 ml |
| 78923 |
S(-)-1-Phenyl-1-propanol |
1 ml, 5 ml |
| 82669 |
1-Pyrene butyric hydrazide |
100 mg, 500 mg |
| 86550 |
2,3,4,6-Tetra-O-acetyl-b-D-glucopyranosyl isothiocyanate |
100 mg, 1 g |
| 86729 |
2,3,4,6-Tetra-O-benzoyl-b-D-glucopyranosyl isothiocyanate |
100 mg, 500 mg |
| 88102 |
2,3,4,6-Tetra-O-pivaloyl-b-D-galactopyranosyl isothiocyanate |
100 mg, 500 mg |
| 90245 |
2,3,4-Tri-O-acetyl-a-D-arabinopyranosyl isothiocyanate |
100 mg, 500 mg |
| 93509 |
(R)-TroloxTM methyl ether |
50 mg |
| 93510 |
(S)-TroloxTM methyl ether |
50 mg |
Back to Top
Back to LC Home |
| |
| |
| |
| |
|