Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
LC Solvents
Liquid Chromatography Solvents
Fluka Solvents for HPLC
HPLC CHROMASOLV® - Solvents for HPLC Analysis
LC-MS CHROMASOLV®
- Solvents for LC-MS Analysis
LC-NMR CHROMASOLV®
- Solvents for LC-NMR Analysis
Fluka Solvents for HPLC
In liquid chromatography the quality and reproducibility of the separation depends not only on the properties of the stationary phase, but decisively on the quality of the solvents used.
Fluka has been optimized all HPLC solvents for liquid chromatography applications. HPLC solvents are lot-tested and precisely controlled to meet the high quality requirements of successful HPLC separations:
  • Low UV-absorbance, assuring optimum sensitivity
  • Guaranteed suitability for fluorescence detection
  • Controlled low content of non-volatile components like water, free acid and free alkali, assuring long pump-seal life
  • Low particulates, pre-filtration is not necessary
  • High assay, assuring high purity
Gradient grade HPLC solvents are specified with a gradient elution test to illustrate the interference-free baselines. Both UV absorbance and fluorescence signals are monitored.
All HPLC solvents are delivered with an expiration date.
Product Range
Catalog No.
Product
Purity
Package Size
00692
Acetonitrile * for HPLC; >99.8% (GC) 500 ml, 1 l, 2.5 l
00683
Acetonitrile for HPLC; gradient grade+ suitable for PAK-analysis; >99.9% (GC) 1 l, 2.5 l
12549
Benzene for HPLC; >99.5% (GC) (dried material) 1 l, 2.5 l
19417
1-Butanol * for HPLC; >99.8% (GC) 1 l, 2.5 l
20398
2-Butoxyethanol for HPLC - GC; >99.8% (GC) 500 ml, 1 l
20247
tert-Butyl methyl ether * for HPLC; >99.8% (GC) 1 l, 2.5 l
87036
Carbon tetrachloride for HPLC; >99.8% (GC) 1 l
19748
1-Chlorobutane * for HPLC; >99.8% (GC) 1 l
25685
Chloroform * for HPLC; >99.8% (GC); stab. with ~1% ethanol 1 l, 2.5 l
59379
2-Chloropropane for HPLC; >99.5% (GC) 500 ml, 1 l
28931
Cyclohexane * for HPLC; >99.5% (GC) 1 l, 2.5 l
29681
Cyclopentane for HPLC; >75% (GC) 1 l, 2.5 l
03522
1,2-Dichloroethane for HPLC; >99.8% (GC) 500 ml, 1 l
66746
Dichloromethane * for HPLC; >99.8% (GC) 1 l, 2.5 l
31675
Diethyl ether, stab. for HPLC; >99.8% (GC); stab. with 2,6-di-tert-butyl-p-cresol 1 l, 2.5 l
38569
1,2-Dimethoxyethane for HPLC; >99.5% (GC) 500 ml, 1 l
38842
N,N-Dimethylacetamide for HPLC; >99.8% (GC) 1 l, 2.5 l
42505
Dioxane * for HPLC; >99.8% (GC) 500 ml, 1 l, 2.5 l
02854
Ethanol for HPLC; gradient grade >99.8% (v/v) 1 l, 2.5 l
02855
Ethanol for HPLC; >99.8% (v/v) 1 l, 2.5 l
45763
Ethyl acetate * for HPLC; >99.8% (GC) 1 l, 2.5 l
51742
Heptane * for HPLC; >99.5% (GC) 500 ml, 1 l, 2.5 l
52762
Hexane * for HPLC; >95% (GC) 1 l, 2.5 l
59042
Isooctane * for HPLC; >99.5% (GC) 1 l, 2.5 l
65548
Methanol for HPLC; gradient grade >99.8% (GC) 1 l, 2.5 l
65541
Methanol * for HPLC; >99.8% (GC) 1 l, 2.5 l
45999
Methyl acetate for HPLC; >99.8% (GC) 250 ml, 1 l, 2.5 l
73479
Nitromethane for HPLC; >97.0% (GC) 250 ml, 1 l
76875
Pentane * for HPLC; >98.0% (GC) 500 ml, 1 l
82091
1-Propanol * for HPLC; >99.8% (GC) 1 l, 2.5 l
59298
2-Propanol for HPLC; gradient grade >99.8% (GC) 1 l, 2.5 l
59307
2-Propanol * for HPLC; >99.8% (GC) 1 l, 2.5 l
86968
Tetrachloroethylene for HPLC; gradient grade >99.5% (GC) 1 l, 2.5 l
87367
Tetrahydrofuran * for HPLC; >99.8% (GC) 500 ml, 1 l, 2.5 l
89683
Toluene * for HPLC; gradient grade >99.5% (GC) 1 l, 2.5 l
95270
Water for gradient elution 1 l, 5 l, 10 l
95304
Water for HPLC; filtered through a 0.2 mm membrane filter 1 l, 2.5 l
* becomes deleted with replacement, please click here for details
Back to top
Concept of High-Purity Solvents for HPLC
In high performance liquid chromatography the quality and reproducibility of the separation depends not only on the properties of the stationary phase, but decisively on the quality of the solvents used. The solvent line CHROMASOLV® offers guaranteed quality specially tailored to chromatographic requirements, in conjunction with batch consistency. Please find below the different product groups with description and detailed product lists.
G CHROMASOLV® solvents for gradient elution
CHROMASOLV® solvents for HPLC
E CHROMASOLV® solvents for HPLC
The G CHROMASOLV® qualities were developed for sensitive gradient elutions at short wavelengths and with very high requirements with regard to UV-transmittance and fluorescence detection. The CHROMASOLV® solvents are characterized by high UV-transmittance, consistently gradient testing for interfering peaks and base line drift, guaranteed suitability for fluorescence detection, low non-volatile components, free acid and free alkali and an exactly defined low water content. This quality is suitable for measurements in the low wavelength range and has also been tested for suitability for gradient applications and for fluorescence detection.
P CHROMASOLV® solvents for preparative LC
R CHROMASOLV® solvents for routine measurements in LC
The specifications of the
P CHROMASOLV® qualities are tailored fully to the requirements of preparative liquid chromatography. In particular this means low contents of non-volatile impurities and of free alkali.
This quality is available for routine analysis in which chromatography is not carried out with gradient elution at short wavelengths. It is suitable for isocratic analysis, for example at 210 nm or with gradients at 254 nm.
Example Specification
Acetonitrile
G CHROMASOLV® Prod.No. 34998
Acetonitrile CHROMASOLV® Prod.No. 34851
Acetonitrile
E CHROMASOLV® Prod.No. 34888
assay (GC) min. 99.9 % min. 99.9 % min. 99.9 %
non-volatile matter max. 0.0002 % max. 0.0005 % max. 0.0005 %
water (Karl Fischer) max. 0.01 % max. 0.02 % max. 0.02 %
free acid (as CH3COOH) max. 0.001 % max. 0.001 % max. 0.002 %
free alkali (as NH3) max. 0.0001 % max. 0.0002 % max. 0.0002 %
transmittance at 195 nm min. 80 % min. 75 % min. 65 %
transmittance at 200 nm min. 95 % min. 93 % min. 88 %
transmittance at 210 nm - - min. 93 %
transmittance from 228 nm min. 99 % - -
transmittance from 230 nm - min. 99 % -
transmittance from 235 nm - - min. 99 %
HPLC-gradient at 210 nm max. 1 mAU max. 3 mAU -
baseline drift at 210 nm max. 12 mAU max. 15 mAU -
HPLC-gradient at 254 nm max. 0.2 mAU max. 0.5 mAU -
fluorescence at 254 nm max. 0.5 ppb max. 1 ppb max. 1 ppb
fluorescence at 365 nm max. 0.5 ppb max. 0.5 ppb max. 0.5 ppb
Acetonitrile
P CHROMASOLV® Prod.No. 34989
Acetonitrile
R CHROMASOLV® Prod.No. 34881
assay (GC) min. 99.8 % min. 99.8 %
non-volatile matter max. 0.0001 % max. 0.0005 %
water (Karl Fischer) max. 0.01 % max. 0.01 %
free acid (as CH3COOH) - max. 0.001 %
free alkali (as NH3) max. 0.0005 % -
transmittance at 200 nm - min. 50 %
transmittance at 220 nm min. 50 % min. 90 %
transmittance from 240 nm min. 99 % min. 99 %
Product Range
G CHROMASOLV®
34998
Acetonitrile G CHROMASOLV® super gradient grade 1; 2.5; 7; 18; 45; 200 L
34963
Ethanol G CHROMASOLV® 1; 2.5 L
34964
Ethanol G CHROMASOLV® (for export) 1; 2.5 L
34885
Methanol G CHROMASOLV® 1; 2.5; 7; 18; 45; 200 L
34959
2-Propanol G CHROMASOLV® 1; 2.5 L
34877
Water G CHROMASOLV® 1; 2.5 L
CHROMASOLV®
34850
Aceton CHROMASOLV® 1; 2.5; 45 L
34851
Acetonitrile CHROMASOLV® gradient grade 1; 2.5; 7; 18; 45; 200 L
34867
1-Butanol CHROMASOLV® 2.5 L
34861
Butanone CHROMASOLV® 1; 2.5 L
34875
tert.-Butylmethylether CHROMASOLV® 1; 2.5; 7 L
34958
1-Chlorobutane CHROMASOLV® 1 L
34855
Cyclohexane CHROMASOLV® 1; 2.5; 7 L
34872
1,2-Dichloroethane CHROMASOLV® 1 L
34856
Dichloromethane CHROMASOLV® 1; 2.5; 7 L
34889
Dimethoxymethane CHROMASOLV® 2.5; 7 L
34869
Dimethylsulfoxide CHROMASOLV® 1; 2.5 L
34857
1,4-Dioxane CHROMASOLV® 1; 2.5 L
34858
Ethylacetate CHROMASOLV® 1; 2.5 L
34852
ethanol CHROMASOLV® 1; 2.5; 7 L
34870
ethanol CHROMASOLV® (for export) 1; 2.5 L
34873
n-Heptane CHROMASOLV® 1; 2.5; 7; 45 L
34994
Iso-Hexane CHROMASOLV® 1; 2.5 L
34859
n-Hexane CHROMASOLV® 1; 2.5; 7; 45 L
34860
methanol CHROMASOLV® 1; 2.5; 7; 18; 45; 200 L
34862
Iso-Octane CHROMASOLV® 2.5 L
34956
n-Pentane CHROMASOLV® 1; 2.5 L
34871
1-Propanol CHROMASOLV® 1; 2.5 L
34863
2-Propanol CHROMASOLV® 1; 2.5; 7; 45 L
34864
Carbon tetrachloride CHROMASOLV® 1 L
34865
Tetrahydrofuran CHROMASOLV® 1; 2.5; 7; 45 L
34866
Toluene CHROMASOLV® 1; 2.5; 18; 45 L
34854
Trichloromethane CHROMASOLV® 1; 2.5; 45 L
34874
1,1,2-Trichlorotrifluoroethane CHROMASOLV® 1; 2.5 L
E CHROMASOLV®
34888
Acetonitrile E CHROMASOLV®, far UV 1; 2.5; 18 L
P CHROMASOLV®
34989
Acetonitrile P CHROMASOLV® 2.5; 18; 45; 185 L
34973
tert.-Butylmethylether P CHROMASOLV® 18; 45; 200 L
34977
Dichloromethane P CHROMASOLV® 45; 200 L
34979
Ethylacetate P CHROMASOLV® 2.5; 18; 45; 185 L
34981
n-Heptane P CHROMASOLV® 2.5; 18; 45 L
34969
iso-Hexane P CHROMASOLV® 2.5; 45; 200 L
34993
n-Hexane P CHROMASOLV® 2.5; 200 L
34983
Methanol P CHROMASOLV® 2.5; 18; 45; 185 L
34991
1-Propanol P CHROMASOLV® 2.5; 200 L
34985
2-Propanol P CHROMASOLV® 18; 45; 185 L
34987
Toluene P CHROMASOLV® 18; 45; 200 L
34975
Trichloromethane P CHROMASOLV® 18; 45; 200 L
R CHROMASOLV®
34881
Acetonitrile R CHROMASOLV®, far UV 1; 2.5; 7; 18; 45; 200 L
Back to top
Concept of LC-MS CHROMASOLV® Solvents for LC-MS Analysis
IMPROVE STABILITY OF LC-MS BASELINES BY USING HIGH-QUALITY SOLVENTS
Using LC-MS CHROMASOLV® eliminates impurities and the need to lot-select your solvents.
LC-MS is fast becoming the dominant analytical tool for researchers in a wide range of industries and application areas. LC-MS combines the advantages of a chromatographic separation with mass detection by MS. Low detection limits and qualified analysis of molecular structures brought this application a new and growing popularity especially in the field of biotechnology and within the R&D departments of the pharmaceutical industry.
Besides the column and instrument, the solvent can seriously affect the performance of an LC-MS system. Impurities arising from the solvent can mask low levels of target ions, or give false peaks, resulting in faulty interpretation of spectra. Sigma-Aldrich's Riedel-de Haën brand of LC-MS CHROMASOLV solvents is manufactured specifically to meet the needs of modern LC-MS instruments because they are designed to have low contents (max. 100 ppb) of alkaline impurities, such as calcium, magnesium, potassium and sodium, which can form clusters with the analyte, especially when using electro spray ionization. All LC-MS CHROMASOLV solvents undergo a battery of specific UV-spectroscopic quality control tests to guarantee the traditional Riedel-de Haën CHROMASOLV specifications and to ensure the reproducibility. They are also checked for LC-MS use by a special test and for particles, which could clog up Nano-LC inlet filters.
For more details see our latest researches: pdf 370 KB.
39253
Water LC-MS CHROMASOLV®
1 L
Specifications/Product Range
LC-MS solvent Water Acetonitrile Methanol 2-Propanol Ethylacetate
product number 39253 34967 34966 34965 34972
pack sizes 1 L 1 L / 2.5 L 1 L / 2.5 L 1 L / 2.5 L 1 L / 2.5 L
assay (GC) - min. 99.9 % min. 99.9 % min. 99.9 % min. 99.7 %
fluorescence at 254 nm max. 1 ppb max. 0.5 ppb max. 1 ppb max.1 ppb -
fluorescence at 365 nm max. 1 ppb max. 0.5 ppb max. 1 ppb max. 1ppb -
Chlorid (Cl) max. 0.000001% - - - -
Fluorid (F) max. 0.000001% - - - -
Nitrate (NO3) max. 0.00001% - - - -
Sulfate (SO4) max. 0.00001% - - - -
free acid - max. 0.001 % max. 0.001 % max. 0.001 % -
free alkali (as NH3) - max. 0.0002 % max. 0.0005 % max. 0.0005 % max. 0.0005 %
non-volatile matter max. 0.001 % max. 0.0002 % max. 0.0005 % max. 0.0005 % max. 0.0005 %
water (Karl Fischer) - max. 0.01 % max. 0.02 % max. 0.05 % max. 0.03 %
transmittance at 200 nm min. 95% min. 95% - - -
transmittance at 230 nm min. 99% min. 99% min. 75% min. 75% -
transmittance at 260 m - - min. 98% min. 98% min. 50%
HPLC gradient (254 nm) max. 1 mAU max. 0.2 mAU max. 2 mAU max. 2 mAU -
Silver (Ag) max. 0.1 ppm max. 0.1 ppm max. 0.1 ppm max. 0.1 ppm -
Aluminum (Al) max. 0.5 ppm max. 0.5 ppm max. 0.5 ppm max. 0.5 ppm -
Barium (Ba) max. 0.1 ppm max. 0.1 ppm max. 0.1 ppm max. 0.1 ppm -
Calcium (Ca) max. 0.1 ppm max. 0.1 ppm max. 0.1 ppm max. 0.1 ppm max. 0.1 ppm
Cadmium (Cd) max. 0.05 ppm max. 0.05 ppm max. 0.05 ppm max. 0.05 ppm -
Cobalt (Co) max. 0.02 ppm max. 0.02 ppm max. 0.02 ppm max. 0.02 ppm -
Chromium (Cr) max. 0.02 ppm max. 0.02 ppm max. 0.02 ppm max. 0.02 ppm -
Copper (Cu) max. 0.02 ppm max. 0.02 ppm max. 0.01 ppm max. 0.02 ppm -
Iron (Fe) max. 0.1 ppm max. 0.1 ppm max. 0.1 ppm max. 0.1 ppm -
Potassium (K) max. 0.1 ppm max. 0.1 ppm max. 0.1 ppm max. 0.1 ppm max. 0.1 ppm
Magnesium (Mg) max. 0.1 ppm max. 0.1 ppm max. 0.1 ppm max. 0.1 ppm max. 0.1 ppm
Manganese (Mn) max. 0.02 ppm max. 0.02 ppm max. 0.01 ppm max. 0.02 ppm -
Sodium (Na) max. 0.1 ppm max. 0.1 ppm max. 0.1 ppm max. 0.1 ppm max. 0.1 ppm
Nickel (Ni) max. 0.02 ppm max. 0.02 ppm max. 0.02 ppm max. 0.02 ppm -
Lead (Pb) max. 0.1 ppm max. 0.1 ppm max. 0.02 ppm max. 0.1 ppm -
Tin (Sn) max. 0.1 ppm max. 0.1 ppm max. 0.1 ppm max. 0.1 ppm -
Zinc (Zn) max. 0.1 ppm max. 0.1 ppm max. 0.1 ppm max. 0.1 ppm -
particle test + + + - -
LC-MS suitability test + + + + +
Back to top
LC-MS CHROMASOLV® - Solvent Blends
LC-MS blends
LC-MS allows the detection and quantification of many analytes. The minimisation of artefacts requires very well specified solvents spiked with ultra pure salts and acids. These additives are used to improve the chromatographic peak shape and to optimise ionisation in the MS interface. The most commonly used solvents in LC-MS techniques are acetonitrile, methanol and water. Additives can include trifluoracetic acid, formic acid, acetic acid and ammonium acetate.
For more details see our latest Labinfo: pdf 104 KB
Catalog No.
Maktx
Package Size
Specification
34976
ACETONITRILE WITH 0.1 VOL.% TRIFLUORACETIC ACID LC-MS CHROMASOLV® 2.5L amber bottle
LC gradient testing in UV and MS, metals impurities (Na < 2ppm, K, Mg, Ca
< 0.5ppm), UV-Absorption, additive content: 0.093-0.107 (TFA, FA, AA: v/v, Ammoniumacetat: g/v) Solvents content: (GC): > 99.0% (exeption 34669 Acetonitrile with 0.1% ammonium acetate:
> 98%)
34678
ACETONITRILE WITH 0.1% ACETIC ACID LC-MS CHROMASOLV® 2.5L amber bottle
34669
ACETONITRILE WITH 0.1% AMMONIUM ACETATE LC-MS CHROMASOLV® 2.5L amber bottle
34668
ACETONITRILE WITH 0.1% FORMIC ACID LC-MS CHROMASOLV® 2.5L amber bottle
34672
METHANOL WITH 0.1% ACETIC ACID LC-MS CHROMASOLV® 2.5L amber bottle
34670
METHANOL WITH 0.1% AMMONIUM ACETATE LC-MS CHROMASOLV® 2.5L amber bottle
34974
METHANOL WITH 0.1% TRIFLUOROACETIC ACID LC-MS CHROMASOLV® 2.5L amber bottle
34978
WATER WITH 0.1 VOL.% TRIFLUORACETIC ACID LC-MS CHROMASOLV® 2.5L amber bottle LC gradient testing in UV and MS, metals impurities (Na < 2ppm, K, Mg, Ca
< 0.5ppm), UV-Absorption, additive content: 0.093-0.107 (TFA, FA, AA: v/v, Ammonium acetate: g/v), pH: effective
+/- 0.1
34675
WATER WITH 0.1% ACETIC ACID LC-MS CHROMASOLV® 2.5L amber bottle
34674
WATER WITH 0.1% AMMONIUM ACETATE LC-MS CHROMASOLV® 2.5L amber bottle
34673
WATER WITH 0.1% FORMIC ACID LC-MS CHROMASOLV® 2.5L amber bottle
34689
WATER/2-PROPANOL LC-MS CHROMASOL LC-MS CHROMASOLV® 1L white glass bottle rinsing agent for
LC-MS
Back to top
LC-NMR CHROMASOLV® Solvents for LC-NMR Analysis
Although an efficient separation method is available by HPLC, no structure determination is possible when detecting separated compounds by UV-detector. Thus NMR spectroscopy is a better universial detection method. A combination of both methods was evident and LC-NMR was born.
Solvents that meet the high requirements of HPLC and proton NMR are needed. But the high throughput of solvents in HPLC does not allow the usage of expensive deuterated solvents as usually required for NMR. HPLC Acetonitrile is often contaminated with impurities containing protons such as propionitrile which is no problem in UV but NMR detection. Therefore a dramatic decrease of proton containing contaminants for LC-NMR solvents is essential.
Acetonitrile LC-NMR CHROMASOLV® (34955) from Riedel-de Haën is the solvent of choice. It was developed to obtain the superior specifications required for LC-NMR. There are no further signals in 1H-NMR that exceed the size of methyl signals (more than 5% at 3.78 ppm NMR of 0.0006% trimethoxybenzene) at typical LC-NMR conditions.
Acetonitrile/ Prod.No. 34955 / 0.5 L / 1 L
assay (GC) min. 99.9 %
non-volatile matter max. 0.0002 %
water (Karl Fischer) max. 0.01 %
free acid (as CH3COOH) max. 0.001 %
free alkali (as NH3) max. 0.0001 %
propionitrile (GC) max. 0.0002 %
For additional information, please contact your local Sigma-Aldrich office.
Back to top
Back to LC home