Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Liquid Chromotography
Liquid Chromatography Standards
Introduction
1. Phytopharmaca derived from naturally occurring ingredients are actively being investigated for their health-promoting potential. Because modern medicinal forms in therapy have become a main topic in clinical science, phytopharmaca are widely used in the pharmaceutical production of medicaments. Many Phytopharmacas are also used as nutritional supplements in food. For performing reliable quality control, standards of Phytopharmaca are needed. Fluka has been innovated in this product range and provides you a selection of the most important analytical standards (see Table 1).
2. Amino Acid calibration standards
The concentration of each standard is verified by amino acid analysis methods. The single amino acid solution are supplied at a concentration of 100 mM in 0.1 N HCl; multicomponent solutions are available in concentrations from 0.01 mM to 1 mM (each component). Take a look at Table 2!
Table 1. Phytopharmaca HPLC Standards (New!)
Catalog No.
Product Name
Quality
Cas Number
Package Size
39393
Aloe-emodine purum, ≥ 95.0% (HPLC)
481-72-1
25 mg
53017
alpha-Amyrin puriss. p.a., ≥98.5% (HPLC)
638-95-9
10 mg
09236
beta-Amyrin puriss. p.a., ≥98.5% (HPLC)
559-70-6
10 mg
44692
Apigenin-7-glucosid purum, ≥98.0% (HPLC)
578-74-5
5 mg, 25 mg
43191
Asiaticosid puriss. p.a., ≥98.5% (HPLC) for HPLC
16830-15-2
1.5 mg
55561
Aucubin puriss., ≥99.0% (HPLC)
479-98-1
5 mg, 25 mg
01338
Bergamottin purum, ≥98.0% (DC)
7380-40-7
5 mg
74581
Boldin purum, ≥99.0% (HPLC)
476-70-0
25 mg, 100 mg
88527
Citropten puriss., ≥99.0% (HPLC)
487-06-9
5 mg, 25 mg
28078
9,11-Didehydroestriol purum, ≥97.0% (HPLC)
246021-20-5
1 mg, 5 mg
09258
Erythrodiol purum p.a., ≥97.0% (HPLC)
545-48-2
10 mg, 50 mg
59780
DL-Kavain puriss., ≥99.0% (HPLC)
3155-48-4
0.5 g, 5 g
78564
Ginkgolid A BioChemika purum, ≥95.0% (HPLC)
15291-75-5
10 mg, 50 mg
05108
Ginkgolid B BioChemika purum, ≥95.0% (HPLC)
15291-77-7
10 mg, 50 mg
44349
Ginkgolid C BioChemika purum, ≥90.0% (HPLC)
15291-76-6
5 mg, 25 mg
97151
Hederacosid C purum, ≥95.0% (HPLC)
14216-03-6
10 mg, 50 mg
36483
Loganin purum, ≥97.0% (HPLC)
18524-94-2
10 mg, 50 mg
09237
Myristicin purum, ≥97.0% (GC)
607-91-0
10 mg, 50 mg
69249
Quercetin dihydrate ≥99.0% (HPLC)
6151-25-3
1 mg, 5 mg
17799
Rhamnetin BioChemika, ≥99.0% (HPLC)
90-19-7
1 mg, 5 mg
44699
Rosmarinic acid purum, ≥97.0% (HPLC)
20283-92-5
50 mg, 250 mg
78095
Rutin trihydrate purum, ≥95.0% (HPLC)
250249-75-3
25 mg, 100 mg
68909
Sennosid A purum, ≥98.0% (HPLC)
128-57-4
25 mg
75412
Sennosid B purum, ≥98.0% (HPLC)
81-27-6
5 mg, 25 mg
78666
Taxifolin BioChemika, ≥85% (HPLC)
17654-26-1
25 mg, 100 mg
72477
Thymol puriss. p.a., ≥99.9% (GC)
89-83-8
500 mg
18143
trans-Nerolidol purum, ≥98.0%
40716-66-3
100 mg
55608
Vitexin-2"-O-rhamnoside purum, ≥98.0% (DC)
64820-99-1
5 mg, 25 mg
Table 2. Amino Acid Callibration Standards (New!)
Catalog No.
Product Name
Description
Cas Number
Package Size
55329
L-Alanine hydrochloride solution 100 mM in 0.1 M HCl
6003-05-0
5 ml
08163
L-Arginine hydrochloride solution 100 mM in 0.1 M HCl
15595-35-4
5 ml
74392
L-Aspartic acid hydrochloride solution 100 mM in 0.1 M HCl
17585-59-0
5 ml
57579
L-Cystine hydrochloride solution 10 mM in 0.1 M HCl
34760-60-6
5 ml
96658
L-Glutamic acid hydrochloride solution 100 mM in 0.1 M HCl
138-15-8
5 ml
55097
Glycine hydrochloride solution 100 mM in 0.1 M HCl
6000-43-7
5 ml
43011
L-Histidine hydrochloride solution 100 mM in 0.1 M HCl
1007-42-7
5 ml
50271
L-Isoleucine hydrochloride solution 100 mM in 0.1 M HCl
17694-98-3
5 ml
80687
L-Leucine hydrochloride solution 100 mM in 0.1 M HCl
760-84-9
5 ml
44208
L-Lysine hydrochloride solution 100 mM in 0.1 M HCl
10098-89-2
5 ml
50272
L-Methionine hydrochloride solution 100 mM in 0.1 M HCl
6003-05-1
5 ml
44026
L-Phenylalanine hydrochloride solution 100 mM in 0.1 M HCl
17585-69-2
5 ml
94321
L-Proline hydrochloride solution 100 mM in 0.1 M HCl
7776-34-3
5 ml
61227
L-Serine hydrochloride solution 100 mM in 0.1 M HCl
16428-75-4
5 ml
69356
L-Threonine hydrochloride solution 100 mM in 0.1 M HCl
82650-07-5
5 ml
56967
L-Tyrosine hydrochloride solution 100 mM in 0.1 M HCl
16870-43-2
5 ml
19515
L-Valine hydrochloride solution 100 mM in 0.1 M HCl
17498-50-9
5 ml
Back to top
Back to LC home
Back to Analytical Standards home