Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Liquid Chromatography
Liquid Chromatography Supports
All chromatographic methods -with the exception of TLC- use columns for the separation process. Column chromatography has found it's place in many laboratories for preparative purposes as well as for reaction control in organic syntheses. The importance of column chromatography is mainly due to following factors:
  • Simple packing procedure
  • Low operating pressure
  • Low expense for instrumentation
The following table gives you an overview of all supports and adsorbents for column chromatography offered by Fluka. When choosing a column packing, you have to consider the properties of substances to be separated-including impurities or components of no interest-and then select the proper adsorbent. As a rule of thumb you can consider:
  • For lipophilic substances: aluminum oxide, silica, acetylated cellulose, polyamide
  • For hydrophilic substances: cellulose, kieselgur, polyamide
In addition to this, it's important to consider possible reactions between the compounds and the adsorbent or eluent. In practice, silica gel is the preferred stationary phase for separation of organics.
Cat.No
Name
Package size
05074
Agarose 2 B for chromatography 25 ml
05076
Agarose 4 B for chromatography 25 ml
05078
Agarose 6 B for chromatography 25 ml
06290
Aluminum oxide Fluka, type 5016 A basic 1 kg, 5 kg, 25 kg
06300
Aluminum oxide Fluka, type 507 C neutral, 100-125 mesh 1 kg, 5 kg, 25 kg
06310
Aluminum oxide Fluka, type 506 C weakly acidic 1kg
06320
Aluminum oxide Fluka, type 504 C acidic 1 kg, 2.5 kg
09297
Aminopropyl silica gel, for preparative chromatography 50 g, 250 g
09298
Aminopropyl silica gel, for preparative chromatography 100g
11365
Avicel® PH-101, ~50 mm 250 g, 1kg
15685
Boric acid Gel, for column chromatography, 0.1-0.4 mm 5 ml, 25 ml
21900
Carboxymethylcellulose Sodium salt; low viscosity 100 g, 250 g, 1kg
21902
Carboxymethylcellulose Sodium salt; medium viscosity 100 g, 250 g, 1kg
21903
Carboxymethylcellulose Sodium salt; high viscosity 100 g, 250 g, 1kg
21904
Carboxymethylcellulose Sodium salt; ultra high viscosity 100 g, 250 g, 1kg
21901
Carboxymethylcellulose Sodium salt; ultra low viscosity 100 g, 250 g, 1kg
22182
Cellulose powder, spruce, for column chromatography, acid washed 250 g, 1kg
22181
Cellulose powder from spruce, for column chromatography 250 g, 1kg
22184
Cellulose powder, for column chromatography, acid washed 250 g, 1kg
22183
Cellulose powder, for column chromatography 250 g, 1kg
02563
Cellulose acetate SAC 20, acetylated 250g
22192
Cellulose acetate phthalate 100 g, 500 g
22200
Cellulose triacetate 50 g, 250 g, 1kg
22503
Chelite P, Na+-form, 25-40 mesh 100 g, 500 g
26960
Cholestyramine resin, for column chromatography 50 g, 250 g
27560
CM-Dextran Sodium salt 5 g, 25 g
CM Dextranomer C-25-120 Synonym Carboxymethyl-dextran 5 g. 25 g, 100 g
CM Dextranomer C-50-120 Synonym Carboxymethyl-dextran 5 g. 25 g, 100 g
27563
CM-Sepharose® CL-6B, for chromatography 50 ml, 250 ml
27565
CM-Sepharose® 'Fast Flow', for chromatography 100 ml, 500 ml
30477
DEAE-Cellulose, 100-200 mm 25 g, 100 g
30461
DEAE-Dextran hydrochloride, for molecular biology 25 g, 100 g
30462
DEAE-Dextran hydrochloride 25 g, 100 g
30473
DEAE-Sepharose® CL-6B, for chromatography 25 ml, 100 ml, 500 ml
30474
DEAE-Sepharose® 'Fast Flow', for chromatography* 50 ml, 250 ml
31399
Dextrangel N 25 for column chromatography 5 g, 25 g, 100 g
44785
Ecteola-23 Cellulose 100 g, 500 g
29227
FlashTubeTM 2002 1ST
29229
FlashTubeTM 2008 1ST
03286
Florisil® acc. to ISO 9377-2/H53, for Water Analysis; further details 100 g, 500 g
46381
Florisil® for Chromatography, 16-30 mesh 100 g, 500 g
46384
Florisil® for Chromatography, 30-60 mesh 100 g, 500 g
46385
Florisil® for Chromatography, 60-100 mesh 100 g, 500 g
46386
Florisil® for Chromatography, 100-200 mesh 100 g, 500 g
46382
Florisil® PR, for pesticide residue analysis 60-100 mesh 100 g, 500 g
55496
Hydroxylapatite 'high resolution' 10 g, 50 g
55497
Hydroxylapatite 'fast flow' 10 g, 50 g
02395
Polyamide 6, for column chromatography 250 g
83332
QAE-Sepharose® 'Fast Flow', for chromatography 100 ml, 500 ml
84936
Sephacryl® S-200, for chromatography 100 ml, 500 ml
84937
Sephacryl® S-300, for chromatography 100 ml, 500 ml
84939
Sephacryl® S-300-HR, for chromatography 100 ml, 500 ml
84941
Sephadex® G-10 for chromatography, 40-120 mm 25 g, 100 g, 500 g
84942
Sephadex® G-25 fine for chromatography, 20-80 mm 25 g, 100 g, 500 g
84943
Sephadex® G-25 medium for chromatography, 50-150 mm 10 g, 50 g, 250 g
84944
Sephadex® G-25 coarse for chromatography, 100-300 mm 10 g, 50 g, 250 g
84946
Sephadex® G-50 superfine for chromatography, 20-50 mm 10 g, 50 g, 250 g
84947
Sephadex® G-50 fine for chromatography, 20-80 mm 10 g, 50 g, 250 g
84948
Sephadex® G-50 medium for chromatography, 50-150 mm 10 g, 50 g, 250 g
84949
Sephadex® G-75 for chromatography, 40-120 mm 10 g, 50 g, 250 g
84934
Sephadex® G-75-50 superfine, for chromatography 10 g, 50 g, 250 g
84951
Sephadex® G-100 for chromatography, 40-120 mm 10 g, 50 g, 250 g
84961
Sephadex® G-200 superfine, for chromatography 10 g, 25 g, 100 g
84954
Sephadex® CM C-25 for chromatography, 40-120 mm 10 g, 50 g, 250 g
84955
Sephadex® CM C-50 for chromatography, 40-120 mm 10 g, 50 g, 250 g
84956
Sephadex® DEAE A-25 for chromatography, 40-120 mm 10 g, 50 g, 250 g
84957
Sephadex® DEAE A-50 for chromatography, 40-120 mm 10 g, 50 g, 250 g
84952
Sephadex® LH-20 for chromatography, 25-100 mm 25 g, 100 g, 500 g
84962
Sepharose® 4B for chromatography 100 ml, 250 ml, 1l
84964
Sepharose® 6B forchromatography 250 ml, 1l
84963
Sepharose® CL-4B forchromatography 250 ml, 1l
84965
Sepharose® CL-6B forchromatography 250 ml, 1l
84967
Sepharose® 6B epoxy-activated for chromatography 5 g, 10 g
85636
S Sepharose® 'Fast Flow', for chromatography 100 ml, 500 ml
85328
Silica-Alox 1kg
60734
Silica gel 40 for column chromatography 0.015-0.035 mm 100 g, 500 g
60735
Silica gel 40 for column chromatography 0.063-0.2 mm 250 g, 1kg
60736
Silica gel 40 for column chromatography 0.2-0.5 mm 250 g, 1kg
60750
Silica gel 40 for preparative column chromatography 0.04-0.063 mm 100 g, 500 g
60737
Silica gel 60 for production column chromatography 0.04-0.063 mm 500 g, 2.5kg
60738
Silica gel 60 for column chromatography 0.035-0.070 mm 250 g, , 25 kg, 1kg
60739
Silica gel 60 for column chromatography <0.063 mm 250 g, 1 kg
60740
Silica gel 60 for column chromatography 0.063-0.2 mm 100 g, 500 g
60741
Silica gel 60 for column chromatography 0.063-0.2 mm 250 g, 1 kg, 25 kg
60742
Silica gel 60 for column chromatography 0.2-0.5 mm 250 g, 1 kg, 25 kg
60751
Silica gel 60, for for preparative column chromatography <0.04 mm 100 g, 500 g
60752
Silica gel 60 for preparative column chromatography 0.040-0.063 mm 500 g, 1 kg, 25 kg
00223
Silica gel 60, for column chromatography, 20-40 mm 250g
60743
Silica gel 60 F254 for column chromatography. 0.063-0.2 mm 250 g, 1 kg
60744
Silica gel 100 for column chromatography 0.015-0.035 mm 100 g, 500 g
60745
Silica gel 100 for column chromatography 0.063-0.2 mm 250 g, 1 kg
60746
Silica gel 100 for column chromatography 0.2-0.5 mm 250 g, 1 kg
60753
Silica gel 100 for or preparative column chromatography 0.04-0.063 mm 100 g, 500 g
60747
Silica gel 100 CN for column chromatography 0.040-0.063 mm 50 g, 250 g
60748
Silica gel 100 CN for column chromatography0.015-0.035 mm 50 g, 250 g
60755
Silica gel 100 C8-Reversed phase, 0.015-0.035 mm 50 g, 250 g
60755
Silica gel 100 C8-Reversed phase, 0.015-0.035 mm 50 g, 250 g
60759
Silica gel 100 C8-Reversed phase, 0.040-0.063 mm 50 g, 250 g
60754
Silica gel 100 C18-Reversed phase for column chromatography 50 g, 250 g
60756
Silica gel 100 C18-Reversed phase for column chromatography 50 g, 250 g
60757
Silica gel 100 C18-Reversed phase for column chromatography 50 g, 250 g
60758
Silica gel 100 C18-Reversed phase for column chromatography 50 g, 250 g
03563
Silica gel E, 0.5-1.0 mm, according ToIxfeld and Buck 250 g, 1kg
03559
Silica gel FIA, 0.071-0.63 mm, acc. to DIN 51791 250 g, 1kg
03561
Silica gel FIA, 0.071-0.16 mm, acc. to DIN 51791 250 g, 1kg
Silica gel, spherical, click for further information  
60780
Silicic acid Hydrate, powder, particle size 100 mesh 100 g, 1kg
60780
Silicic acid Hydrate, powder, particle size 100 mesh 250 g
02388
Tribenzoylcellulose Suspension, 10-20 mm, ~40% in Methanol/Water 100 ml, 500 ml
Spherical Silica Gel
These high value products are mechanically very stable and ensure excellent column packing reproducibility and column performance. The wide variety of pore size from 70 Å to 300 Å allows the application for a broad range of separation processes including separations of peptides and proteins. Convince yourself about improved chromatography results!
  • Porous spherical particles
  • Narrow pore and particle size distribution
  • High mechanical Stability
  • High column packing reproducibility
Spheical Silica Gel
Figure 1: Particle size distribution of Silica Gel 100 Å
93875
95021
86981
89752
Nominal pore diam.
Å
70
100
200
300
Specific surface area
m2/g
480
270
130
90
Specific pore volume
ml/g
0.8
0.9
0.9
0.9
pH (10% aqueous solution)  
6
6
6
6
Res. Water (160°C)
%
<2
<2
<2
<2
Fe
ppm
30
30
30
30
Pb
ppm
1
1
1
1
Particle sizes
mm
40
40
40
40
Florisil® for Water Analysis acc. EN ISO 9377-2/H53
Hydrocarbons in water have to be determined according guideline EN ISO 9377-2/H53.
The method requires a clean-up step using Florisil® as adsorbent. We responded to many inquiries from customers and developed Florisil® which is approved according EN ISO 9377-2/H53.
Specifications
  • Recovery Rate in Mineral Oil: >80.0 %
  • Relation Peak Area Stearylstearate Treated/Untreated: <1.0
  • Particle Size: 60-100 mesh
Product List
Prod.No.
Name
Pack Size
03286
Florisil® acc. to ISO 9377-2/H53
100 g, 500 g
Back to top
Back to LC Home