HPCE Buffers
IEF Markers
Anion HPCE-Kit
HPCE Buffers
Introduction
The rapidly growing capabilities of HPCE have created a great demand for reagents of an appropriate quality.
With the introduction of a series of ready-to-use buffers covering the pH range from 2.5 to 11, Fluka now meets exactly the requirements of many analysts dealing with HPCE.
Quality Guarantee
We guarantee for each reagent:
- No insoluble impurities—the HPCE buffers are filtered through a 0.2 mm filter membrane after production
- Minimal absorption over a wide wave length range—see figure 1 below
- Virtually no fluorescent impurities—see figure 2 below
- Tested for application—see figure 3 below
 Figure 1: UV/VIS absorption of the buffer solution pH 2.5 for HPCE (Product No. 82581) in the wave-length range 200-800 nm. |
|
 Figure 2: Residual fluorescence of the buffer solution pH 2.5 for HPCE (Product No. 82581). The fluorescence is checked by exitation at 200, 230 and 260 nm. This illustrates the excellent suitability of this buffer for applications with fluorescence detection. Standard (S): 10-7M ovalene, embedded in a polymethylmethacrylate (PMMA) matrix, excited at 342 nm. |
|
 Figure 3: Separation of the two peptides aprotinin (A) and soybean trypsin inhibitor (STI). Sample: containing 50 ng/ml of each peptide. Separation buffer: 20 nm sodium citrate pH 2.5 (Product No. 82581). Capillary: fused silica, 60 cm in length, 50 mm i.d. Injection: hydrostatic 3 sec, DH =9.8 cm. Field: 20 kV. Detection: 200 nm |
Product List: Buffer solutions for HPCE
| Product No. |
Description |
Package Size |
Add to Cart |
| 82609 |
Buffer Solution pH 2.5 for HPCE [20 mM citric acid-NaOH] |
50 ml/100 ml |
|
| 82581 |
Buffer Solution pH 2.5 for HPCE [20 mM sodium citrate] |
50 ml/100 ml/500 ml |
|
| 82582 |
Buffer Solution pH 3.0 for HPCE [20 mM sodium citrate] |
50 ml/100 ml/500 ml |
|
| 82621 |
Buffer Solution pH 3.0 for HPCE [Cupric electrolyte buffer pH 3.0] special quality for the detection of cations (alkali and alkaline earth metals and amines) |
50 ml/100 ml |
|
| 82622 |
Buffer Solution pH 3.0 for HPCE [150 mM potassium phosphate] |
100 ml/500 ml |
|
| 82582 |
Buffer Solution pH 3.0 for HPCE [20 mM sodium citrate] |
50 ml/100 ml/500 ml |
|
| 82583 |
Buffer Solution pH 3.5 for HPCE [20 mM sodium citrate] |
50 ml/100 ml/500 ml |
|
| 82584 |
Buffer Solution pH 4.0 for HPCE [20 mM sodium citrate] |
50 ml/100 ml/500 ml |
|
| 82585 |
Buffer Solution pH 4.5 for HPCE [20 mM sodium citrate] |
50 ml/100 ml/500 ml |
|
| 82586 |
Buffer Solution pH 5.0 for HPCE [20 mM sodium citrate] |
50 ml/100 ml/500 ml |
|
| 82587 |
Buffer Solution pH 5.5 for HPCE [20 mM sodium citrate] |
50 ml/100 ml/500 ml |
|
| 82588 |
Buffer Solution pH 6.0 for HPCE [20 mM sodium citrate] |
50 ml/100 ml/500 ml |
|
| 82589 |
Buffer Solution pH 6.5 for HPCE [20 mM sodium phosphate] |
50 ml/100 ml/500 ml |
|
| 82591 |
Buffer Solution pH 7.0 for HPCE [20 mM borate-phosphate] |
50 ml/100 ml/500 ml |
|
| 82614 |
Buffer Solution pH 7.0 with SDS f.HPCE [100 mM boric acid/50 mM sodium phosphate, with 50 mM SDS] special quality for Micellar Electrokinetic CapillaryChromatography (MECC) |
50 ml/100 ml |
|
| 82591 |
Buffer Solution pH 7.0 for HPCE [20 mM sodium citrate] |
50 ml/100 ml/500 ml |
|
| 82636 |
Buffer Solution pH 7.0 for HPCE [50 mM sodium citrate] |
50 ml/100 ml |
|
| 82637 |
Buffer Solution pH 7.0 for HPCE [100 mM sodium citrate] |
50 ml/100 ml |
|
| 82592 |
Buffer Solution pH 7.5 for HPCE [20 mM sodium phosphate] |
50 ml/100 ml |
|
| 82619 |
Buffer Solution pH 7.7 for HPCE [Pyromellitic acid electrolyte buffer pH 7.7] special quality for the detection of inorganic and low molecular weight organic acid anions |
50 ml/100 ml |
|
| 82593 |
Buffer Solution pH 8.0 for HPCE [20 mM sodium phosphate] |
50 ml/100 ml |
|
| 82594 |
Buffer Solution pH 8.0 for HPCE [20 mM sodium tetraborate] |
50 ml/100 ml/500 ml |
|
| 82615 |
Buffer Solution pH 8.0 with methylcellulose for HPCEM [50 mM TRIS-borate/ 2.5 mM EDTA, 0.5% methylcellulose] special quality for separation of DNA restrictions fragments |
50 ml/100 ml |
|
| 82633 |
Buffer Solution pH 8.0 for HPCE [50 mM sodium borate] |
50 ml/100 ml |
|
| 82634 |
Buffer Solution pH 8.0 for HPCE [100 mM sodium borate] |
100 ml/500 ml |
|
| 82601 |
Buffer Solution pH 8.5 for HPCE [20 mM sodium phosphate] |
50 ml/100 ml/500 ml |
|
| 82602 |
Buffer Solution pH 8.5 for HPCE [20 mM sodium tetraborate] |
50 ml/100 ml/500 ml |
|
| 82616 |
Buffer Solution pH 8.6 with urea for HPCE [100 mM TRIS/100 mM boric acid/2 mM EDTA/7 M urea] special quality for separation of nucleic acids |
50 ml/100 ml |
|
| 82603 |
Buffer Solution pH 9.0 for HPCE [20 mM sodium phosphate] |
50 ml/100 ml/500 ml |
|
| 82604 |
Buffer Solution pH 9.0 for HPCE [20 mM sodium tetraborate] |
50 ml/100 ml/500 ml |
|
| 82605 |
Buffer Solution pH 9.5 for HPCE [20 mM sodium phosphate] |
50 ml/100 ml/500 ml |
|
| 82606 |
Buffer Solution pH 10.0 for HPCE [20 mM CAPS] |
50 ml/100 ml/500 ml |
|
| 82607 |
Buffer Solution pH 10.5 for HPCE [20 mM CAPS] |
50 ml/100 ml/500 ml |
|
| 82608 |
Buffer Solution pH 11.0 for HPCE [20 mM CAPS] |
50 ml/100 ml/500 ml |
|
| 82617 |
Buffer Solution pH 11.0 for HPCE [20 mM glycine-NaOH] |
50 ml/100 ml |
|
| 84428 |
Hydrochloric acid Solution for HPCE |
100 ml/500 ml |
|
| 72079 |
Sodium hydroxide Solution for HPCE |
100 ml/500 ml |
|
| 95283 |
Water for HPCE |
250 ml/1 l |
|
back to top
IEF Markers
Application
IEF (Isoelectric Focusing) is a powerful analytical tool for the separation of ampholytes, mainly proteins. In order to ensure the high performance of analysis, standards of pI (pI markers) are needed. In addition to classical protein based standards, low molecular compounds were developed and successfully examined in capillary IEF and IEF-Gel electrophoresis. For capillary IEF, UV absorption is the most poular method in use. UV induced fluorescence emission is of interest if derivatizations of proteins with e.g. dansyl chloride, fluorescamine, o-phtaldialdehyde or coumarin moieties are used to increase sensitivity. Another advantage for IEF gel electrophoresis with Fluorescent IEF-Marker is the possibility to control the formation of gradient without further staining (using illumination UV).
Fluorescent IEF markers can also be detected by UV-absorption at 280 nm (20°C), allthough the signal is not as strong as with fluorescence detection. The absorption maxima of the individual markers are between 308 and 350 nm. For fluorescence detection an excitation wavelength of 310 nm (individual excitation maxima: 310 to 400 nm) is suggested; the emission maximum of the individual markers lies between 410 and 500 nm.
Product Description
Solids:
- 1 mg packages
- Fluorescenz: Emmax and Exc. (see table 1)
- Storage at 4°C
Stock Solution:
- 200 ml packages
- Fluorescenz: Emmax, Exc. and conditions (see table 1)
- Concentration: 1, 2, 3 mg /ml
- Solution: aqueous (see table 1)
- Filtered through 0.45 mm membrane filter
- Store at 4°C; stable for at least 6 months
Analysis Conditions for CIEF using Pressure Mobilization
- Capillary: neutral capillary
- Anolyte: 91 mM phosphoric acide in gel buffer
- Catholyte: 20 mM sodium hydroxide in water
- Detection: 280 nm
- Temperature: 20 °C
- Injection: 20 psi, 1 min
- Polarity: inlet anode, outlet cathode
- Focussing voltage: 500 V/cm
- Focussing time: 2 min
- Mobilization: 0.5 psi, 500 V/cm anolyte -> catholyte (Mobilization should be stopped after the last marker is eluted to avoid the filling of the capillary with anolyte.)
Figure 4. Example for CE-IEF with fluorescent pI Markers
pI Marker:
Peak 1: 8.7
Peak 2: 7.6
Peak 3: 6.6
Peak 4: 6.2
Figure 5. The emission spectrum and emission maximum of the Fluorescent IEF-Marker pI 7.6
Figure 6. The UV spectrum of the Fluorescent IEF-Marker pI 7.6 (three different concentration)
Table 1. Conditions of Fluorescent IEF Markers stock solutions.
Additional information on Fluorescent IEF Markers in chapter Fluorescent Probes.
| Catalog Number |
Description |
pl |
Stock Solution Concentration in ultrapure water |
Fluorescence |
| Marker [mg/ml] |
HCl [mM] |
2-propanol (%) |
Emmax [nm] |
Exc. [nm] |
Buffer for measurement pH=pl |
| 35096 74169 |
solid stock solution |
2.1 |
3 |
- |
50 |
430 |
340 |
50mM Citrate, 50mM KCl |
| 17952 72172 |
solid stock solution |
3.0 |
2 |
5 |
50 |
440 |
360 |
0.1M Citrate |
| 17953 40677 |
solid stock solution |
3.5 |
2 |
5 |
50 |
415 |
318 |
0.1M Citrate |
| 17954 89827 |
solid stock solution |
4.0 |
1 |
- |
- |
415 |
310 |
0.1M Citrate |
| 17955 89149 |
solid stock solution |
4.5 |
1 |
5 |
- |
424 |
336 |
0.1M Citrate |
| 17956 89478 |
solid stock solution |
5.1 |
2 |
10 |
- |
415 |
330 |
0.1M Citrate |
| 17957 77866 |
solid stock solution |
5.5 |
3 |
15 |
- |
412 |
325 |
0.1M Citrate |
| 17958 73938 |
solid stock solution |
6.2 |
1 |
10 |
- |
500 |
394 |
0.1M Phosphate |
| 17959 73376 |
solid stock solution |
6.6 |
1 |
10 |
- |
500 |
396 |
0.1M Phosphate |
| 17961 89508 |
solid stock solution |
6.8 |
1 |
5 |
- |
418 |
338 |
0.1M Phosphate |
| 17962 89951 |
solid stock solution |
7.2 |
1 |
4 |
- |
500 |
387 |
0.1M Phosphate |
| 17963 89952 |
solid stock solution |
7.6 |
1 |
10 |
- |
495 |
385 |
0.1M Tris |
| 17964 75734 |
solid stock solution |
8.1 |
3 |
- |
- |
420 |
340 |
0.1M Tris |
| 17966 89357 |
solid stock solution |
8.7 |
1 |
10 |
- |
500 |
390 |
0.1M Tris |
| 17967 90699 |
solid stock solution |
9.0 |
1 |
10 |
- |
495 |
385 |
0.1M Tris |
| 46276 89268 |
solid stock solution |
9.5 |
3 |
8 |
- |
415 |
325 |
0.1M Carbonate |
| 17968 77672 |
solid stock solution |
10.3 |
1 |
10 |
- |
495 |
388 |
0.1M Carbonate |
| 17951 |
stock solution of Marker-Mix |
- |
2 (of each) |
5 |
50 |
- |
- |
- |
Directions for the use of Fluorescent IEF-Markers
The protocol for the use of Fluorescent IEF-Marker is following: Prepare the IEF-marker stock solution by dissolving the substance in the solvent according to Table 1. Sonicate for up to 10 min if necessary. Store at 4 °C (the solution is stable for approximately 6 months). For use in HPCE this stock solution should be diluted 1:100 in a suitable ampholyte solution (2% ampholyte 3-10; e.g. 10046). For uses in IEF-gels the stock solution can be loaded directly onto the gel (max. 1 ml per lane).
Reference
M. Horka, Th. Willimann, M. Blum, P. Nording, Z.Friedl, K. Slais, Capillary isoelectric focusing with UV-induced fluorescence detection, J. of Chromatography A, 916, 65-71 (2001).
back to top
Anion HPCE-Kit – Product No. 29244
General Information
Capillary electrophoresis is an efficient analytical separation technique for analysis of minute amounts of sample and has several advantages, including fast separation and high resolution. The Fluka pH 7.7 buffer system is designed for the analysis of anions by indirect UV detection with reversed electro-osmotic flow. This buffer was validated for the analysis of eight common anions: fluoride, chloride, bromide, sulfate, nitrate, nitrite, thiosulfate and phosphate [1].
Contents
- 100 ml Buffer solution pH 7.7 for HPCE (Product No. 82619)
[Pyromellitic acid electrolyte buffer pH 7.7; composition: 2.25 mM pyromellitic acid, 6.50 mM sodium hydroxide, 0.75 mM hexamethonium hydroxide, 1.60 mM triethanolamine,
pH: 7.7+/- 0.2 (250C); manufactured under clean room conditions; filtered through a 0.2 mm filter]
- 100 ml Water for HPCE (Product No. 95283)
[manufactured under clean room conditions; filtered through a 0.2 mm filter]
- 10 ml Multielement Anion HPCE Standard Solution (Product No. 29235; 100 ppm)
[composition: 100 mg/l fluoride, 100 mg/l bromide, 100 mg/l chloride, 100 mg/l phosphate, 100 mg/l sulfate, 100 mg/l nitrate; manufactured from highly pure salts and water; filtered through a 0.2 mm filter]
When handling buffer solution pH 7.7 avoid contact with eyes or skin. Wear safety glasses. Store tightly closed.
- Needed but not supplied in this kit:
0.1 M sodium hydroxide solution for HPCE (Product No. 72079) or an equivalent quality.
Application
- The buffer solution pH 7.7 for HPCE is supplied ready to use. No dilutions or other treatments are required.
- The Multielement Anion Standard is supplied as a 100 ppm concentrate (0.1 g/l) of the six anions. Dilute the Multielement Anion Standard with Water for HPCE to your desired concentration (typically 1 ppm to 10 ppm). This working standard solution may be used as capillary test solution or for peak identification and quantification.
- Sample preparation:
Samples are either used directly or diluted with water for HPCE (Product No. 95283) to a concentration of approximately 1 to 10 mg/l of the anion of interest. Detection limits are approximately at 0.5 mg/l of the anion of interest.
Working Instructions
- Capillary treatment:
For anion separation, use a fused silica capillary with an inner diameter of 50-75 mm, 50-60 cm total length (40-50 cm from inlet to detector). Rinse the capillary with 0.1 M sodium hydroxide solution for HPCE (Product No. 72079) for at least 10 min, then for 10 to 20 min with buffer solution pH 7.7. Do not use this capillary with other buffer systems, especially not with phosphate buffers (see also trouble shooting guide).
- Sample Injection:
two modes of sample injection are frequently used in capillary electrophoresis: pressure injection or electrokinetic injection. For anion analysis commonly pressure injection is chosen; the injected sample has the same composition as the material in the sample vial. When using electrokinetic injection a preconcentration of faster migrating anions occurs: the composition of the injected sample has a higher concentration of faster migrating anions than the material in the sample vial.
- Electrophoresis conditions:
| Injection: |
5 sec. at 0.5 PSI (corresponds to 30.2 mbar) |
| Separation voltage: |
- 30 kV ( [-] at the inlet side to [+] at the outlet side) |
| Detection: |
UV at 254 nm, inverted signal |
| Run time: |
5 to 15 min |
| Expected current: |
15 – 20 mA |
- Capillary treatment between single runs:
Between single runs a 3 min rinsing step with Buffer solution pH 7.7 for HPCE is sufficient.
- Capillary treatment at the end of the day:
If the capillary will be used the next day, store the capillary in Buffer solution pH 7.7 for HPCE. If the capillary is not used for longer periods, rinse the capillary with 0.1 M sodium hydroxide solution for HPCE (Product No. 72079) for 5 min, then for 2 min with Water for HPCE (Product No. 95283). Blow the capillary dry for 2 min and store the capillary dry.
Peak Identification
Single peaks are best identified by comparing the migration times with those of known anions. For final identification a standard anion should be added to the sample. The signal of the peak must be enhanced by the amount of the standard added (spiking).
Additional to the Multielement Anion HPCE Standard Solution (Product No. 29235) Fluka offers various anion standard solutions and multielement anion standard solutions for quantification or identification of detected anions. These standards are listed in the Fluka/Riedel catalogue as "Ion Chromatography Standard Solutions", "Multielement Atomic Spectroscopy Standard Solutions" and "Ionselective Electrode Standard Solutions".
Quantification
For quantification of a specific anion in your sample prepare a calibration plot from a standard solution of the anion of interest:
Analyze serial dilutions of the anion standard solution and calculate the corrected peak areas (peak area / migration time). Plot the areas versus the concentrations of the diluted standard solution (figure 7). From this calibration plot the concentration of your sample may be determined by comparing its corrected peak area with the calibration curve.
| ppm |
corr. area |
| 0 |
0 |
| 2 |
97 |
| 5 |
454 |
| 10 |
832 |
| 20 |
1720 |
Figure 7. Calibration plot of nitrate (at 0 ppm, 1 ppm, 5 ppm, 10 ppm, 20 ppm); without thiosulfate as reference anion.
For more accurate determinations, the use of a reference anion may be advantageous. This is done to omit run-to-run deviations caused by slightly variing injection amounts, buffer or run conditions. Chose a reference anion which is not contained in the samples analyzed. Add to all serial dilutions of the standard anion (e.g.: 1 ppm, 5 ppm, 10 ppm, 20 ppm nitrate) the choosen reference anion (e.g. 2 ppm thiosulfate). Add the same amount of reference anion also to your sample.
Calculate the quotients:
Plot the quotients Qstandard versus the concentrations of the diluted standard anions. From this calibration plot the concentration of an unknown sample may be determined by comparing its quotient Qsample with the calibration curve. When run-to-run variations cause problems, this calibration curve will show a better correlation coefficient (R2).
Literature
[1] Rhemrev-Boom, M. M. (1994): Determination of anions with capillary electrophoresis and indirect ultra violet detection. J. Chromatography 680(2), 675–84.
Trouble Shooting Guide
| Observation |
Matter |
Recommended Action |
| Migration times of the anions are not reproducible |
Capillary not fully reconstituted |
Rinse the capillary with 0.1 M NaOH and buffer, 20 min each. Perform 2 to 3 runs. Reproducible migration times usually are achieved after more than two runs. |
| Base line is dropping dramatically during run |
Capillary coated with components of other buffer systems |
Replace capillary and use the new capillary only for anion separations with the buffer solution pH 7.7 for HPCE |
| Very broad peaks, adjacent peaks are poorly resolved |
Sample concentration too high |
Dilute sample with water for HPCE |
| Current is dropping, no current observed |
Capillary is blocked |
Rinse with high pressure (with buffer or 0.1 M NaOH) |
| Cracked capillary |
Replace capillary |
| No peaks detected |
Polarity not reversed |
Use polarity from [-] to [+] |
Electropherogram
Separation of bromide (1), chloride (2), sulfate (3), nitrate (4), fluoride (5), and phosphate (6) each at a concentration of 6 mg/l (6 ppm).
Figure 8
back to top
Electrophoresis Home
Fluorescent Probes Home
Analytical Reagents Home