Sigma-Aldrich
Analytical / Chromatography > Analytical Reagents > Electrophoresis > HPCE
Analytical Reagents
HPCE
HPCE Buffers
IEF Markers
Anion HPCE-Kit
HPCE Buffers
  • Introduction
  • Quality Guarantee
  • Product List
Introduction
The rapidly growing capabilities of HPCE have created a great demand for reagents of an appropriate quality.
With the introduction of a series of ready-to-use buffers covering the pH range from 2.5 to 11, Fluka now meets exactly the requirements of many analysts dealing with HPCE.
Quality Guarantee
We guarantee for each reagent:
  • No insoluble impurities—the HPCE buffers are filtered through a 0.2 mm filter membrane after production
  • Minimal absorption over a wide wave length range—see figure 1 below
  • Virtually no fluorescent impurities—see figure 2 below
  • Tested for application—see figure 3 below
Figure 1 - HPCE Electrophoresis: UV/VIS absorption of the buffer solution pH 2.5 for HPCE in the wave-length range 200-800 nm.
Figure 1: UV/VIS absorption of the buffer solution pH 2.5 for HPCE (Product No. 82581) in the wave-length range 200-800 nm.
  Figure 2 - HPCE Electrophoresis: Residual fluorescence of the buffer solution pH 2.5 for HPCE.
Figure 2: Residual fluorescence of the buffer solution pH 2.5 for HPCE (Product No. 82581). The fluorescence is checked by exitation at 200, 230 and 260 nm. This illustrates the excellent suitability of this buffer for applications with fluorescence detection. Standard (S): 10-7M ovalene, embedded in a polymethylmethacrylate (PMMA) matrix, excited at 342 nm.
  Figure 3 - HPCE Electrophoresis: Separation of the two peptides aprotinin (A) and soybean trypsin inhibitor (STI).
Figure 3: Separation of the two peptides aprotinin (A) and soybean trypsin inhibitor (STI). Sample: containing 50 ng/ml of each peptide. Separation buffer: 20 nm sodium citrate pH 2.5 (Product No. 82581). Capillary: fused silica, 60 cm in length, 50 mm i.d. Injection: hydrostatic 3 sec, DH =9.8 cm. Field: 20 kV. Detection: 200 nm
Product List: Buffer solutions for HPCE
Product No.
Description
Package Size
Add to
Cart
82609 Buffer Solution pH 2.5 for HPCE
[20 mM citric acid-NaOH]
50 ml/100 ml
82581 Buffer Solution pH 2.5 for HPCE
[20 mM sodium citrate]
50 ml/100 ml/500 ml
82582 Buffer Solution pH 3.0 for HPCE
[20 mM sodium citrate]
50 ml/100 ml/500 ml
82621 Buffer Solution pH 3.0 for HPCE
[Cupric electrolyte buffer pH 3.0]
special quality for the detection of cations
(alkali and alkaline earth metals and amines)
50 ml/100 ml
82622 Buffer Solution pH 3.0 for HPCE
[150 mM potassium phosphate]
100 ml/500 ml
82582 Buffer Solution pH 3.0 for HPCE
[20 mM sodium citrate]
50 ml/100 ml/500 ml
82583 Buffer Solution pH 3.5 for HPCE
[20 mM sodium citrate]
50 ml/100 ml/500 ml
82584 Buffer Solution pH 4.0 for HPCE
[20 mM sodium citrate]
50 ml/100 ml/500 ml
82585 Buffer Solution pH 4.5 for HPCE
[20 mM sodium citrate]
50 ml/100 ml/500 ml
82586 Buffer Solution pH 5.0 for HPCE
[20 mM sodium citrate]
50 ml/100 ml/500 ml
82587 Buffer Solution pH 5.5 for HPCE
[20 mM sodium citrate]
50 ml/100 ml/500 ml
82588 Buffer Solution pH 6.0 for HPCE
[20 mM sodium citrate]
50 ml/100 ml/500 ml
82589 Buffer Solution pH 6.5 for HPCE
[20 mM sodium phosphate]
50 ml/100 ml/500 ml
82591 Buffer Solution pH 7.0 for HPCE
[20 mM borate-phosphate]
50 ml/100 ml/500 ml
82614 Buffer Solution pH 7.0 with SDS f.HPCE
[100 mM boric acid/50 mM sodium phosphate, with 50 mM SDS]
special quality for Micellar Electrokinetic CapillaryChromatography (MECC)
50 ml/100 ml
82591 Buffer Solution pH 7.0 for HPCE
[20 mM sodium citrate]
50 ml/100 ml/500 ml
82636 Buffer Solution pH 7.0 for HPCE
[50 mM sodium citrate]
50 ml/100 ml
82637 Buffer Solution pH 7.0 for HPCE
[100 mM sodium citrate]
50 ml/100 ml
82592 Buffer Solution pH 7.5 for HPCE
[20 mM sodium phosphate]
50 ml/100 ml
82619 Buffer Solution pH 7.7 for HPCE
[Pyromellitic acid electrolyte buffer pH 7.7]
special quality for the detection of inorganic and low molecular weight organic acid anions
50 ml/100 ml
82593 Buffer Solution pH 8.0 for HPCE
[20 mM sodium phosphate]
50 ml/100 ml
82594 Buffer Solution pH 8.0 for HPCE
[20 mM sodium tetraborate]
50 ml/100 ml/500 ml
82615 Buffer Solution pH 8.0 with methylcellulose for HPCEM
[50 mM TRIS-borate/ 2.5 mM EDTA, 0.5% methylcellulose]
special quality for separation of DNA restrictions fragments
50 ml/100 ml
82633 Buffer Solution pH 8.0 for HPCE
[50 mM sodium borate]
50 ml/100 ml
82634 Buffer Solution pH 8.0 for HPCE
[100 mM sodium borate]
100 ml/500 ml
82601 Buffer Solution pH 8.5 for HPCE
[20 mM sodium phosphate]
50 ml/100 ml/500 ml
82602 Buffer Solution pH 8.5 for HPCE
[20 mM sodium tetraborate]
50 ml/100 ml/500 ml
82616 Buffer Solution pH 8.6 with urea for HPCE
[100 mM TRIS/100 mM boric acid/2 mM EDTA/7 M urea]
special quality for separation of nucleic acids
50 ml/100 ml
82603 Buffer Solution pH 9.0 for HPCE
[20 mM sodium phosphate]
50 ml/100 ml/500 ml
82604 Buffer Solution pH 9.0 for HPCE
[20 mM sodium tetraborate]
50 ml/100 ml/500 ml
82605 Buffer Solution pH 9.5 for HPCE
[20 mM sodium phosphate]
50 ml/100 ml/500 ml
82606 Buffer Solution pH 10.0 for HPCE
[20 mM CAPS]
50 ml/100 ml/500 ml
82607 Buffer Solution pH 10.5 for HPCE
[20 mM CAPS]
50 ml/100 ml/500 ml
82608 Buffer Solution pH 11.0 for HPCE
[20 mM CAPS]
50 ml/100 ml/500 ml
82617 Buffer Solution pH 11.0 for HPCE
[20 mM glycine-NaOH]
50 ml/100 ml
84428 Hydrochloric acid Solution for HPCE 100 ml/500 ml
72079 Sodium hydroxide Solution for HPCE 100 ml/500 ml
95283 Water for HPCE 250 ml/1 l
back to top
IEF Markers
  • Application
  • Product Description
  • Analysis Conditions
  • Product List
  • Directions
  • Reference
Application
IEF (Isoelectric Focusing) is a powerful analytical tool for the separation of ampholytes, mainly proteins. In order to ensure the high performance of analysis, standards of pI (pI markers) are needed. In addition to classical protein based standards, low molecular compounds were developed and successfully examined in capillary IEF and IEF-Gel electrophoresis. For capillary IEF, UV absorption is the most poular method in use. UV induced fluorescence emission is of interest if derivatizations of proteins with e.g. dansyl chloride, fluorescamine, o-phtaldialdehyde or coumarin moieties are used to increase sensitivity. Another advantage for IEF gel electrophoresis with Fluorescent IEF-Marker is the possibility to control the formation of gradient without further staining (using illumination UV).
Fluorescent IEF markers can also be detected by UV-absorption at 280 nm (20°C), allthough the signal is not as strong as with fluorescence detection. The absorption maxima of the individual markers are between 308 and 350 nm. For fluorescence detection an excitation wavelength of 310 nm (individual excitation maxima: 310 to 400 nm) is suggested; the emission maximum of the individual markers lies between 410 and 500 nm.
Product Description
Solids:
  • 1 mg packages
  • Fluorescenz: Emmax and Exc. (see table 1)
  • Storage at 4°C
Stock Solution:
  • 200 ml packages
  • Fluorescenz: Emmax, Exc. and conditions (see table 1)
  • Concentration: 1, 2, 3 mg /ml
  • Solution: aqueous (see table 1)
  • Filtered through 0.45 mm membrane filter
  • Store at 4°C; stable for at least 6 months
Analysis Conditions for CIEF using Pressure Mobilization
  • Capillary: neutral capillary
  • Anolyte: 91 mM phosphoric acide in gel buffer
  • Catholyte: 20 mM sodium hydroxide in water
  • Detection: 280 nm
  • Temperature: 20 °C
  • Injection: 20 psi, 1 min
  • Polarity: inlet anode, outlet cathode
  • Focussing voltage: 500 V/cm
  • Focussing time: 2 min
  • Mobilization: 0.5 psi, 500 V/cm anolyte -> catholyte (Mobilization should be stopped after the last marker is eluted to avoid the filling of the capillary with anolyte.)
Example for CE-IEF with fluorescent pI Markers
Figure 4. Example for CE-IEF with fluorescent pI Markers
pI Marker:
Peak 1: 8.7
Peak 2: 7.6
Peak 3: 6.6
Peak 4: 6.2
The emission spectrum and emission maximum of the Fluorescent IEF-Marker pI 7.6
Figure 5. The emission spectrum and emission maximum of the Fluorescent IEF-Marker pI 7.6
The UV spectrum of the Fluorescent IEF-Marker pI 7.6 (three different concentration)
Figure 6. The UV spectrum of the Fluorescent IEF-Marker pI 7.6 (three different concentration)
Table 1. Conditions of Fluorescent IEF Markers stock solutions.
Additional information on Fluorescent IEF Markers in chapter Fluorescent Probes.
Catalog Number Description pl Stock Solution Concentration in ultrapure water Fluorescence
Marker [mg/ml] HCl [mM] 2-propanol (%) Emmax [nm] Exc. [nm] Buffer for measurement pH=pl
35096
74169
solid stock solution 2.1 3 - 50 430 340 50mM Citrate, 50mM KCl
17952
72172
solid stock solution 3.0 2 5 50 440 360 0.1M Citrate
17953
40677
solid stock solution 3.5 2 5 50 415 318 0.1M Citrate
17954
89827
solid stock solution 4.0 1 - - 415 310 0.1M Citrate
17955
89149
solid stock solution 4.5 1 5 - 424 336 0.1M Citrate
17956
89478
solid stock solution 5.1 2 10 - 415 330 0.1M Citrate
17957
77866
solid stock solution 5.5 3 15 - 412 325 0.1M Citrate
17958
73938
solid stock solution 6.2 1 10 - 500 394 0.1M Phosphate
17959
73376
solid stock solution 6.6 1 10 - 500 396 0.1M Phosphate
17961
89508
solid stock solution 6.8 1 5 - 418 338 0.1M Phosphate
17962
89951
solid stock solution 7.2 1 4 - 500 387 0.1M Phosphate
17963
89952
solid stock solution 7.6 1 10 - 495 385 0.1M Tris
17964
75734
solid stock solution 8.1 3 - - 420 340 0.1M Tris
17966
89357
solid stock solution 8.7 1 10 - 500 390 0.1M Tris
17967
90699
solid stock solution 9.0 1 10 - 495 385 0.1M Tris
46276
89268
solid stock solution 9.5 3 8 - 415 325 0.1M Carbonate
17968
77672
solid stock solution 10.3 1 10 - 495 388 0.1M Carbonate
17951
stock solution of Marker-Mix - 2 (of each) 5 50 - - -
Directions for the use of Fluorescent IEF-Markers
The protocol for the use of Fluorescent IEF-Marker is following: Prepare the IEF-marker stock solution by dissolving the substance in the solvent according to Table 1. Sonicate for up to 10 min if necessary. Store at 4 °C (the solution is stable for approximately 6 months). For use in HPCE this stock solution should be diluted 1:100 in a suitable ampholyte solution (2% ampholyte 3-10; e.g. 10046). For uses in IEF-gels the stock solution can be loaded directly onto the gel (max. 1 ml per lane).
Reference
M. Horka, Th. Willimann, M. Blum, P. Nording, Z.Friedl, K. Slais, Capillary isoelectric focusing with UV-induced fluorescence detection, J. of Chromatography A, 916, 65-71 (2001).
back to top
Anion HPCE-Kit – Product No. 29244
  • General Information
  • Contents
  • Application
  • Working Instructions
  • Peak Identification
  • Quantification
  • Literature
  • Trouble Shooting Guide
  • Electropherogramm
General Information
Capillary electrophoresis is an efficient analytical separation technique for analysis of minute amounts of sample and has several advantages, including fast separation and high resolution. The Fluka pH 7.7 buffer system is designed for the analysis of anions by indirect UV detection with reversed electro-osmotic flow. This buffer was validated for the analysis of eight common anions: fluoride, chloride, bromide, sulfate, nitrate, nitrite, thiosulfate and phosphate [1].
Contents
  • 100 ml Buffer solution pH 7.7 for HPCE (Product No. 82619)
    [Pyromellitic acid electrolyte buffer pH 7.7; composition: 2.25 mM pyromellitic acid, 6.50 mM sodium hydroxide, 0.75 mM hexamethonium hydroxide, 1.60 mM triethanolamine,
    pH: 7.7+/- 0.2 (250C); manufactured under clean room conditions; filtered through a 0.2 mm filter]
  • 100 ml Water for HPCE (Product No. 95283)
    [manufactured under clean room conditions; filtered through a 0.2 mm filter]
  • 10 ml Multielement Anion HPCE Standard Solution (Product No. 29235; 100 ppm)
    [composition: 100 mg/l fluoride, 100 mg/l bromide, 100 mg/l chloride, 100 mg/l phosphate, 100 mg/l sulfate, 100 mg/l nitrate; manufactured from highly pure salts and water; filtered through a 0.2 mm filter]
    When handling buffer solution pH 7.7 avoid contact with eyes or skin. Wear safety glasses. Store tightly closed.
  • Needed but not supplied in this kit:
    0.1 M sodium hydroxide solution for HPCE (Product No. 72079) or an equivalent quality.
Application
  • The buffer solution pH 7.7 for HPCE is supplied ready to use. No dilutions or other treatments are required.
  • The Multielement Anion Standard is supplied as a 100 ppm concentrate (0.1 g/l) of the six anions. Dilute the Multielement Anion Standard with Water for HPCE to your desired concentration (typically 1 ppm to 10 ppm). This working standard solution may be used as capillary test solution or for peak identification and quantification.
  • Sample preparation:
    Samples are either used directly or diluted with water for HPCE (Product No. 95283) to a concentration of approximately 1 to 10 mg/l of the anion of interest. Detection limits are approximately at 0.5 mg/l of the anion of interest.
Working Instructions
  • Capillary treatment:
    For anion separation, use a fused silica capillary with an inner diameter of 50-75 mm, 50-60 cm total length (40-50 cm from inlet to detector). Rinse the capillary with 0.1 M sodium hydroxide solution for HPCE (Product No. 72079) for at least 10 min, then for 10 to 20 min with buffer solution pH 7.7. Do not use this capillary with other buffer systems, especially not with phosphate buffers (see also trouble shooting guide).
  • Sample Injection:
    two modes of sample injection are frequently used in capillary electrophoresis: pressure injection or electrokinetic injection. For anion analysis commonly pressure injection is chosen; the injected sample has the same composition as the material in the sample vial. When using electrokinetic injection a preconcentration of faster migrating anions occurs: the composition of the injected sample has a higher concentration of faster migrating anions than the material in the sample vial.
  • Electrophoresis conditions:
    Injection: 5 sec. at 0.5 PSI (corresponds to 30.2 mbar)
    Separation voltage: - 30 kV ( [-] at the inlet side to [+] at the outlet side)
    Detection: UV at 254 nm, inverted signal
    Run time: 5 to 15 min
    Expected current: 15 – 20 mA
  • Capillary treatment between single runs:
    Between single runs a 3 min rinsing step with Buffer solution pH 7.7 for HPCE is sufficient.
  • Capillary treatment at the end of the day:
    If the capillary will be used the next day, store the capillary in Buffer solution pH 7.7 for HPCE. If the capillary is not used for longer periods, rinse the capillary with 0.1 M sodium hydroxide solution for HPCE (Product No. 72079) for 5 min, then for 2 min with Water for HPCE (Product No. 95283). Blow the capillary dry for 2 min and store the capillary dry.
Peak Identification
Single peaks are best identified by comparing the migration times with those of known anions. For final identification a standard anion should be added to the sample. The signal of the peak must be enhanced by the amount of the standard added (spiking).
Additional to the Multielement Anion HPCE Standard Solution (Product No. 29235) Fluka offers various anion standard solutions and multielement anion standard solutions for quantification or identification of detected anions. These standards are listed in the Fluka/Riedel catalogue as "Ion Chromatography Standard Solutions", "Multielement Atomic Spectroscopy Standard Solutions" and "Ionselective Electrode Standard Solutions".
Quantification
For quantification of a specific anion in your sample prepare a calibration plot from a standard solution of the anion of interest:
Analyze serial dilutions of the anion standard solution and calculate the corrected peak areas (peak area / migration time). Plot the areas versus the concentrations of the diluted standard solution (figure 7). From this calibration plot the concentration of your sample may be determined by comparing its corrected peak area with the calibration curve.
figure1_hpce_kit
ppm
corr. area
0
0
2
97
5
454
10
832
20
1720
Figure 7. Calibration plot of nitrate (at 0 ppm, 1 ppm, 5 ppm, 10 ppm, 20 ppm); without thiosulfate as reference anion.
For more accurate determinations, the use of a reference anion may be advantageous. This is done to omit run-to-run deviations caused by slightly variing injection amounts, buffer or run conditions. Chose a reference anion which is not contained in the samples analyzed. Add to all serial dilutions of the standard anion (e.g.: 1 ppm, 5 ppm, 10 ppm, 20 ppm nitrate) the choosen reference anion (e.g. 2 ppm thiosulfate). Add the same amount of reference anion also to your sample.
Calculate the quotients:
HPCE Kit Equation Image
Plot the quotients Qstandard versus the concentrations of the diluted standard anions. From this calibration plot the concentration of an unknown sample may be determined by comparing its quotient Qsample with the calibration curve. When run-to-run variations cause problems, this calibration curve will show a better correlation coefficient (R2).
Literature
[1] Rhemrev-Boom, M. M. (1994): Determination of anions with capillary electrophoresis and indirect ultra violet detection. J. Chromatography 680(2), 675–84.
Trouble Shooting Guide
Observation Matter Recommended Action
Migration times of the anions are not reproducible Capillary not fully reconstituted Rinse the capillary with 0.1 M NaOH and buffer, 20 min each. Perform 2 to 3 runs. Reproducible migration times usually are achieved after more than two runs.
Base line is dropping dramatically during run Capillary coated with components of other buffer systems Replace capillary and use the new capillary only for anion separations with the buffer solution pH 7.7 for HPCE
Very broad peaks, adjacent peaks are poorly resolved Sample concentration too high Dilute sample with water for HPCE
Current is dropping, no current observed Capillary is blocked Rinse with high pressure (with buffer or 0.1 M NaOH)
Cracked capillary Replace capillary
No peaks detected Polarity not reversed Use polarity from [-] to [+]
Electropherogram
Separation of bromide (1), chloride (2), sulfate (3), nitrate (4), fluoride (5), and phosphate (6) each at a concentration of 6 mg/l (6 ppm).
Electropherogram:  Separation of bromide (1), chloride (2), sulfate (3), nitrate (4), fluoride (5), and phosphate (6)
Figure 8
back to top
Electrophoresis Home
Fluorescent Probes Home
Analytical Reagents Home