Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Food Analysis
Food Analysis
Fluka Riedel Banner
logo_new1
Bioactive Nutrient Explorer
During the last decade, food protection from potentially hazardous residues has become a major topic of public interests. Food regulation authorities in the European Community and Switzerland produced proposals for restrictions of unwanted residues in food. Fluka can contribute to this trend by offering a comprehensive product range of speciality standards designed for the food and beverage industry. The product range includes unique items such as bisphenol-A-diglycidyl ether(BADGE) and bisphenol-F-diglycidyl ether standards (BFDGE) as well as food irradiation- and phytoestrogene standards.
Please take a look at one of the following topics:
  • Determination of Acrylamide, Testkit
  • Standards for food analysis: BADGE, BFDGE
  • Flavors and Fragrances (SAFC)
  • Food Irradiation
  • Phytoestrogens
  • GMO Certified Reference Materials from IRMM
  • See also:
    Kits and Reagents for Food Analysis
    Ready-to-use Kit for Acrylamide Analysis
    Last year the Swedish National Food Administration published its concerns regarding the high concentrations of acrylamide detected in foods processed at high temperatures. This discovery was particularly disturbing since the acrylamide-containing food stuff included products that are consumed on a regular basis at rather large quantities, such as potato chips, French fries, roast potatoes, breakfast cereals and crisp bread.
    The most common method for the determination of acrylamide in food is the Grob-Method. This method requires several high quality reagents and standards that are not available from the same supplier. Fluka has combined efforts and is now presenting a ready-to-use kit that makes acrylamide determination faster and easier. The Acrylamide Kit (Cat. No 72615) contains all the necessary standards and solvents for the extraction and accurate quantification of acrylamide as proposed by Grob1.
    Your convenient solution: Acrylamide Kit
    It was an exciting challenge for us to provide you with a ready-to-use kit for fast and easy determination of acrylamide. The Kit (Cat. No 72615) contains all reagents and standards needed for 12 determinations. The suitability of all 8 reagents was thoroughly tested. The concentrations and specifications of the Acrylamide-, Acrylamide-D3- and Methacrylamide-Standard Solutions were specially tailored for this method which reduces the time required significantly. The kit contains all the instructions required for sample preparation. Table 1 provides an overview of the kit and the reagents, their specifications and pack sizes. Please click here to download the method (168 KB).
    Composition of Acrylamide Kit (Cat. No 72615)
    Component
    Specification
    Pack Size
    Acrylamide Standard Solution 500 ppm in Acetonitrile
    5 ml (Certan® vial)
    Acrylamide-D3 Standard Solution 500 ppm in Acetonitrile
    5 ml (Certan® vial)
    Methylacrylamide Standard Solution 500 ppm in Acetonitrile
    5 ml (Certan® vial)
    Butyramide Standard Solution 25 ppm in Acetonitrile
    5 ml (Certan® vial)
    1-Propanol puriss.p.a., >99.5 % (GC)
    250 ml glass bottle
    n-Hexane puriss., >99.0 % (GC)
    100 ml glass bottle
    Acetonitrile for residue analysis, >99.9 % (GC)
    50 ml glass bottle
    Oil tested to determine Acrylamide
    according to Grob
    50 ml glass bottle
    Table 1: Components of Acrylamide Kit (Cat. No 72615). Standard solutions are available as kit only and can not be ordered individually. All components last for 12 determinations.
    Literature References
    1 M. Biedermann, S. Biedermann-Brem, A. Noti and K. Grob; P. Egli and H. Mändli; Mitt. Lebensm. Hyg. 2002, 93, 638-652
    Back to Top
    Standards for food analysis: BADGE, BFDGE
    Epoxy based resins are widely used for lacquer coatings on the inside of food cans and food storage vessels. BADGE and/or BFDGE are normally added to these coatings. Studies have shown that the hydrochloric media in the edible oil reacts with the epoxy based resins of the coating, resulting in the corresponding HCl-adducts, of which its toxicities are unknown. BADGE and/or BFDGE are added to the coating in order to prevent these reactions by reacting with hydrochloric acid itself.
    There has been performed considerable research in Europe to develop and apply methods of analysis for the food analysis standards. In 2000 the European Scientific Committee for Food produced a proposal for a limit of 1 mg/kg for BADGE, BFDGE (and their adducts) as well as hydrolysates in canned food. For performing a reliable analysis, standards of BADGE, BFDGE and its derivatives are needed. Fluka offers 9 unique, carefully synthesized standards including: BADGE, BFDGE, the HCl-adducts and hydrolysates (see Table 1). There is a large target group of people working with these standards, including all analytical chemists within the food industry (control laboratories), can producing companies, governmental laboratories, private analytical service laboratories and university laboratories.
    Table 1. Standards for Food Analysis: BADGE, BFDGE
    Catalog No.
    Product Name
    CAS No.
    Description
    Package Size
    15136
    Bisphenol A bis(3-chloro-2-hydroxypropyl) ether
    4809-35-2
    purum p.a. qualitative standard; >99.0% (HPLC)
    250 mg, 1g
    15137
    Bisphenol A bis(2,3-dihydroxypropyl)ether
    5581-32-8
    purum p.a. qualitative standard; >97.0% (HPLC)
    250 mg
    92427
    Bisphenol A (3-chloro-2-hydroxypropyl) (2,3-dihydroxypropyl) ether
    227947-06-0
    purum p.a. qualitative standard; >98.0% (HPLC)
    25 mg, 100 mg
    73124
    Bisphenol A (3-chloro-2-hydroxypropyl) glycidyl ether
    13836-48-1
    purum p.a. qualitative standard; ~95% (HPLC)
    25 mg, 100 mg
    15138
    Bisphenol A diglycidyl ether, standardfor quantification
    1675-54-3
    purum p.a. qualitative standard; >97.0% (GC)
    500 mg
    73417
    Bisphenol A (2,3-dihydroxypropyl) glycidyl ether
    76002-91-0
    purum p.a. qualitative standard; ~97% (HPLC)
    25 mg, 100 mg
    15139
    Bisphenol F bis(3-chloro-2-hydroxy-propyl) ether
    -
    p.a. qualitative standard; mixture of 3 isomers ortho-ortho, ortho-para, para-para; ~95% (HPLC) total assay of the 3 isomers
    250 mg
    15142
    Bisphenol F bis(2,3-dihydroxypropyl)ether
    72406-26-9
    purum p.a. qualitative standard; mixture of 3 isomers ortho-ortho, ortho-para, para-para; >97.0% (HPLC) total assay of the 3 isomers
    250 mg
    15144
    Bisphenol F diglycidyl ether
    2095-03-6
    purum p.a. qualitative standard; mixture of 3 isomers ortho-ortho, ortho-para, para-para; ~97% (GC) total assay of the 3 isomers
    500 mg
    Back to Top
    Food Irradiation
    Treatment of food-stuff with ionized radiation is used to eliminate microbiological activity. Such food-stuff, with a few exceptions, cannot be sold in certain European countries and need to be declared in many other countries. Irradiation produces certain substances which are absent in untreated food stuff and have been used as indicators of irradiation. Irradiation of fat-containing foodstuff (e.g. chicken meat, hazelnut) produces free radicals, which partially degrade fat (triglycerides). This process produces alkanes, alkenes, aldehydes and alkylcyclobutanones. Irradiation of water-containing albuminous foodstuffs (meat) produces reactive hydroxyradicals, which react with phenylalanine to produce ortho- and meta-tyrosine. Detection of typical radiation byproducts is used to prove treatment with ionized radiation. Fluka offers selected indicator substances which can be used as standards for GC-analysis (Table 2).
    Table 2. Standards for the Detection of Food Irradiation
    Catalog No.
    Product Name
    CAS No.
    Description
    Package Size
    52206
    1,7-Hexadecadiene
    125110-62-5
    ~ 95% (GC), standard for food analysis
    5 mg, 25 mg
    87141
    1,7-Tetradecadiene
    150767-46-7
    ~ 95% (GC), standard for food analysis
    5 mg, 25 mg
    44197
    2-Dodecylcyclobutanone
    35493-46-0
    > 95% (GC), standard for food analysis
    5 mg, 25 mg
    93851
    DL-o-Tyrosine
    2370-61-8
    ~ 98% (NT), standard for food analysis
    100 mg
    93853
    DL-m-Tyrosine
    775-06-4
    > 98.0% (NT), standard for food analysis
    100 mg
    Back to Top
    Phytoestrogens
    It is well established, that plants contain substances with estrogenic effects, so called phytoestrogens. These substances have been found in numerous plant food products, e.g. from carrots, peas, lentil, rice, cabbage and soy. They are structurally similar to the mammalian estrogen or estradiol, and show estrogenic properties. However, their estrogenic activity is generally much less than that of human estrogens (estrogenic activity ranges from 1/500 to 1/1000 of the activity of oestradiol). Hence phytoestrogens can act as anti-oestrogenic agents by blocking the oestrogen receptors and exerting a much weaker oestrogenic effect compared with the hormone. As a consequence it has been suggested that they might partly suppress or inhibit normal oestrogenic activity in oestrogen-responsive tissues such as breast tissue and may reduce the risk of breast cancer. Fluka offers a large number of selected pure flavones, flavonones and isoflavones for the examination of phytoestrogenic effects and analysis of Phytoestrogene contents (see Table 3)
    Table 3. Standards for the Detection of Phytoestrogens
    Catalog No.
    Product Name
    CAS No.
    Description
    Package Size
    00017
    Acacetin
    480-44-4
    BioChemika; >99.0% (TLC)
    25 mg, 100 mg
    10798
    Apigenin
    520-36-5
    BioChemika; ~98% (HPLC)
    25mg, 100 mg
    14385
    Biochanin A
    491-80-5
    BioChemika; >97.0% (TLC)
    100 mg, 1g
    27214
    Chrysin
    480-40-0
    BioChemika; ~98% (TLC)
    100 mg, 1g
    30405
    Daidzein
    486-66-8
    purum; >98.0% (TLC)
    5mg, 25 mg
    30408
    Daidzin
    552-66-9
    BioChemika; ~97% (HPLC)
    1mg, 5mg
    38763
    4',7-Dimethoxyisoflavone
    1157-39-7
    BioChemika; ~98% (TLC)
    1mg, 5mg
    45405
    Equol
    94105-90-5
    BioChemika; >99.0% (TLC )
    1mg, 5mg
    46340
    Fisetin
    528-48-3
    BioChemika; >99.0% (TLC)
    5 mg, 100 mg
    48754
    Genistein 4',7-dimethyl ether
    34086-51-6
    BioChemika; ~95% (TLC)
    1mg, 5mg
    48756
    Genistin
    529-59-9
    BioChemika; >99.0% (TLC)
    1mg, 5mg
    60010
    Kaempferol
    520-18-3
    BioChemika; >96% (HPLC)
    25mg, 100 mg
    75375
    Ononin
    486-62-4
    BioChemika; >99.0% (TLC)
    1mg, 5mg
    80614
    Pinostrobin
    480-37-5
    BioChemika; >99.0% (TLC)
    5mg, 25 mg
    82415
    Prunetin
    552-59-0
    BioChemika; >99.0% (TLC)
    5mg, 25 mg
    82435
    Puerarin
    3681-99-0
    BioChemika; >99.0% (TLC)
    1mg, 5mg
    83370
    Quercetin Dihydrate
    6151-25-3
    BioChemika; ~99% (HPLC)
    25g, 100 g
    85447
    Sissostrin
    5928-26-7
    BioChemika; ~95% (TLC)
    1 mg
    91953
    3',4',7-Trihydroxyisoflavone
    485-63-2
    BioChemika; >99.0% (TLC)
    1mg, 5mg
    91955
    4',5,7-Trihydroxyisoflavone
    485-63-2
    BioChemika; >98.0% (HPLC)
    25mg, 100 mg
    91956
    4',6,7-Trihydroxyisoflavone
    17817-31-1
    BioChemika; ~98% (TLC)
    1mg, 5mg
    92182
    4',6,7-Trimethoxyisoflavone
    798-61-8
    BioChemika; >99.0% (TLC)
    1mg, 5mg
    96093
    Zearalenone
    17924-92-4
    BioChemika; >98.0% (TLC)
    5mg, 25 mg
    GMO Certified Reference Material from IRMM
    The Certified Reference Materials (CRMs) for detection of material derived from genetically modified plants (GMOs) were developed and produced by IRMM (Inst. for Reference Materials and Measurement, Geel, Belgium) on behalf of Fluka GmbH (Buchs, Switzerland) in the frame of a collaboration with the Environment Institute (EI) of the Joint Research Centre of the European Comission (Ispra, Italy). They consist of dried flour containing different mass fractions of powder from genetically modified soya beans or maize kernels respectively.
    New Generation of CRMs for Roundup ReadyTM Soya:
    Five CRMs of dried soya bean powder with different mass fractions (0 nominal (< 0.1), 0.5, 1, 2 and 5 %) of genetically modified "Roundup ReadyTM" soya are available now. These CRMs, IRMM-410S, are replacing IRMM-410R, which was sold out in January 2002. In contrary to IRMM-410R, IRMM-410S has been produced with the help of dry-mixing techniques (3rd generation) in order to minimise DNA and protein degradation occurring during the production. The reference material has been produced from whole seeds of a non-modified soya line (Asgrow line A1900) and genetically modified event 40-3-2 Roundup ReadyTM soya (Asgrow line AG5602 RR), both delivered by Monsanto, St.Louis, MO, USA.
    The non-GMO raw material used for the production con-tains traces of Roundup ReadyTM soya GMO below 0.03 %. IRMM-410S is intended to be used for the validation of methods for the detection of genetically modified food and is available in the form of glass bottles containing 1 g of soya bean powder packed under argon atmosphere.
    Table 4. Roundup ReadyTM Soya Standards
    Catalog No.
    Product Name
    Description
    Package Size
    83063
    Soya Bean Powder, Cert. Ref. Material IRMM-410S-0 Replacement for 53198
    0 % (nominal, < 0.03%) genetically modified RRTM soya
    1 g
    72647
    Soya Bean Powder, Cert. Ref. Material IRMM-410S-1 Replacement for 49232
    0.1 % genetically modified RRTM soya
    1 g
    81751
    Soya Bean Powder, Cert. Ref. Material IRMM-410S-2 Replacement for 57452
    0.5 % genetically modified RRTM soya
    1 g
    76913
    Soya Bean Powder, Cert. Ref. Material IRMM-410S-3 Replacement for 68202
    1 % genetically modified RRTM soya
    1 g
    78767
    Soya Bean Powder, Cert. Ref. Material IRMM-410S-4 Replacement for 49215
    2 % genetically modified RRTM soya
    1 g
    93109
    Soya Bean Powder, Cert. Ref. Material IRMM-410S-5 Replacement for 44386
    5 % genetically modified RRTM soya
    1 g
    94162
    Soya Bean Powder set, Cert. Ref. Material IRMM-410S Replacement for 89305
    0, 0.1, 0.5, 1, 2 and 5 % genetically modified RRTM soya
    1 set
    Back to Top
    Table 5. Maize MON 810 Standards
    Catalog No.
    Product Name
    Description
    Package Size
    77119
    IRMM-413-0 for 0% MON 810 0% (nominal) genetically modified MON 810 maize
    1 g
    74411
    IRMM-413-1 for 0.1% MON 810 0.1% genetically modified MON 810 maize
    1 g
    79521
    IRMM-413-2 for 0.5% MON 810 0.5% genetically modified MON 810 maize
    1 g
    71981
    IRMM-413-3 for 1% MON 810 1% genetically modified MON 810 maize
    1 g
    79112
    IRMM-413-4 for 2% MON 810 2% genetically modified MON 810 maize
    1 g
    76182
    IRMM-413-5 for 5% MON 810 5% genetically modified MON 810 maize
    1 g
    78761
    IRMM-413-Set for 0%, 0.1%, 0.5%, 1%, 2% and 5% MON 810 -
    set
    Back to Top
    Table 6. IRMM Certified Standards for Maize Bt-11 and Maize Bt-176
    Catalog No.
    Product
    Pack Size
    Maize Bt-176
    03828
    Maize GMO Standard IRMM 411R, Set for 0%, 0.1%, 0.5%, 1%, 2%, 5% Bt-176
    1 set
    91528
    Maize GMO Standard IRMM 411R-0, 0% Bt-176
    1 g
    91516
    Maize GMO Standard IRMM 411R-1, 0.1% Bt-176
    1 g
    14724
    Maize GMO Standard IRMM 411R-2, 0.5% Bt-176
    1 g
    95772
    Maize GMO Standard IRMM 411R-3, 1% Bt-176
    1 g
    89092
    Maize GMO Standard IRMM 411R-4, 2% Bt-176
    1 g
    19074
    Maize GMO Standard IRMM 411R-5, 5% Bt-176
    1 g
    Maize Bt-11
    03831
    Maize GMO Standard IRMM 412R, Set for 0%, 0.1%, 0.5%, 1%, 2%, 5% Bt-11
    1 set
    91524
    Maize GMO Standard IRMM 412R-0, 0% Bt-11
    1 g
    54703
    Maize GMO Standard IRMM 412R-1, 0.1% Bt-11
    1 g
    52492
    Maize GMO Standard IRMM 412R-2, 0.5% Bt-11
    1 g
    19577
    Maize GMO Standard IRMM 412R-3, 1% Bt-11
    1 g
    19558
    Maize GMO Standard IRMM 412R-4, 2% Bt-11
    1 g
    65944
    Maize GMO Standard IRMM 412R-5, 5% Bt-11
    1 g
    Back to Top
    GMO Qualitative Standards
    In April 2004 the legislation regarding the labelling of genetically modified foods and feed products changed. This new legislation requires that even highly modified foods such as lecithin, starch and oil, as well as feed products containing GMOs are labelled clearly as such.Therefore, maize and canola are now subject to the new labelling legislation, requiring more stringent control and labelling than was previously required.
    However, in order to measure the presence or absence of GMOs a suitable standard is required. Fluka is now presenting a new range of GMO Maize Standards (NK603-, GA21- and CBH-3511 Starlink varieties) and Canola (RT73-, OXY235-, LibertyLink Falcon®- or MS8xRf3- varieties). These will enable you to easily identify manipulated material
    There are numerous benefits to be gained from the use of Fluka GMO standards:
    - The process of DNA extraction for use as a positive control can be avoided, thereby eliminating the need for time-consuming manual extraction procedures.
    - Since the DNA is extracted from plant material, the application of these standards is no longer limited to a certain sequence information or protocol. Therefore, it is possible to use them with all DNA–based methods and even with your own optimized protocols
    - The world wide availability of GMO standards having the same characteristics and composition can ensure the standardization of measurements and procedures, therefore allowing comparison of results obtained in different facilities.
    Table 7. GMO Qualitative Standards
    Catalog No.
     Product
    55231
    Canola GMO Genomic DNA Standard
    <<PDF>>
    69407
    Maiz GMO Genomic DNA Standard
    <<PDF>>
    Back to Top
    Back to Analytical