Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Microscopy
Embedding Media
Fluka is pleased to offer an extended range of embedding media kits and ready-made mounting media for both light and electron microscopy. Due to the increasing importance of fluorescence microscopy non-fluorescent aqueous mounting media that delay fluorescence fading have been introduced. PVA-NPD is the medium of choice for AMCA or Cascade blue-labeled tissue sections and cell cultures. Either PVA-NPD or PVA-DABCO are recommended for FITC, TRITC, DTAF, Cy-3 labeled sections or cell cultures.
Fluka2003_05
Prod. No.
Mounting/Embedding media and immersion oils
10951
Araldite M
10952
Araldite M Accelerator 960,accelerator DY 060
10953
Araldite M Hardener 964
31184
D.E.R.(R) 330
31185
D.E.R.(R) 332
31190
D.E.R.(R) 732
31191
D.E.R.(R) 736
30424
Dammar Resin*
33581
Benzoyl peroxide moistened with water(H2O ~25%)
43910
Divinylbenzene, mixture of isom., stab.
44160
Dodecenylsuccinic anhydride, forelectron microscopy
44581
DPX Mountant for histology*
44610
Durcupan(R) ACM Fluka, set for 1 lembedding mixture
44611
Durcupan(R) ACM Fluka, component A/Mepoxy resin
44612
Durcupan(R) ACM Fluka, component Bhardener 964
44613
Durcupan(R) ACM Fluka, component Caccelerator 960
44614
Durcupan(R) ACM Fluka, component Dplasticiser
44602
Durcupan(R) water-soluble Fluka, singlecomp. A, pge 100 ml
44600
Durcupan(R) water-soluble Fluka, setfor min. 10 embedd.
44604
Durcupan(R) water soluble Fluka,single component B
44591
Durcupan(R) water-soluble Fluka,single component C
45345
Epoxy Embedding Medium
45348
Epoxy Embedding Medium, Accelerator,DMP 30
45346
Epoxy Embedding Medium, Hardener DDSA
45347
Epoxy Embedding Medium, Hardener MNA
45359
Epoxy Embedding Medium Kit
03808
Ethylene glycol dimethacrylate, stab.
03989
Eukitt(R) quick-hardening mounting me-dium, f. microscopy*
49927
Glycerol-Gelatin, package with 15 ml*
50586
Glycol methacrylate Kit, water soluble
53402
Histocryl embedding resin*
17238
HPMA Embedding Kit
56821
Immersion oil fluorescence-free forfluoresc.microscopy
56822
Immersion oil for microscopy
10976
Immersion oil, for microscopy, droppingbottle with 20 ml
60610
Canada balsam, for microscopy*
03984
Canada balsam Solution, for microscopy*
60895
Colophony
62636
Lowicryl HM20 reagent kit, formicroscopy*
62637
Lowicryl HM23 reagent kit, formicroscopy*
62640
Lowicryl K11M reagent kit, formicroscopy*
62639
Lowicryl K4M reagent kit, formicroscopy*
62660
LR Accelerator*
10972
LR Gold Embedding Kit*
62659
LR Gold resin*
62663
LR Catalyst*
62662
LR White Embedding Kit
10973
LR White Embedding Kit, modified*
10971
LR White resin ready-to-use*
62661
LR White resin*
64050
Methacrylic acid, monomer, stab.
64112
Butyl methacrylate, stab.
64180
Hydroxypropyl methacrylate, mixture ofisomers, stab.
64200
Methyl methacrylate, stab. with0.0025% hydroquinone
74378
2-Nonen-1-ylsuccinic anhydride, forelectron microscopy
74432
Nonylphenyl-polyethyleneglycol acetate,for histology*
03987
Paraffin wax, for histology, largelumps, sp 55-600C
76257
Paraplast regular, for electron micros-copy*
76258
Paraplast plus, for electron microscopy*
76259
Paraplast x-tra, for electron microscopy*
81190
Polyethylene glycol 1'000
81188
Polyethylene glycol 1'000*
81150
Polyethylene glycol 200
10981
Polyvinyl alcohol mounting medium withDABCO, antifading*
10979
Polyvinyl alcohol mounting medium,antifading*
85960
Styrene monomer stab.
90403
Vestopal
94885
Vestopal(R) 310
94886
Vestopal Kit
94956
Vinylcyclohexene dioxide, mixture ofisomers
96090
Cedar oil, for microscopy
Back to top
Back to Microscopy home
Back to Analytical home
Back to Bioscience home