Sigma-Aldrich
Analytical / Chromatography > Analytical Reagents > Trace Analysis
Analytical Reagents
Trace Analysis
Fluka® offers high-purity reagents, solvents, and standards for inorganic trace analysis, and for the determination of organic residues in food & environmental applications. As a supplier of analytical reagents and standards, quality assurance is of prime importance to Fluka. We employ a comprehensive Quality Assurance System according to EN 29001 and ISO 9001. Each of our products is manufactured to clear, guaranteed specifications. The large variety of modern analytical methods (e.g., ICP-OES, ICP-MS, AAS) allows for specifically tailored control and analysis procedures for each product.
Inorganic Trace Analysis
Organic Residue Analysis
Search Tools
Product Literature
TraceCERT Inorganic CRMs brochure
TraceCERT® Inorganic CRMs brochure
Inorganic Trace Analysis brochure
Inorganic Trace Analysis brochure
(Reagents and Standards for Sample Preparation, Calibration and Analysis)
Inorganic Trace Analysis
Sensitive trace analysis applications (AAS, ICP-AES, polarography, ion chromatography, etc.) require extremely pure sample preparation reagents to assure low blank values. Digestion reagents cannot contain metal ions or other impurities. Further, complete decomposition of the sample is required in order to achieve reproducible and accurate elemental results by analytical instrumental methods.
Fluka’s portfolio of high-purity and ultra-high purity reagents and solvents for contamination-free sample preparation includes the TraceSELECT® and TraceSELECT® Ultra lines of acids, bases, salts and solvents.
  • Ultra High Purity Reagents (TraceSELECT Ultra)
  • High Purity Acids for Trace Analysis (TraceSELECT)
  • High Purity Bases for Trace Analysis (TraceSELECT)
  • High Purity Reagents for Trace Analysis
  • High Purity Salts for Trace Analysis
  • Solvents for Metal Speciation Analysis
High quality analytical reference standards for reproducible, traceable measurements include:
  • TraceCERT® certified reference materials for ICP, AAS and IC
  • AAS Reference Materials
  • ICP-OES/-MS Reference Materials
  • Ion Chromatography CRMs
  • Proficiency Testing Studies by Sigma-Aldrich RTC
Request a Quote for Custom Standards or Custom Packaging
Organic Residue Analysis
Organic residue testing entails the analysis of persistent organic pollutants, and highly toxic compounds, down to ultratrace levels in food and environmental applications.  Most of the organic pollutants identified in environmental analysis are extracted using solvents, from matrices such as food, water or soil. The high purity solvents, additives and standards provided by Fluka ensure low detection levels and reproducible results for sample preparation; as well as, for separation and analysis of organic contaminants in biologicals, soil, sludge, water, and air matrices. In food and environmental applications, the focus is on substances such as dioxins, polychlorinated biphenyls (PCBs), and pesticides.
High-purity Solvents & Standards for sample preparation and analysis of trace levels of organic pollutants:
  • Solvents for Organic Residue Analysis
  • TraceCERT Organic Certified Reference Materials
  • Food Residue Analysis Standards
  • Standards for Environmental Applications
  • Environmental Matrix CRMs
  • Pesticides & Metabolites
  • GMO Reference Materials, Qualitative
  • Mycotoxin Standards
  • Proficiency Testing Studies by Sigma-Aldrich RTC
Request a Quote for Custom Standards or Custom Packaging
Use the Applications Search feature to find chromatograms and method conditions for GC, HPLC, SPE and SPME applications involving organic trace analysis.
Search Tools
Perform a quick search for a suitable analytical reagent / solvent or an analytical standard using our dedicated search tools.
Use the Applications Search feature to find chromatograms and method conditions for GC, HPLC, SPE and SPME applications.
Analytical Reagents Search
Search for High-Purity Reagents & Solvents
Analytical Standards Search
Search for Chemical Reference Standards, CRMs, PT
Product Literature
  • Overview of Trace Analysis Reagents (Presentation PDF)
  • New High Purity TraceSELECT Reagents for Voltammetry and Ion Chromatography (to be published; Analytix 1/2012, p. 19; PDF)
  • High Purity Reagents for AAS - auxiliary reagents for interference-free measurements (Analytix 1/2011, p. 14; PDF)
  • High Purity Salts for melting digestion in environmental, water, and food analysis (Analytix 5/2010, p. 22; PDF)
  • High Purity Digestion Reagents for Ultra-Trace Analysis - Ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis (Analytix 4/2010, p. 10; PDF)