Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Trace Analysis
Dissolution Reagents
Dissolution Reagents for Sample Preparation: Mercury and Arsenic-free Reagents
Sample preparation is an important, basic method in the analysis of inorganic and organic specimens. Acid dissolution, with or without an oxidizing agent has proved to be an efficient, commonly used procedure for the decomposition of solids, sediments and sludges.
In general, strong inorganic acids are employed such as nitric, perchloric, hydrochloric and sulfuric acid. In most cases the dissolution process requires more than one acid e.g. aqua regia, Leffort (Lunge) mixture, or the Dixon mixture in soil analysis with a certain, predominating acid, determining the rate and completeness of the decomposition.
Mercury and arsenic-free reagents are particularly suitable for the determination of those metal ions, in the presence of others, in e.g. industrial and waste waters, polluted soils, and sludge. They can be used for sample preparation (dissolution) and measuring the prepared samples with ICP-OES or –MS
Acid, Acid Mixture
Additive
Metal to be analyzed in matrix
Matrix to be analyzed
Method
HCl / HNO3
-
As, Hg
Waste sludge, soils, sediments, rocks
Extraction
HNO3 / H2SO4
Br2
As
Pb, Cu, Zn, Mo ores, concentrates
Dissolution
HNO3 / H2SO4 (2:1)
-
As
Soils
Decomposition
HNO3 / H2SO4
KMnO4
Hg
Soils
Decomposition
HNO3 / H2SO4
KMNO4
Hg
Water, waste-water, sludge
DIN 38406
HNO3 / H2O2
-
As
Water, wast-water, sludge
DIN 38406
1. HNO3 / HClO4
2. KMnO4 / K2S2O8
-
As
Materials rich in organic compounds
Two-step decomposition
1. HNO3 / HClO4
2. HCl
-
As
Sulfidic minerals
Decomposition
H2SO4
-
As
Allemontite
Dissolution
View Product List and Ordering information
Back to Trace Analysis home
Back to Analytical home