Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Analytical / Chromatography > Analytical Reagents > Trace Analysis > Electrochemistry Reagents
Trace Analysis
Electrochemistry Reagents
Supporting Electrolytes for Electrochemistry
Within the range of electroanalytical methods, specially polarograhic and voltammetric methods have gained more and more importance. The range of application has increased considerably and includes trace analysis of metals and organic compounds, as well as environmental analytic chemistry, pharmacy and medicine. Supporting electrolytes are extensively employed in many areas of electrochemistry. Tetraalkylammonium salts are very suitable because of their excellent solubility characteristics in most organic solvents.
Fluka offers specially purified tetraalkylammoniium salts, tested for the absence of impurities, which would lead to undesirable residual currents.
Catalog No.
Product Name
Assay
Cas No.
Package Size
17045
Tetrabutylammonium fluorosulfate
> 98
88504-81-8
1g, 5g
86606
Tetraethylammonium bromide
> 99.0
71-91-0
5 g, 25 g
86616
Tetraethylammonium chloride
> 99.0
56-34-8
5 g, 25 g
86618
Tetraethylammonium tetrafluoroborate
> 99.0
429-06-1
5 g, 25 g
86624
Tetraethylammonium hydrogen phthalate
> 99.0
41606-94-4
5 g, 25 g
86625
Tetraethylammonium hexafluorophosphate
> 99.0
429-07-2
5 g, 25 g
86638
Tetraethylammonium iodide
> 99.0
68-05-3
5 g, 25 g
86643
Tetraethylammonium nitrate
> 99.0
1941-26-0
5 g, 25 g
86646
Tetraethylammonium perchlorate
> 99.0
2567-83-1
5 g, 25 g
86653
Tetraethylammonium trifluoromethanesulfonate
> 99.0
35895-69-3
5 g, 25 g
86654
Tetraethylammonium toluene-4-sulfonate
> 99.0
733-44-8
5 g, 25 g
86835
Tetrabutylammonium acetate
> 99.0
10534-59-5
5 g, 25 g
86836
Tetrabutylammonium bromide
> 99.0
1643-19-2
10 g, 50 g
86837
Tetrabutylammonium benzoate
> 99.0
18819-89-1
5 g, 25 g
86838
Tetrabutylammonium bis-trifluoromethane-sulfonimidate
> 99.0
210230-40-3
1 g, 5 g
86879
Tetrabutylammonium hexafluorophosphate
> 99.0
3109-63-5
25 g, 100 g
86893
Tetrabutylammonium perchlorate
> 99.0
1923-70-2
10 g, 50 g
86895
Tetrabutylammonium trifluoromethane sulfonate
> 99.0
35895-70-6
5 g, 25 g
86896
Tetrabutylammonium tetrafluoroborate
> 99.0
429-42-5
5 g, 25 g
86897
Tetrabutylammonium tetraphenylborate
> 99.0
15522-59-5
5 g, 25 g
86912
Tetrabutylammonium iodide
> 99.0
311-28-4
5 g, 25 g
86913
Tetrabutylammonium nitrate
> 99.0
1941-27-1
5 g, 25 g
86916
Tetrabutylammonium toluene-4-sulfonate
> 99.0
7182-86-7
5 g, 25 g
86927
Tetrabutylphosphonium hexafluorophosphate
> 99.0
111928-21-3
5 g, 25 g
86934
Tetrabutylphosphonium tetrafluoroborate
> 99.0
1813-60-1
5 g, 25 g
87259
Tetraethylammonium benzoate
> 99.0
16909-22-1
5 g, 25 g
87267
Tetraethylammonium bis-trifluoromethane-sulfonimidate
> 99.0
161401-26-9
1 g, 5 g
87311
Tetrahexylammonium perchlorate
> 99.0
4656-81-9
5 g, 25 g
87697
Tetramethylammonium benzoate
> 99.0
25255-90-7
5 g, 25 g
87709
Tetramethylammonium bromide
> 99.0
64-20-0
5 g, 25 g
87719
Tetramethylammonium chloride
> 99.0
75-57-0
5 g, 25 g
87722
Tetramethylammonium hydrogen phthalate
> 99.0
79723-02-7
5 g, 25 g
87737
Tetramethylammonium nitrate
> 99.0
1941-24-8
5 g, 25 g
87753
Tetramethylammonium perchlorate
> 99.0
2537-36-2
5 g, 25 g
88013
Tetrapentylammonium perchlorate
> 99.0
4328-09-0
5 g, 25 g
88104
Tetrapropylammonium bromide
> 99.0
1941-30-6
5 g, 25 g
88121
Tetrapropylammonium perchlorate
> 99.0
15780-02-6
5 g, 25 g
Back to Top
Back to Trace Analysis home
Back to Analytical home