Sigma-Aldrich
Analytical / Chromatography > Analytical Reagents > Trace Analysis > Ion Chromatography
Trace Analysis
Ion Chromatography
Certified Standards for Ion Chromatography
  • Multiion Standards (PRIMUS)
  • Anion Standards (BAM and Sigma-Aldrich certified)
  • TraceCERT® Standards for IC *new*
Certified Eluent Concentrates
Water for Ion Chromatography
Standards for Ion Chromatography
  • Cation Standards
  • Anion Standards
  • Multiion Standards
  • Eluent concentrates
  • Organic Acid Standards
Certified Standards for Demanding Ion Chromatography Applications
PRIMUS (Primary Multiions Standards for Ion Chromatography) - A Corporate Project of:
Sigma Aldrich logo
PRIMUS
The need of using CRM's
Today, Ion Chromatography (IC) is an important technique within the analytical laboratory. It is used for the aqueous determination of ions down to the ppt range, being one of the best methods that can provide quantitative analysis of ions at this level. Moreover, the development of modern IC apparatus and columns makes this technique a fast, uncomplicated and reliable tool.
The quality of IC standards had to be improved to keep up with the advances in the available hardware. The selection of appropriate standards for the calibration of the IC system is important. Fluka already offers a wide range of IC standards, starting from single elements for anion and cation analytic to multielement standards. These standards are frequently used by the Environmental, Food and Beverage industries. However, other industries like Pharmaceutical or Semiconductor, may need to work with even higher qualifications and certified standards to comply with GMP. This trend is furthermore reinforced by official regulations of the European Community within the guideline 96/23/EG on the use of Certified Reference Materials (CRM):
...In paragraph 3.1.1.2., the determination of trueness (one component of accuracy) is described: trueness can only be established by means of CRM's. A CRM should be used whenever available] (1).
The American FDA (Food and Drug Administration) demands in their Inspection Guide for analytic methods the usage of reference standards.
ISO (International Organization for Standardization) has published more then 6 papers about reference materials.
CRM's are guaranteed as validation standards. This means that they have to be traceable to a SI unit and standard deviation has to be determined. In accordance to ISO Guide 17025 (2), the preparation of a CRM ideally is carried out by metrological institutes such as NIST (National Institute of Standards and Technology), EMPA (Swiss Federal Laboratories for Materials Testing and Research) or BAM in Germany (Federal Institute for Materials Research and Testing).
Single- and Multielement Standards developed in collaboration with EMPA and BAM
For Ion Chromatography, the number of CRM's available is still small. In order to fill this gap, Fluka launched a few months ago their first multiion CRM's for Ion Chromatography (Table 1).
The development was carried out in collaboration with EMPA, located in St. Gallen, Switzerland. Based on the competence of this highly regarded metrological institute, the traceability to SI is guaranteed and standard deviation is determined. Therefore, these CRM's are produced completely at EMPA, giving you the guarantee of certification you desire.
Following a positive customer response, we were also asked to produce certified multielement standards and we are pleased to offer two new certified multielement standard solutions, PRIMUS (Primary Multiion Standards) (Table 2).
High precision weighing and high purity salts for highest quality
EMPA determines the purity of the starting materials based on high purity salts (Table 2). The content of these primary materials is determined by two methods for each salt. The analysis is done in collaboration with BAM. The results are verified and cross-checked at state-of-the-art levels to ensure the quality you require. The really novel part is the production procedure. First, the solid salts are determined and then they are dissolved in high purity water. Although it sounds simple, one of the core competences of EMPA is the extremely precise and accurate weighing of these salts. This allows us to offer our customers CRM's standards having a very low content of ion species (10mg/kg for each ion). To ensure the quality of our products, each bottle is delivered in a sealed aluminium bag to minimise the loss of water and to keep the concentration stable over more than two years. All standards are delivered with a Certificate of Analysis, including a detailed report with the calculation of the measurement uncertainty.
Did you know that ...
... each new Metrohm IC apparatus contains a pack of our PRIMUS multication and
multianion standards?
Metrohm highly recommends the use of PRIMUS standards for the internal calibration and validation of their instruments.
References
  1. Commission decision of 12 August 2002 implementing Council Directive 96/23/EC concerning the performance of analytical methods and the interpretation of results. Official Journal of the European Communities, L 221/8
  2. ISO Guides 30 (1992): Terms and definitions used in connection with reference materials; 31(2000): Reference materials - contents of certificates and labels; 32 (1997): Calibration in analytical chemistry and use of certified reference materials; 33 (2000): Uses of certified reference materials; 34 (2000): General requirements for the competence of reference material producers; 35 (1989): Certification of reference materials - General statistical principles
www.sigma-aldrich.com/certified_standards
Table 1: PRIMUS: Multielement Standard Solutions for Ion Chromatography (10 mg/kg)
Catalog No.
Product Name
Ions
Produced from
Example Certificate
89316
Certified Multication Standard Solution Lithium, Sodium, Potassium, Magnesium, Calcium Li2CO3, NaCl, KCl, MgO, CaCO3
PDF
89886
Certified Multianion Standard Solution Fluoride, Chloride, Bromide, Nitrate, Phosphate, Sulfate NaF, NaCl, NaBr, NaNO3, Na2SO4, Na2HPO4
PDF
Back to Top
PRIMUS standards supplied in 50 mL HDPE bottles. A detailed certificate is provided with each bottle including:
SI traceable content, uncertainty, trace impurities and expiry date.
Table 2: These anionic standard solutions for Ion Chromatography (1000 mg/L) are
certified in a cooperation project with BAM and Sigma-Aldrich
Catalog No.
Product Name
Produced from
Example Certificate
87969
Bromide NaBr
PDF
87603
Chloride NaCl
PDF
80373
Fluoride NaF
PDF
86576
Nitrate NaNO3
PDF
81193
Phosphate Na2HPO4
PDF
80218
Sulfate Na2SO4
PDF
Back to Top
These certified standards are prepared with high-purity salts and water and are supplied in 100 mL HDPE bottles.
TraceCERT® Standards for IC
click here for Background Information
Cat. No.
Ion Composition
Example Certificate
Additional Information
59755
Ammonium NH4Cl in water
replaces cat. no. 95023
43147
Bromide NaBr in water
replaces cat. no. 17355
39883
Chloride NaCl in water
replaces cat. no. 24889
40121
Chromate KCrO4 in water
replaces cat. no. 27072
90157
Cyanide      
77365
Fluoride NaF in water
replaces cat. no. 47070
41271
Iodide KI in water
new product
74246
Nitrate NaNO3 in water
replaces cat. no. 72547
67276
Nitrite NaNO2 in water
replaces cat. no. 77012
38364
Phosphate Na2HPO4 in water
replaces cat. no. 79409
90071
Sulfate Na22SO4 in water
replaces cat. no. 86126
Back to Top
Certified Eluent Concentrate for Ion Chromatography
Our new certified eluent concentrates for ion chromatography are traceable by potentiometric titration
to NIST Standard Reference Material. It is certified in accordance with ISO Guide 31. All details about
exact content, uncertainty, traceability and expiry date are described in the certificate.
Table 1: TraceCERT® calibration standards for IC
Cat. No.
Product Name Concentration (mMole)
Example Certificate
50972
2,6-pyridinedicarboxylic acid 20
50433
DL-tartaric acid 100
55517
Methanesulfonic acid 100
16355
Nitric acid 100
50439
Perchloric acid 10
61699
Potassium hydroxide 100
36486
Sodium bicarbonate 100
56169
Sodium carbonate 100
43617
Sodium hydroxide 100
68279
Sulfuric acid 100
Back to Top
Packaging: 1L HDPE bottles with 45mm screw thread
Water for Ion Chromatography
Fluka has developed a water quality especially for the needs of ion chromatography. This water is
also suitable for trace analysis of anions, cations and also some organics which are usually analyzed
by IC.
The new product 00612 is available in 2.5L and 5L package size and specified as follows:
Organic Traces:
Acetate, formiate, glycolate, oxalate: ≤ 10 µg/kg each
Cation Traces:
Al, Ba, Bi, Cd, Co, Cr, Cu, Fe, Li, Mg, Mn, Mo, Ni, Pb, Sr, Zn: ≤ 5 µg/kg each. Ca, K, Na: ≤ 10 µg/kg
each NH4+: ≤ 50µg/kg
Anion Traces:
Bromide, chloride, fluoride, iodide, nitrate, nitrite, phosphate, sulfate: 1 µg/kg each
Conductivity:
≤ 2 µS/cm
Catalog No.
Product Name
Pack Size
00612
Water for Ion Chromatography
2.5 L, 5L
Packaging: 2.5L and 5L HDPE bottles with 45mm thread
Back to Top
Standards for Ion Chromatography
Ion chromatographic standard solutions require slightly different specification than spectroscopic solutions:
  • The solution is filtered through a 0.45 mm membrane filter to prevent the column from clogging
  • pH > 3
All the solutions are carefully prepared from high-purity salts, preferably nitrates; their concentration is 1000 mg/L. For packaging high-density polyethylene bottles are used. On each label the expiry date of the solution is stated.
Cation Standards for Ion Chromatography (1000mg/L)
Catalog No.
Product Name
CAS No.
Additional information
95023
Ammonium
12125-02-9
replaced by 59755
11719
Barium
10022-31-8
14203
Beryllium
-
20892
Cadmium
10325-94-7
21041
Calcium
10124-37-5
20951
Cesium
7647-17-8
27570
Cobalt
10141-05-6
61151
Copper
3251-23-8
15316
Lead
10099-74-8
62369
Lithium
7790-69-4
63041
Magnesium
10377-60-3
63533
Manganese
10377-66-9
72224
Nickel
13138-45-9
100%37
Potassium
7757-79-1
71167
Sodium
7631-99-4
85889
Strontium
10042-76-9
96456
Zinc
7779-88-6
Back to Top
IC standards supplied in 100 mL HDPE bottles.
Matrix: water or 0.001M nitric acid solution.
Anion Standards for Ion Chromatography (1000 mg/L)
Catalog No.
Product Name
CAS No.
Additional information
17355
Bromide
7647-15-6
replaced by 43147
24889
Chloride
7647-14-5
replaced by 39883
27072
Chromate
7789-00-6
replaced by 40121
28491
Cyanide
151-50-8
47070
Fluoride
7681-49-4
replaced by 77365
72547
Nitrate
7631-99-4
replaced by 74246
77012
Nitrite
-
replaced by 67276
79409
Phosphate
7778-77-0
replaced by 38364
86126
Sulfate
7757-82-6
replaced by 90071
Back to Top
IC standards supplied in 100 mL HDPE bottles. Matrix: water
Multiion Standards for Ion Chromatography
Catalog No.
Product Name
Ion Concentration
CAS No.
87029
Multielement Anion Br (20mg/kg), Cl(10mg/kg), F(3mg/kg), NO3(20mg/kg), PO4(30mg/kg), SO4(20mg/kg)
-
63597
Multielement I Br (1000mg/L), Cl(1000mg/L), F(1000mg/L)
-
63598
Multielement II NO3(1000mg/L), PO4(1000mg/L), SO4(1000mg/L)
-
Back to Top
IC standards supplied in 100 mL HDPE bottles. Matrix: water
Eluent Concentrates for Ion Chromatography
Catalog No.
Product Name
Pack Size
CAS No.
72064
Sodium hydroxide solution, eluent, 50-52% 500 mL
1310-73-2
86854
Tetrabutylammonium hydroxide, ~40% 100 mL, 500mL, 2.5L
2052-49-5
Back to Top
Organic Acid Standards for Ion Chromatography (1000mg/L)
Due to the omnipresence of organic acids in foods and beverages such as fruit drinks and wines and their link to certain nutritional, quality and safety aspects of the food, interest in measuring their concentration is growing rapidly. Ion chromatography is a common method for the analysis of organic acids.
To meet the requirements of our analytical customers, we developed a group of organic acids standards for ion chromatography (Table 1). The concentration is 1.000 g/L in water. All standards are carefully prepared from high purity starting materials, dissolved in water 17749 for analytical usage, filtered through a 0.22µm membrane filter to prevent column fouling and clogging. All standards packaged in high density polyethylene bottles under sterile conditions. An expiration date is clearly provided on each bottle.
Catalog No.
Brand
IC Standard
13669
Fluka Acetate
38730
Fluka Citrate
49897
Fluka Formate
49323
Fluka Malonate
04621
Fluka Oxalate
04924
Fluka Phthalate
02717
Fluka Propionate
50824
Fluka Tartrate
Back to Top
IC standards supplied in 100 mL HDPE bottles.
Back to Top
Back to Trace Analysis home
Back to Analytical home