|
Trace Analysis
Ion Standard Solutions
|
|
|
Atomic Absorption Spectroscopy
ICP-OES / ICP-MS Spectroscopy
View all AAS & ICP products in the Online Catalog
Atomic Absorption Spectroscopy
| Standard Solutions (1000 mg/kg) |
| Cat.No. |
Brand |
Product |
Medium |
Package |
| 06154 |
Fluka |
Aluminum |
0.05 M HCl, stab. With 0.4% HCHO |
100 ml, 500 ml |
| 06155 |
Fluka |
Aluminum |
~0.5 M HNO3 |
100 ml, 500 ml |
| 10773 |
Fluka |
Antimony |
~ 5 M HCl |
100 ml, 500 ml |
| 11082 |
Fluka |
Arsenic |
~ 0.05 M HNO3 + NaOH |
100 ml, 500 ml |
| 206962 |
Aldrich |
Arsenic |
2% KOH |
100 ml, 500 ml |
| 11722 |
Fluka |
Barium |
H2O |
100 ml, 500 ml |
| 11723 |
Fluka |
Barium |
~ 0.5 M HNO3 |
100 ml, 500 ml |
| 206970 |
Aldrich |
Barium |
1 wt. % HCl |
100 ml, 500 ml |
| 14205 |
Fluka |
Beryllium |
~ 0.5 M HNO3 |
100 ml, 500 ml |
| 206989 |
Aldrich |
Beryllium |
1 wt. % HCl |
100 ml, 500 ml |
| 95375 |
Fluka |
Bismuth |
~ 0.5 M HNO3 |
100 ml, 500 ml |
| 15579 |
Fluka |
Boron |
H2O |
100 ml, 500 ml |
| 20895 |
Fluka |
Cadmium |
~ 0.5 M HNO3 |
100 ml, 500 ml |
| 21049 |
Fluka |
Calcium |
~ 0.5 M HCl |
100 ml, 500 ml |
| 21058 |
Fluka |
Calcium |
~ 0.5 M HNO3 |
100 ml, 500 ml |
| 22236 |
Fluka |
Cer |
~ 0.5 M HNO3 |
100 ml, 500 ml |
| 20958 |
Fluka |
Cesium |
~ 0.5 M HNO3 |
100 ml, 500 ml |
| 20952 |
Fluka |
Cesium |
H2O |
100 ml, 500 ml |
| 27067 |
Fluka |
Chromium(III) |
~ 0.05 M HNO3 |
100 ml, 500 ml |
| 207055 |
Aldrich |
Chromium(III) |
1 wt. % HCl |
100 ml, 500 ml |
| 27068 |
Fluka |
Chromium(VI) |
H2O |
100 ml, 500 ml |
| 292044 |
Aldrich |
Chromium(VI) |
1 wt. % HCl |
100 ml, 500 ml |
| 60785 |
Fluka |
Cobalt |
~ 0.05 M HNO3 |
100 ml, 500 ml |
| 61147 |
Fluka |
Copper |
~ 0.05 M HNO3 |
100 ml, 500 ml |
| 207128 |
Aldrich |
Europium |
1 wt. % HNO3 |
100 ml, 500 ml |
| 207136 |
Aldrich |
Gadolinium |
1 wt. % HNO3 |
100 ml, 500 ml |
| 48341 |
Fluka |
Gallium |
~ 0.5 M HNO3 |
100 ml |
| 48843 |
Fluka |
Germanium |
~ 1 M HCl |
100 ml, 500 ml |
| 50766 |
Fluka |
Gold |
~1 M HCl |
100 ml |
| 57081 |
Fluka |
Indium |
~ 0.5 M HNO3 |
100 ml |
| 58195 |
Fluka |
Iridium |
~1 M HCl |
100 ml |
| 44903 |
Fluka |
Iron |
~ 0.05 M HNO3 |
100 ml, 500 ml |
| 305952 |
Aldrich |
Iron |
1 wt. % HCl |
100 ml, 500 ml |
| 207225 |
Aldrich |
Lanthanum |
1 wt. % HNO3 |
100 ml, 500 ml |
| 15314 |
Fluka |
Lead |
~ 0.5 M HNO3 |
100 ml, 500 ml |
| 62372 |
Fluka |
Lithium |
H2O |
100 ml, 500 ml |
| 62373 |
Fluka |
Lithium |
~ 0.5 M HNO3 |
100 ml, 500 ml |
| 207241 |
Aldrich |
Lithium |
1 wt. % HCl |
100 ml, 500 ml |
| 62701 |
Fluka |
Lutetium |
~ 0.5 M HNO3 |
100 ml |
| 207268 |
Aldrich |
Lutetium |
1 wt. % HNO3 |
100 ml |
| 63046 |
Fluka |
Magnesium |
~ 0.05 M HCl, stab. With 0.04% HCHO |
100 ml, 500 ml |
| 63044 |
Fluka |
Magnesium |
~ 0.5 M HNO3 |
100 ml, 500 ml |
| 63534 |
Fluka |
Manganese |
~ 0.5 M HNO3 |
100 ml, 500 ml |
| 83357 |
Fluka |
Mercury |
~ 0.5 M HNO3 |
100 ml, 500 ml |
| 69859 |
Fluka |
Molybdenum |
~ 0.5 NH4OH |
100 ml, 500 ml |
| 69876 |
Fluka |
Molybdenum |
~ 0.5 M HNO3 |
100 ml, 500 ml |
| 207306 |
Aldrich |
Molybdenum |
water |
100 ml, 500 ml |
| 207314 |
Aldrich |
Neodymium |
1 wt. % HNO3 |
100 ml, 500 ml |
| 72223 |
Fluka |
Nickel |
~ 0.05 M HNO3 |
100 ml, 500 ml |
| 274917 |
Aldrich |
Niobium |
water |
100 ml, 500 ml |
| 75612 |
Fluka |
Osmium |
~ 1 M HCl |
100 ml |
| 207349 |
Aldrich |
Palladium |
5% HCl |
100 ml, 500 ml |
| 76035 |
Fluka |
Palladium. |
~ 1 M HNO3 |
100 ml |
| 207357 |
Aldrich |
Phosphorus |
water |
100 ml, 500 ml |
| 80964 |
Fluka |
Platinum |
~ 2 M HCl |
100 ml |
| 60032 |
Fluka |
Potassium |
H2O |
100 ml, 500 ml |
| 60029 |
Fluka |
Potassium |
~ 0.5 M HNO3 |
100 ml, 500 ml |
| 316946 |
Aldrich |
Potassium |
1 wt. % HCl |
100 ml, 500 ml |
| 83672 |
Fluka |
Rhenium |
~ 1 M HNO3 |
100 ml |
| 83722 |
Fluka |
Rhodium |
~ 1 M HCl |
100 ml |
| 83955 |
Fluka |
Rubidium |
~ 0.5 M HNO3 |
100 ml, 500 ml |
| 207438 |
Aldrich |
Rubidium |
1 wt. % HCl |
100 ml, 500 ml |
| 84033 |
Fluka |
Ruthenium |
~ 1 M HCl |
100 ml, 500 ml |
| 84615 |
Fluka |
Scandium |
~ 0.5 M HNO3 |
100 ml, 500 ml |
| 84896 |
Fluka |
Selenium. |
~ 0.5 M HNO3 |
100 ml, 500 ml |
| 85351 |
Fluka |
Silicium |
~ 1 M HNO3 + 2% HF |
100 ml, 500 ml |
| 316822 |
Aldrich |
Silicon |
2% NaOH |
100 ml, 500 ml |
| 85137 |
Fluka |
Silver |
~ 0.5 M HNO3 |
100 ml, 500 ml |
| 71177 |
Fluka |
Sodium |
H2O |
100 ml, 500 ml |
| 71173 |
Fluka |
Sodium |
~ 0.5 M HNO3 |
100 ml, 500 ml |
| 207500 |
Aldrich |
Sodium |
1 wt. % HCl |
100 ml, 500 ml |
| 85891 |
Fluka |
Strontium |
H2O |
100 ml, 500 ml |
| 85891 |
Fluka |
Strontium |
H2O |
100 ml, 500 ml |
| 207519 |
Aldrich |
Strontium |
1 wt. % HCl |
100 ml, 500 ml |
| 333298 |
Aldrich |
Sulfur |
water |
100 ml, 500 ml |
| 86275 |
Fluka |
Tantalum |
~ 2 M HF |
100 ml |
| 305928 |
Aldrich |
Terbium |
1 wt. % HNO3 |
100 ml, 500 ml |
| 88203 |
Fluka |
Thallium |
~ 0.5 M HNO3 |
100 ml, 500 ml |
| 207578 |
Aldrich |
Thorium |
5% HNO3 |
100 ml, 500 ml |
| 207586 |
Aldrich |
Thulium |
1 wt. % HNO3 |
100 ml, 500 ml |
| 96524 |
Fluka |
Tin |
~ 5 M HCl |
100 ml, 500 ml |
| 89475 |
Fluka |
Titanium |
~ 2 M HCl |
100 ml, 500 ml |
| 274933 |
Aldrich |
Titanium |
water |
100 ml, 500 ml |
| 207624 |
Aldrich |
Uranium |
1 wt. % HNO3 |
100 ml, 500 ml |
| 94693 |
Fluka |
Vanadium |
~ 0.05 M NH4OH |
100 ml, 500 ml |
| 94695 |
Fluka |
Vanadium |
~ 0.5 M H2SO4 |
100 ml, 500 ml |
| 94694 |
Fluka |
Vanadium |
~ 0.5 M HNO3 |
100 ml, 500 ml |
| 317608 |
Aldrich |
Vanadium |
5% HCl |
100 ml, 500 ml |
| 207640 |
Aldrich |
Ytterbium |
1 wt. % HNO3 |
100 ml, 500 ml |
| 95802 |
Fluka |
Yttrium |
~ 0.5 M HNO3 |
100 ml, 500 ml |
| 96457 |
Fluka |
Zinc |
~ 0.5 M HNO3 |
100 ml, 500 ml |
| 207667 |
Aldrich |
Zinc |
1 wt. % HCl |
100 ml, 500 ml |
| 96568 |
Fluka |
Zirconium |
~ 2 M HCl |
100 ml, 500 ml |
| Standard Solutions (10,000 mg/kg) |
| Cat.No. |
Brand
|
Product |
Medium |
Package |
| 02457 |
Fluka |
Calcium |
~ 0.5 M HNO3 |
100 ml, 500 ml |
| 02583 |
Fluka |
Iron |
~ 0.5 M HNO3 |
100 ml, 500 ml |
| 02398 |
Fluka |
Magnesium |
~ 0.5 M HNO3 |
100 ml, 500 ml |
| 02498 |
Fluka |
Potassium |
~ 0.5 M HNO3 |
100 ml, 500 ml |
| 02397 |
Fluka |
Sodium |
~ 0.5 M HNO3 |
100 ml, 500 ml |
| 02584 |
Fluka |
Zinc |
~ 0.5 M HNO3 |
100 ml, 500 ml |
| Concentrates: Each ampoule contains 1 g metal |
| Cat.No. |
Brand
|
Product |
Medium |
Package |
| 02781 |
Fluka |
Aluminum |
H2O |
1 piece |
| 02782 |
Fluka |
Antimony * |
HCl, H2O |
1 piece |
| 02783 |
Fluka |
Arsenic |
H2O |
1 piece |
| 02784 |
Fluka |
Barium |
H2O |
1 piece |
| 02786 |
Fluka |
Cadmium |
H2O |
1 piece |
| 02672 |
Fluka |
Calcium |
HCl, H2O |
1 piece |
| 02787 |
Fluka |
Cesium |
H2O |
1 piece |
| 02673 |
Fluka |
Chromium(VI) |
H2O |
1 piece |
| 02683 |
Fluka |
Cobalt |
Sulfuric acid, H2O |
1 piece |
| 02684 |
Fluka |
Copper |
HCl, H2O |
1 piece |
| 02681 |
Fluka |
Gold |
HCl, H2O |
1 piece |
| 02679 |
Fluka |
Iron* |
HCl, H2O |
1 piece |
| 02785 |
Fluka |
Lead |
H2O |
1 piece |
| 02685 |
Fluka |
Lithium |
H2O |
1 piece |
| 02656 |
Fluka |
Magnesium |
H2O |
1 piece |
| 02654 |
Fluka |
Manganese |
H2O |
1 piece |
| 02587 |
Fluka |
Mercury |
H2O |
1 piece |
| 02653 |
Fluka |
Molybdenum |
H2O |
1 piece |
| 02588 |
Fluka |
Nickel |
H2O |
1 piece |
| 03366 |
Fluka |
Phosphate |
H2O |
1 piece |
| 02682 |
Fluka |
Potassium |
H2O |
1 piece |
| 02687 |
Fluka |
Selenium |
H2O |
1 piece |
| 03793 |
Fluka |
Silicon |
H2O |
1 piece |
| 02688 |
Fluka |
Silver |
Nitric solution |
1 piece |
| 02589 |
Fluka |
Sodium |
H2O |
1 piece |
| 03794 |
Fluka |
Strontium |
H2O |
1 piece |
| 02762 |
Fluka |
Tin |
HCl |
1 piece |
| 03796 |
Fluka |
Titanium* |
H2O |
1 piece |
| 03799 |
Fluka |
Tungsten |
Sodiumhydroxide solution |
1 piece |
| 03797 |
Fluka |
Vanadium |
Sodiumhydroxide solution |
1 piece |
| 02761 |
Fluka |
Zinc |
H2O |
1 piece |
| 02763 |
Fluka |
Zirconium |
Hydrochloric acid |
1 piece |
| Concentrates: Each ampoule contains 10g metal |
| Cat.No. |
Brand
|
Product |
Medium |
Package |
| 03382 |
Fluka |
Aluminum |
H2O |
1 piece |
| 03381 |
Fluka |
Antimony |
Hydrochloric solution |
1 piece |
| 03356 |
Fluka |
Cadmium |
Hydrochloric solution |
1 piece |
| 03379 |
Fluka |
Calcium |
H2O |
1 piece |
| 03378 |
Fluka |
Chloride |
H2O |
1 piece |
| 03377 |
Fluka |
Chromium(VI) |
H2O |
1 piece |
| 03374 |
Fluka |
Cobalt |
Sulfuric solution |
1 piece |
| 03372 |
Fluka |
Copper (CuCl2.2H2O) |
Hydrochloric solution |
1 piece |
| 03373 |
Fluka |
Copper (CuSO4.5H2O) |
H2O |
1 piece |
| 03376 |
Fluka |
Iron |
H2O |
1 piece |
| 03399 |
Fluka |
Lead |
H2O |
1 piece |
| 03371 |
Fluka |
Magnesium |
H2O |
1 piece |
| 03357 |
Fluka |
Manganese |
H2O |
1 piece |
| 03368 |
Fluka |
Nickel (Ni(SO4)2.7H2O |
H2O |
1 piece |
| 03369 |
Fluka |
Nickel (NiCl2.6H2O) |
H2O |
1 piece |
| 03367 |
Fluka |
Phosphate |
H2O |
1 piece |
| 03375 |
Fluka |
Potassium |
H2O |
1 piece |
| 03370 |
Fluka |
Sodium |
H2O |
1 piece |
| 03364 |
Fluka |
Sulfate |
H2O |
1 piece |
| 03362 |
Fluka |
Zinc (ZnSO4.7H2O) |
H2O |
1 piece |
| 03363 |
Fluka |
Zinc (ZnCl2) |
Hydrochloric solution |
1 piece |
* the ready-to-use solution should be acidified with 100 ml HCl conc.
Back to top
ICP-OES/ICP-MS Spectroscopy
| Concentrates: Each ampoule contains 1 g metal |
| Cat.No. |
Brand
|
Product |
Medium |
Package |
| 02769 |
Fluka |
Boron |
H2O |
1 piece |
| 02776 |
Fluka |
Cobalt |
Nitric solution |
1 piece |
| 02777 |
Fluka |
Copper |
Nitric solution |
1 piece |
| 02772 |
Fluka |
Iron |
Nitric solution |
1 piece |
| 02778 |
Fluka |
Magnesium |
Nitric solution |
1 piece |
| 02797 |
Fluka |
Sodium |
Nitric solution |
1 piece |
| 02696 |
Fluka |
Zinc |
Nitric solution |
1 piece |
| 02794 |
Fluka |
Multielement Cd,Cr,Cu,Fe,Mn,Ni,Pb,Sb,Sn,Zn Strd Conc.,1mg each |
Nitric solution |
1 piece |
| Master Standard Solutions (1000 mg/kg) |
| Cat.No. |
Brand
|
Product |
Medium |
Package |
| 17042 |
Fluka |
Aluminum |
~ 0.5 M HNO3 |
100 ml |
| 17056 |
Fluka |
Barium |
H2O |
100 ml |
| 17033 |
Fluka |
Beryllium |
~ 0.5 M HNO3 |
100 ml |
| 17034 |
Fluka |
Bismuth |
~ 0.5 M HNO3 |
100 ml |
| 17061 |
Fluka |
Cadmium |
~ 0.5 M HNO3 |
100 ml |
| 17062 |
Fluka |
Calcium |
~ 0.5 M HNO3 |
100 ml |
| 17069 |
Fluka |
Cesium |
~ 0.5 M HNO3 |
100 ml |
| 17059 |
Fluka |
Chromium |
H2O |
100 ml |
| 17057 |
Fluka |
Cobalt |
~ 0.5 M HNO3 |
100 ml |
| 17068 |
Fluka |
Copper |
~ 0.5 M HNO3 |
100 ml |
| 17067 |
Fluka |
Gallium |
~ 0.5 M HNO3 |
100 ml |
| 17071 |
Fluka |
Indium |
~ 0.5 M HNO3 |
100 ml |
| 17066 |
Fluka |
Iron |
~ 0.5 M HNO3 |
100 ml |
| 17043 |
Fluka |
Lead |
~ 0.5 M HNO3 |
100 ml |
| 17073 |
Fluka |
Lithium |
~ 0.5 M HNO3 |
100 ml |
| 17074 |
Fluka |
Magnesium |
~ 0.5 M HNO3 |
100 ml |
| 17075 |
Fluka |
Manganese |
~ 0.5 M HNO3 |
100 ml |
| 17039 |
Fluka |
Molybdenum |
~ 0.5 M HNO3 |
100 ml |
| 17054 |
Fluka |
Nickel |
~ 0.5 M HNO3 |
100 ml |
| 17063 |
Fluka |
Platinum |
~ 2 M HCl |
100 ml |
| 17072 |
Fluka |
Potassium |
~ 0.5 M HNO3 |
100 ml |
| 17064 |
Fluka |
Rubidium |
~ 0.5 M HNO3 |
100 ml |
| 17038 |
Fluka |
Sodium |
~ 0.5 M HNO3 |
100 ml |
| 17048 |
Fluka |
Strontium |
H2O |
100 ml |
| 17051 |
Fluka |
Thallium |
~ 0.5 M HNO3 |
100 ml |
| 17065 |
Fluka |
Tin |
~ 5 M HCl |
100 ml |
| 17046 |
Fluka |
Titanium |
~ 2 M HCl |
100 ml |
| 17052 |
Fluka |
Vanadium |
~ 0.5 M HNO3 |
100 ml |
| 17053 |
Fluka |
Zinc |
~ 0.5 M HNO3 |
100 ml |
| 17044 |
Fluka |
Zirconium |
~ 2 M HCl |
100 ml |
Back to top
| Multi-Element Standard Solutions |
|
Cat.No. 70002 |
Cat.No. 70003 |
Cat.No. 70004 |
Cat.No. 70006 |
Cat.No. 70008 |
Cat.No. 70009 |
Cat.No. 70011 |
Cat.No. 70007 |
| Product |
I |
II |
III |
IV |
V |
VI |
VII |
Tuning Solution |
| Package |
100 ml |
100 ml |
100 ml |
100 ml |
100 ml |
100 ml |
100 ml |
100 ml |
| Application |
ICP-OES |
ICP-OES |
ICP-OES For analysis of drinking and waste water |
ICP-OES For analysis of drinking and waste water |
ICP-MS |
ICP-MS |
ICP-MS |
ICP-MS |
| Element |
Conc. [mg/kg] (~1M HNO3) |
Conc. [mg/kg] (~0.5M HCl) |
Conc. [mg/kg] (~5M HNO3) |
Conc. [mg/kg] (~5M HNO3) |
Conc. [mg/kg] (~5M HNO3) |
Conc. [mg/kg] (~5M HNO3) |
Conc. [mg/kg] (~7.5M HCl) |
Conc. [mg/kg] (~5M HNO3) |
| Aluminium |
50 |
|
|
40 |
10 |
|
|
|
| Arsenic |
|
|
|
40 |
|
| Barium |
10 |
|
|
40 |
10 |
|
|
|
| Beryllium |
|
10 |
|
10 |
10 |
|
|
10 |
| Bismuth |
100 |
|
|
|
10 |
|
|
10 |
| Boron |
50 |
|
|
100 |
|
| Cadmium |
10 |
|
|
10 |
10 |
|
|
|
| Cesium |
|
10 |
|
|
|
| Calcium |
10 |
|
2000 |
|
100 |
10 |
|
|
| Cer |
|
10 |
| Chromium |
50 |
|
|
20 |
10 |
|
|
|
| Cobalt |
10 |
|
|
10 |
10 |
|
|
10 |
| Copper |
10 |
|
|
20 |
10 |
|
|
|
| Gallium |
|
10 |
|
|
|
| Germanium |
|
50 |
|
10 |
|
| Gold |
|
10 |
|
| Indium |
|
10 |
|
|
10 |
| Iron |
10 |
|
|
100 |
100 |
10 |
|
|
| Lithium |
50 |
|
|
|
10 |
|
|
|
| Lead |
100 |
|
|
40 |
10 |
|
|
10 |
| Magnesium |
10 |
|
400 |
|
10 |
|
|
10 |
| Manganese |
10 |
|
|
10 |
10 |
|
|
|
| Molybdenum |
50 |
|
|
|
10 |
|
|
|
| Nickel |
50 |
|
|
20 |
10 |
|
|
|
| Phosphate |
|
| Platinum |
|
10 |
|
| Potassium |
100 |
|
200 |
|
100 |
10 |
|
|
| Rubidium |
|
10 |
|
|
|
| Selenium |
|
|
|
100 |
|
| Silver |
10 |
|
|
|
10 |
|
|
|
| Sodium |
50 |
|
1000 |
|
100 |
|
|
|
| Strontium |
10 |
|
|
|
10 |
|
|
|
| Thallium |
50 |
|
|
100 |
10 |
|
|
|
| Tin |
|
50 |
|
10 |
|
| Titanium |
|
10 |
|
10 |
|
| Vanadium |
|
50 |
|
40 |
10 |
|
|
|
| Zinc |
10 |
|
|
100 |
10 |
|
|
|
| Zirconium |
|
50 |
|
10 |
|
Back to top
Back to Trace Analysis home
Back to Spectroscopy home