|
Reagents for Trace Analysis
Sample prep for trace analysis applications requires reagents of highest purity. Specifically, they must have very low metal content to ensure appropriately low blanks. Fluka-brand TraceSELECT® and TraceSELECT®Ultra reagents from Sigma-Aldrich include high purity acids, bases and salts that are designed for trace and ultra-trace analysis. These reagents are handled under clean-room conditions and supplied in Teflon PFA bottles to ensure long-term purity.
TraceSELECT®Ultra
TraceSELECT®Ultra products are designed for the ultra-trace analysis at ppb and even ppt levels. Typical metal trace impurities are below 0.1 µg/kg (<0.1 ppb), in some cases even as low as the ppq level. To maintain purity, TraceSELECT®Ultra products are delivered in Teflon PFA bottles.
| Catalog No. |
TraceSELECT® Ultra Acids and Bases |
Content |
Package Size |
| 07692 |
Acetic acid |
≥ 99.0% T |
250mL, 1L |
| 16748 |
Ammonium hydroxide solution |
≥ 25% T |
250mL, 1L |
| 23828 |
Hydrobromic acid |
≥ 44% T |
250mL, 1L |
| 96208 |
Hydrochloric acid |
≥ 30% T |
250mL, 1L |
| 02658 |
Hydrofluoric acid |
≥ 49% acidimetric |
250mL, 1L |
| 16911 |
Hydrogen peroxide solution |
≥ 30% RT |
250mL, 1L, 5L |
| 02650 |
Nitric acid |
~ 65% T |
250mL, 1L |
| 12415 |
Perchloric acid
|
67-72% |
250mL, 1L |
| 64957 |
Phosphoric acid |
≥ 85% T |
250mL, 1L |
| 77239 |
Sulfuric acid |
≥ 95% T |
250mL, 1L |
| 14213 |
Tetramethylammonium hydroxide solution |
25% in H2O |
250mL |
| 14211 |
Water |
|
1L |
|
|
TraceSELECT®
The products of the TraceSELECT® series have been developed for sample preparation and analysis in the ppm and ppb level. The blank values for metal Traces are typically below 0.01mg/kg (0.01 ppm)
|
|
|
| Catalog No. |
TraceSELECT® Acids and Bases |
Content |
Package Size |
| 45727 |
Acetic acid |
≥ 99.0% T |
100mL, 500mL |
| 09857 |
Ammonium hydroxide solution |
≥ 25% in water |
100mL, 500mL |
| 06454 |
Formic acid |
≥ 88.0% |
250mL |
| 08256 |
Hydrochloric acid |
≥ 30% |
100mL, 500mL, 1L |
| 84415 |
Hydrochloric acid |
≥ 37% fuming T |
100mL, 500mL |
| 47559 |
Hydrofluoric acid |
47-51% AT |
100mL, 500mL |
| 95321 |
Hydrogen peroxide solution |
≥ 30% RT |
100mL, 500mL |
| 84385 |
Nitric acid |
> 69.5% T |
250mL, 500mL, 1L |
| 79614 |
Phosphoric acid |
≥ 85% T |
100mL, 500mL |
| 18498 |
Phosphorus oxychloride |
≥99.9999% |
250mL |
| 77227 |
Perchloric acid |
67-71% T |
100mL, 500mL |
| 30955 |
Potassium hydroxide solution |
≥ 30% in water T |
250mL |
| 13171 |
Sodium hydroxide solution |
≥ 30% in water T |
250mL |
| 84716 |
Sulfuric acid |
≥ 95% T |
100mL, 500mL |
| 68556 |
Tetramethylammonium hydroxide solution |
25% in H2O |
250mL |
| 95305 |
Water |
|
500ML, 1L, 2.5L, 10L |
| Catalog No. |
TraceSELECT® Salts |
Content |
Package Size |
| 73432 |
Ammonium acetate |
99.9999% |
100g |
| 09725 |
Ammonium chloride |
99.9995% |
25g, 100g |
| 09726 |
Ammonium phosphate monobasic |
99.9999% |
100g |
| 09979 |
Ammonium sulfate |
99.9999% |
100g |
| 40581 |
Cesium bromide |
99.999% |
25g |
| 90033 |
Cesium chloride |
99.9995% |
25g, 100g |
| 62462 |
Lithium carbonate |
99.998% |
25g, 100g |
| 60348 |
Potassium bisulfate |
99.999% |
25g, 100g |
| 60111 |
Potassium carbonate
|
99.995% |
50g |
| 05257 |
Potassium chloride |
99.9995% |
25g, 100g |
| 30533 |
Potassium iodide |
99.999% |
100g |
| 60216 |
Potassium phosphate monobasic |
99.995% |
25g, 100g |
| 60371 |
Potassium hydroxide hydrate |
99.995% |
25g |
| 60429 |
Potassium nitrate |
99.995% |
25g, 100g |
| 60347 |
Potassium phosphate dibasic |
99.999% |
100g |
| 59929 |
Sodium acetate |
99.999% |
25g, 100g |
| 01963 |
Sodium bromide |
99.995% |
25g |
| 71347 |
Sodium carbonate |
99.9999% |
25g, 100g |
| 38979 |
Sodium chloride |
99.999% |
25g, 100g |
| 01968 |
Sodium hydroxide monohydrate |
99.9995% |
25g, 100g |
| 71752 |
Sodium nitrate |
99.999% |
25g, 100g |
| 71629 |
Sodium phosphate dibasic |
99.999% |
100g |
| 71492 |
Sodium phosphate monobasic |
99.999% |
25g, 100g |
| 44355 |
Tin(II) chloride dihydrate |
99.999% |
250g, 2,5kg |
| Catalog No. |
TraceSELECT® Solvents |
Content |
Package Size |
| 01324 |
Acetonitrile |
≥ 99.9% |
1 L |
| 42105 |
Methanol |
≥ 99.9% |
1 L |
| 43729 |
1-Methyl-2-pyrrolidinone |
≥ 99.9% |
1 L |
| Catalog No. |
TraceSELECT® Organics |
Content |
Package Size |
| 05878 |
L-Ascorbic acid |
≥ 99.0% |
100g |
| 94068 |
Citric acid monohydrate |
≥ 99.9998% |
100g |
| 11514 |
Hydroxylamine hydrochloride |
≥ 99.9995% |
100g |
| 78513 |
D-Mannitol |
≥ 99.9999% |
50g |
| 93722 |
Oxalic acid dihydrate |
≥ 99.9999% |
100g |