|
NEW – Available NOW!
Precision Test & Calibration Gases in Disposable Cylinders Fluka Gas Standards
Portable cylinders are fully compatible with European-style fittings (7/16” 28NS/2 male)
Precision gas standards and convenient mixtures for laboratory calibration, field sampling and personnel monitoring
|
|
- Portable, convenient and easy to use
- Packaged in lightweight, non-refillable containers
- Ready-to-use mixtures eliminate the need to buy individual cylinders for each component.
- Extensive range of both standard gases and gas mixtures
- Suitable for testing and calibrating a wide range of gas detection and analysis equipment
- Compliant with all current standards and legislation for non-refillable cylinders
- Manufactured gravimetrically for maximum accuracy (± 2 ppm)
- Available in 5 L and 20 L cylinders with two valve configurations (control valve, syringe adapter)
- Single component gases and mixtures include CO, CO2, methane, ethane, ethylene, acetylene, N2, O2, H2, paraffins, BTEX, vinyl chloride and others
|
Fluka Gas Standards - Available Mixtures
|
Cat. No.
|
Product Name |
Add to Cart
|
|
|
Carbon monoxide (1%), carbon dioxide (1%), methane (1%), ethane (1%), ethylene (1%) and acetylene (0.5%) in nitrogen |
|
|
|
Carbon monoxide (5%), carbon dioxide (5%), nitrogen (5%), oxygen (2%), methane (4%) and hydrogen (2%) in helium |
|
|
|
Methane, 99.0% |
|
|
|
C1-C6 n-Paraffins |
|
|
|
Carbon monoxide, carbon dioxide, hydrogen, methane & oxygen in nitrogen (1% each) |
|
|
|
C1-C6 n-Paraffins , 100 ppm each component in nitrogen |
|
|
|
Ethylene in air 100 ppm |
|
|
|
BTEX Mix in Nitrogen (10ppm) |
|
|
|
1% Carbon Dioxide, 20% Oxygen in N2 |
|
|
|
Methane in nitrogen , 1%, |
|
|
|
Vinyl chloride in nitrogen , 10 ppm |
|
|
|
Miniflow regulator with brass control valve & flow indicator |
|
|
|
Brass Control Valve with threaded outlet |
|
|
|
Septum regulator for syringe samples |
|
Cylinder Specifications
|
?Cylinder capacity (litres)
|
5
|
20
|
|
Pressure (bar/psig)
|
8/120
|
20/300
|
|
Size (mm)
|
213 x 66
|
240 x 76
|
|
Weight (Kg)
|
0.1
|
0.2
|
|
Water capacity (litres)
|
0.68
|
0.97
|
|
Material
|
Tinplate
|
Aluminium alloy
|
|
Accuracy % or ppm nom value
|
± 5 or 2
|
± 5 or 2
|
|
Valve outlet
|
aerosol type push button dispenser
|
7/16” 28NS/2 male
|
Adapters suitable for 20 L cylinder
|
Cat. No.
|
Product Name |
Add to Cart
|
|
|
?Brass Control Valve with threaded outlet |
|
|
|
Miniflo 1 - Brass control valve & flow indicator |
|
|
|
Septaflo - Control valve for samplling with a syringe and needle for GC injection |
|
Back to Top Back to Application Area / Analytical Technique
|