Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Analytical / Chromatography > Analytical Standards  > Application Area & Technique > Spectroscopy Standards for OES and XRF
Application Area & Technique
Spectroscopy Standards for OES and XRF
MBHX-Ray Fluorescence and Optical Emission–Spectroscopy are are widely used in the metal industry and in material control for the determination of the composition and impurity profile of metals and metal alloys.
The new Sigma-Aldrich certified reference materials (CRMs) for spectroscopy are produced in the United Kingdom by MBH Ltd, a recognized manufacturer in the field of spectroscopy reference materials.
Certified values are given for the most relevant impurities for each corresponding metal or alloy. The disks are produced using commercial-purity metals and master alloys. To minimise surface effects, approximately 2mm from the cast faces were removed.
Analysis was carried out on millings taken from samples representative of the product. It was performed by a panel of laboratories mostly operating within the terms of ISO/IEC 17025, using documented standard reference methods and validated by appropriate reference materials ensuring traceability of the results.
Homogeneity was tested by checking sample and batch uniformity using an optical emission spectrometer for at least 15% of all the disks from each batch. The products are delivered together
with a detailed certificate.
Table 1: Spectroscopy Products with Certified Values
Cat. No. Description Certified Values %
97593 Steel (low Alloy) C: 0.261, Si: 0.753, S: 0.0476, P: 0.043, Mn: 1.19, Ni: 0.886, Cr: 2.09, Mo: 0.788, Cu: 0.130, Sn: 0.047, Al: 1.136, V: 0.296, W: 0.358, Co: 0.470, Nb: 0.322, Ti: 0.260, Ta: 0.26
05602 Duplex Steel C: 1.492, Si: 1.89, S: 0.0098, P: 10.97, Mn: 10.97, Ni: 3.03, Cr: 24.71, Mo: 0.086, Cu: 0.114, Al: 0.028, V: 0.126, Nb: 2.47, N: 0.478
51870 Copper-Nickel Alloy C: 0.0051, Si: 0.293, Mn: 1.00, Cu: 30.48, Fe: 1.24, Cr: 0.073, Mo: 0.023, Co: 0.029, Ti: 0.063, Al: 0.086, Mg: 0.035, S: 0.0069, P: 0.0134, Nb: 0.156, Ni: 66.58
51479 Leaded Brass Pb: 1.274, Sn: 0.0912, Zn: 35.34, Fe: 0.299, Ni: 0.282, Al: 0.384, Si: 0.022, As: 0.098, Bi: 0.0257, Sb: 0.104, P: 0.042, Co: 0.0054, Se: 0.0010, Cd: 0.0030, B: 0.0014, Cu: 62.10
56912 Aluminium Bronze Sn: 0.103, Pb: 0.266, Zn: 0.247, Fe: 4.05, Ni: 4.53, Al: 9.60, Si: 0.295, As: 0.0066, Mn: 0.056, P: 0.048, Cr: 0.0031, Mg: 0.0035, Cu: 80.70
97625 Phosphor Bronze Sn: 3.39, Pb: 1.02, Zn: 1.48, Fe: 0.555, Ni: 0.90, Al: 0.001, Si: 0.097, As: 0.199, Mn: 0.195, Bi: 0.0033, Sb: 0.470, P: 0.885, S: 0.023, Co: 0.095, Mg: 0.0035, Cu: 90.6
74808 Gunmetal Sn: 5.22, Pb: 5.13, Zn: 4.19, Fe: 0.136, Ni: 1.008, Al: 0.007, Si: 0.001, As: 0.059, Mn: 0.0018, Bi: 0.0225, Sb: 0.093, P: 0.0112, S: 0.061, Co: 0.0662, Ag: 0.0099, Cu: 83.98
66955 Copper (Impurities) Pb: 0.046, Zn: 0.0024, Fe: 0.0015, Ni: 0.0211, As: 0.0196, Sb: 0.020, Mn: 0.0023, Cd: 0.0023, Bi: 0.0202, S: 0.0056, Se: 0.120, Ag: 0.323, Au: 0.0129, Sn: 0.0037, Co: 0.0002, P: 0.0020, Al: 0.0002, In: 0.0009, Te: 0.024
96687 Galvanizing Alloy Pb: 0.0062, Mg: 0.0034, Al: 0.514, Cd: 0.0006, Fe: 0.0028, Sn: 0.0024, Cu: 0.0321, Ni: 0.0441, Mn: 0.0089, Bi: 0.0061, Sb: 0.0287, As: 0.0003, Cr: 0.0007, Co: 0.0037
95653 Zinc (Impurities) Pb: 0.0026, Mg: 0.0001, Al: 0.0008, Cd: 0.0017, Fe: 0.0077, Sn: 0.0016, Cu: 0.0018, Ni: 0.0013, Mn: 0.0005, Bi: 0.0005, Sb: 0.0006, Tl: 0.0012, In: 0.0006, Hg: 0.00
49892 Zinc-Aluminium-Copper Alloy Pb: 0.0106, Mg: 0.0828, Al: 3.89, Cd: 0.0053, Fe: 0.0081, Sn: 0.0099, Cu: 1.004, Ni: 0.0024, Mn: 0.0086, Bi: 0.0022, Sb: 0.0068, Ti: 0.0008, Cr: 0.0063, Si: 0.011
43998 Aluminium -Magnesium Alloy Cu: 0.093, Mg: 3.97, Si: 0.21, Fe: 0.41, Mn: 0.40, Ni: 0.90, Zn: 0.092, Pb: 0.107, Sn: 0.096, Ti: 0.25, Cr: 0.095, Be: 0.0074
91927 Aluminium-Silicon-Copper Alloy Cu: 1.48, Mg: 0.304, Si: 10.19, Fe: 1.001, Mn: 0.547, Ni: 0.234, Zn: 1.688, Pb: 0.292, Sn: 0.368, Ti: 1.57, Cr: 0.0106, Co: 0.0179, V: 0.0179, Be: 0.0025, Sb: 0.0042, Cd: 0.0032
67226 Tin (Impurities) As: 0.0008, Bi: 0.0025, Sb: 0.0034, Pb: 0.0113, Cu: 0.0013, Fe: 0.0008, Cd: 0.0015, Zn: 0.0034, Ni: 0.0004, Al: 0.0197, Ag: 0.0018, In: 0.0047, Au: 0.0007, Te: 0.0009, Se: 0.0005, Ga: 0.0027, Hg: 0.0044
12242 Tin-Base Solder (Lead free) As: 0.045, Bi: 0.175, Sb: 1.06, Pb: 0.119, Cu: 2.97, Fe: 0.016, Cd: 0.0072, Zn: 0.002, Ni: 0.0203, Al: 0.0008, Ag: 0.562, P: 0.018, Se: 0.0115
80901 Lead (Impurities) Sn: 0.603, Sb: 0.259, Bi: 1.186, Cu: 0.066, As: 0.155, Ag: 0.505, Cd: 0.211, In: 0.224, Ni: 0.0020, Te: 0.0011, Se: 0.0064, Au: 0.0082, Na: 0.0073, Hg: 0.089
30978 Battery Alloy with Calcium Sn: 0.899, Sb: 0.002, Bi: 0.0174, Cu: 0.0009, As: 0.0007, Ag: 0.0082, Zn: 0.0048, Cd: 0.0019, Ni: 0.0004, Al: 0.0240, Te: 0.0005, Ca: 0.0960
87610 Lead (high Alloy) Sn: 1.45, Sb: 6.05, Bi: 0.0194, Cu: 0.0291, As: 0.217, Ag: 0.0071, Fe: 0.010, Zn: 0.0007, Cd: 0.0046, Ni: 0.0062, Te: 0.0071, Se: 0.0149, S: 0.0036