Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Analytical Standards
SCOTTY® Gas Cylinder Specs
scotty 4
  SCOTTY 4
Contents: 4 Liters (0.14 cf)
Pressure: 120 psig (8 bar)
Outlet Fitting: Aerosol-type push button with applicator tube
Weight: 0.2 lbs. (approximately 100 grams)
Dimensions: 2 1/3" X 8" (63.5 mm x 203.2 mm)
U.S. D.O.T. Specs: 2Q
scotty 14
SCOTTY 14
Contents: 14 Liters (0.5 cf)
Pressure: 240 psig (17 bar)
Outlet Fitting: CGA 160 - 1/8" (3.2 mm) NPTF
Weight: 1.5 lbs. (0.7 kg)
Dimensions: 3" X 11 (76.2 mm x 279.4 mm)
U.S. D.O.T. Specs: 4B240
scotty 48
SCOTTY 48
Contents: 48 Liters (1.7 cf)
Pressure: 300 psig (21 bar)
Outlet Fitting: CGA 165
Weight: 1.75 lbs. (0.8 kg)
Dimensions: 4" X 16 1/4" (101.6 mm x 412.8 mm)
U.S. D.O.T. Specs: 39 NRC
scotty 74 el
SCOTTY 74-EL
Contents: 74 Liters (2.6 cf)
Pressure: 480 psig (33 bar)
Outlet Fitting: CGA 170 (aluminum diaphragm packless valves with stainless steel diaphragms)
Weight: 1.75 lbs. (0.8 kg)
Dimensions: 4" X 16 1/4" (101.6 mm x 412.8 mm)
U.S. D.O.T. Specs: 39 NRC
scotty 110
SCOTTY 110 (Pi-marked cylinder*)
Contents: 110 Liters (3.9 cf)
Pressure: 1800 psig (124 bar)
Outlet Fitting: CGA 180
Weight: 2.2 lbs. (1 kg)
Dimensions: 3.25" D X 11.625" H (82.6mm X 295.3mm)
U.S. D.O.T. Specs: 3AL2216
* Pi-marked cylinders comply with the requirements of the Transportable Pressure Equipment Directive (TPED) for movement within the EU