| |
SCOTTY 4 |
| |
| Contents: 4 Liters (0.14 cf) |
| Pressure: 120 psig (8 bar) |
| Outlet Fitting: Aerosol-type push button with applicator tube |
| Weight: 0.2 lbs. (approximately 100 grams) |
| Dimensions: 2 1/3" X 8" (63.5 mm x 203.2 mm) |
| U.S. D.O.T. Specs: 2Q |
|
| SCOTTY 14 |
| |
| Contents: 14 Liters (0.5 cf) |
| Pressure: 240 psig (17 bar) |
| Outlet Fitting: CGA 160 - 1/8" (3.2 mm) NPTF |
| Weight: 1.5 lbs. (0.7 kg) |
| Dimensions: 3" X 11 (76.2 mm x 279.4 mm) |
| U.S. D.O.T. Specs: 4B240 |
|
|
|
| SCOTTY 48 |
| |
| Contents: 48 Liters (1.7 cf) |
| Pressure: 300 psig (21 bar) |
| Outlet Fitting: CGA 165 |
| Weight: 1.75 lbs. (0.8 kg) |
| Dimensions: 4" X 16 1/4" (101.6 mm x 412.8 mm) |
| U.S. D.O.T. Specs: 39 NRC |
|
| SCOTTY 74-EL |
| |
| Contents: 74 Liters (2.6 cf) |
| Pressure: 480 psig (33 bar) |
| Outlet Fitting: CGA 170 (aluminum diaphragm packless valves with stainless steel diaphragms) |
| Weight: 1.75 lbs. (0.8 kg) |
| Dimensions: 4" X 16 1/4" (101.6 mm x 412.8 mm) |
| U.S. D.O.T. Specs: 39 NRC |
|
|
|
| SCOTTY 110 (Pi-marked cylinder*) |
| |
| Contents: 110 Liters (3.9 cf) |
| Pressure: 1800 psig (124 bar) |
| Outlet Fitting: CGA 180 |
| Weight: 2.2 lbs. (1 kg) |
| Dimensions: 3.25" D X 11.625" H (82.6mm X 295.3mm) |
| U.S. D.O.T. Specs: 3AL2216 |
 |
| |
| * Pi-marked cylinders comply with the requirements of the Transportable Pressure Equipment Directive (TPED) for movement within the EU |