| Certified Viscosity and Density Standards from Paragon |
| Product No. |
Description |
Dynamic Viscosity (mPa.s) at 25°C |
Pack size |
| 67348 |
Viscosity and Density Standard N 0.8 |
0.4329 |
500 ml |
| 19044 |
Viscosity and Density Standard N 1.0 |
0.9184 |
500 ml |
| 01446 |
Viscosity and Density Standard N 0.8 |
1.3115 |
500 ml |
| 93835 |
Viscosity and Density Standard N2 |
1.77 |
500 ml |
| 41859 |
Viscosity and Density Standard S 3 |
3.251 |
500 ml |
| 49571 |
Viscosity and Density Standard D5 |
4.975 |
500 ml |
| 05867 |
Viscosity and Density Standard S6 |
7.237 |
500 ml |
| 05854 |
Viscosity and Density Standard D10 |
10.31 |
500 ml |
| 63484 |
Viscosity and Density Standard N10 |
14.43 |
500 ml |
| 05428 |
Viscosity and Density Standard S20 |
28.98 |
500 ml |
| 18964 |
Viscosity and Density Standard N35 |
55.43 |
500 ml |
| 05397 |
Viscosity and Density Standard S60 |
102 |
500 ml |
| 05395 |
Viscosity and Density Standard N100 |
206 |
500 ml |
| 01437 |
Viscosity and Density Standard S200 |
396.2 |
500 ml |
| 69443 |
Viscosity and Density Standard D500 |
502.5 |
500 ml |
| 04870 |
Viscosity and Density Standard N350 |
740.3 |
500 ml |
| 08670 |
Viscosity and Density Standard D1000 |
1010 |
500 ml |
| 08577 |
Viscosity and Density Standard S600 |
1277 |
500 ml |
| 19495 |
Viscosity and Density Standard N1000 |
2618 |
500 ml |
| 38177 |
Viscosity and Density Standard S2000 |
4638 |
500 ml |
| 73284 |
Viscosity and Density Standard D5000 |
4975 |
500 ml |
| 11492 |
Viscosity and Density Standard D7500 |
7686 |
500 ml |
| 09216 |
Viscosity and Density Standard N4000 |
10296 |
500 ml |
| 50989 |
Viscosity and Density Standard S8000 |
20032 |
500 ml |
| 13219 |
Viscosity and Density Standard N15000 |
40006 |
500 ml |
| 41272 |
Viscosity and Density Standard S30000 |
71382 |
500 ml |
| Back to Top |