Sigma-Aldrich
Analytical / Chromatography > Analytical Standards  > Application Area & Technique > Certified Viscosity Standards
Analytical Standards
Certified Viscosity Standards
Certified Viscosity StandardsFor the calibration and verification of viscosity measuring equipment, analytical standards with well known viscosities are needed.
For this purpose, we supply certified viscosity standards produced from Paragon Scientific in the United Kingdom. They are manufactured in an ISO/IEC17025 and ISO Guide 34 accredited laboratory in accordance with ASTM D 2162 (Standard Practice for basic calibration of master viscosity standard oils) and are traceable to NIST.
In the certificate, the viscosity values and the density are stated at various temperatures as well as expiry date and uncertainties. The Viscosity values are given as kinematic viscosity (mm2/s (cSt)) and as dynamic viscosity (mPa*s (cP) for all the temperatures.
The table below shows the Certified Viscosity and Density standards available at Sigma-Aldrich.
Certified Viscosity and Density Standards from Paragon
Product No. Description Dynamic Viscosity (mPa.s) at 25°C Pack size
67348 Viscosity and Density Standard N 0.8 0.4329 500 ml
19044 Viscosity and Density Standard N 1.0 0.9184 500 ml
01446 Viscosity and Density Standard N 0.8 1.3115 500 ml
93835 Viscosity and Density Standard N2 1.77 500 ml
41859 Viscosity and Density Standard S 3 3.251 500 ml
49571 Viscosity and Density Standard D5 4.975 500 ml
05867 Viscosity and Density Standard S6 7.237 500 ml
05854 Viscosity and Density Standard D10 10.31 500 ml
63484 Viscosity and Density Standard N10 14.43 500 ml
05428 Viscosity and Density Standard S20 28.98 500 ml
18964 Viscosity and Density Standard N35 55.43 500 ml
05397 Viscosity and Density Standard S60 102 500 ml
05395 Viscosity and Density Standard N100 206 500 ml
01437 Viscosity and Density Standard S200 396.2 500 ml
69443 Viscosity and Density Standard D500 502.5 500 ml
04870 Viscosity and Density Standard N350 740.3 500 ml
08670 Viscosity and Density Standard D1000 1010 500 ml
08577 Viscosity and Density Standard S600 1277 500 ml
19495 Viscosity and Density Standard N1000 2618 500 ml
38177 Viscosity and Density Standard S2000 4638 500 ml
73284 Viscosity and Density Standard D5000 4975 500 ml
11492 Viscosity and Density Standard D7500 7686 500 ml
09216 Viscosity and Density Standard N4000 10296 500 ml
50989 Viscosity and Density Standard S8000 20032 500 ml
13219 Viscosity and Density Standard N15000 40006 500 ml
41272 Viscosity and Density Standard S30000 71382 500 ml