Sigma-Aldrich
Analytical / Chromatography > Analytical Standards  > Custom Services > Specialty Packaging
Custom Services
Specialty Packaging
  From this:
spclty_pkging_before
To This:
Specialty Packaging benefits
We can accept any reference material repackaging job, large or small, simple or difficult. Our facilities and equipment enable us to package hygroscopic, light sensitive, or other delicate materials under suitable conditions and into inert atmospheres. Our innovative ampule filling procedure eliminates product burning during the sealing process.
We have experience repackaging
• Biocides
• Diagnostic stains
• Drugs
• Solvents
• Vitamins
Formulation and Packaging Capabilities
Ampul Packaging
Liquids
• Nitrogen or argon blanket in amber glass
• 0.2 mL to 50 mL
• Reduced light conditions
• Varying viscosity and hazards
Solids
• Powders or crystalline materials
• Argon blanket in amber glass
• Single use ampuls
• No burning material in ampul neck
• Blending and sieving as necessary
• 10 mg to 1000 mg
Vials
• Solids and liquids
• Crimp and screw caps
• Wide range of septa materials
Metalized Pouches
• Inert argon atmosphere
• Heat-sealed
• 100 mg to 1000 mg
• Compact single use alternative
Documentation
• Batch production sheets
• Cleaning records
• Material inventory records
• Analytical data
• Environmental conditions
Labeling
• Customized labeling
• Sequential labeling of packaged units
Automated Ampul Filling
Automated ampule filling
View more information, or to request a quote contact Technical Services by phone, fax,
or e-mail.
Phone: 800-359-3041/814-359-3041
Fax: 800-359-3044/814-359-5468
e-mail: techserv@sial.com