Return to Web Version Sigma-Aldrich
Microbiology Standards
Vitroids™ and LENTICULE® discs contain viable microorganisms in a certified quantity (ISO 17025), produced under reproducible conditions (ISO Guide 34) with strains obtained from CECT®, NCTC and other collections. The discs consist of bacteria or fungi in a solid water soluble matrix. Microorganisms in this form are stable for at least one year and are in a viable stage (no lag phase or recovery time).
Search by product name/number, CAS number, or molecular formula
Component  
Search Tips:
  • To look up multiple component mixtures, enter component names separated by commas
  • When searching by CAS No., hyphenation is optional
Search by Agency/Method
Custom Capabilities
Our quality products and solutions allow you to work smarter, enabling the community a safe and healthier world.
Featured Application
LC/MS Analysis of Pain Management Opioids on Ascentis® Express Phenyl-Hexyl
Contact Us
Contact us with any comments or questions.
LENTICULE is a registered trademark of United Kingdom Secretary of State for Health.
Vitroids is a trademark of Sigma-Aldrich Co. LLC.