Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Analytical / Chromatography > Analytical Standards  > Physical Property Standards
Analytical Standards
Physical Property Standards
Buffer Standards
Conductance Standards
MS Markers
Buffer Standard Solutions
Buffer Standard solutions requiring rapid availability and high accuracy of the compounds.
Reliable quality, standard-specific certification, high accuracy and a fast delivery service are the hallmark of Sigma-Aldrich products.
Our product range encompasses Buffer Standard Solutions according to the guideline DIN 19266.
View Product List and Ordering Information
Conductance Standard Solutions
Electrical conductivity can be used to monitor the quality of
  • surface waters
  • process waters in water supply plants and
  • waste waters
The International Organisation for Standardization (ISO) specifies in guideline ISO7888 a method for the measurement of electrical conductivity of all types of water. For this method, 3 conductance standard A, B, C solutions are required. Fluka's conductance standard solutions are prepared according to this guideline. Furthermore, the conductivity measuring equipment is calibrated with standard solutions which are traceable to NIST standards.
Conductance Standard Solutions according ISO 7888
Cat. No.
Product
Description
CAS No.
Package Size
60134 Potassium chloride solution puriss. p.a., Conductance standard A, acc. to ISO 7888 12.90 mS/cm at 25°C 250 ml
60136 Potassium chloride solution puriss. p.a., Conductance standard B, acc. to ISO 7888 1.41 mS/cm at 25°C 250 ml
60138 Potassium chloride solution puriss. p.a., Conductance standard C, acc. to ISO 7888 0.147 mS/cm at 25°C 250 ml
Back to top
Back to Analytical Standards home