Sigma-Aldrich
Analytical / Chromatography > Analytical Standards  > Organic Standards Custom Quote
Analytical / Standards
Organic Standards Custom Quote
We can prepare chemical standards for your:
  • environmental
  • food & beverage
  • petroleum
  • chemical
  • pharmaceutical
applications using carefully monitored raw materials and solvents.
We ship most custom products within 7-10 working days of receiving the order.
star First Name :
star Last Name :
star Company :
space Internal Mail Code :
star Street Address:
space Address (Line 2)
space Department :
space Building / Room :
star City :
space State :
space Province :
star Zip / Postal Code :
star Country :
star Phone Number :
space Fax Number :
star E-Mail Address :
star I wish to receive email updates and offers:
Yes
No
Specifications  
space Mixture Description (for label) :
space Solvent(s) :
space Volume or Weight / ampul:  
space 1 mL
5 mL
10 mL
100 mg
500 mg
Other (please specify)
space Number of units :
(typically minimum order quantity is 4)
space Concentration Units :  
space ng/mL
space µg/mL
space mg/mL
space Other (please specify)
vol / vol %
wt. / wt. %
wt. / vol.
Components CAS No.
(Optional)
Concentration
Select the testing and documentation that best meets your requirements:
Gravimetric
  Components are guaranteed to weigh within ± 0.5% of the amounts you specify. A Certificate of Composition (COC) is included. The COC states the exact amount of raw materials used, the purity and lot number of each raw material.
Qualitative
  The composition is verified by GC or LC. A COC is provided along with a chromatogram.
Quantitative
  The standard is compared to a separately prepared reference batch of the same formulation, using the internal standard method (GC) or the external standard method (LC). A Certificate of Analysis (COA) is included. The COA includes a chromatogram, gravimetric measurements, raw material purities, and the concentration of each component and its uncertainty value. Complimentary data packets are available upon request.
Other (We will contact you to discuss testing requirements)
Please enter any additional comments or instructions for preparation of the standards.
Every custom standard includes a Material Safety Data Sheet (MSDS).
Products are handled under the requirements of our Quality Management System which is registered to ISO 9001:2000.