Vendor – NSI Solutions
Oil & Grease Standards
Save 10% on Oil & Grease Standards
US Customers only
Offer valid through Dec 31, 2009
NSI Oil and Grease PAR (Precision and Recovery) Surrogate SNIPS are packaged in exact volumes in ready-to-use inert, sealed PFA tubes. Simply snip off the top of the tube and pour the entire contents into your extraction vessel.
Also available are the more conventional bottles of oil and grease standards, as well as recovery standards.
All of these products are supplied with a certificate of traceability, listing certified concentrations and acceptance limits.
   Oil & Grease Standards
   Ordering Information
Oil & Grease Standards from NSI Solutions
Oil & Grease Standards
Product Name and Description Additional Information Product No. Price Sale Price
Surrogate Standards
SNIPS - Pkg size 25% larger than competitor's product!
Method 1664 Oil and Grease PAR Surrogate (0.2%)
0.2% n-hexadecane, 0.2% stearic acid in acetone delivers 40mg total oil & grease per tube.
25 x 10 tubes NSI-QC-003LSNIP $105.00 94.50
Method 1664 Oil and Grease PAR Surrogate (0.4%)
0.4% n-hexadecane, 0.4% stearic acid in acetone delivers 80mg total oil & grease per tube
25 x 10 tubes NSI-QC-003SNIP $105.00 94.50
Controls
BOTTLES - Exceptional value, priced 30-50% lower than the competition!
Oil and Grease, Ready-to-use bottles
Works with freon and hexane extraction
500 mL NSI-QCI-069 $39.90 35.90
4 x 500 mL NSI-QCI-069C $117.60 105.85
Oil and Grease, Supplied in Boston Round bottle
Designed for use with SPE
1 Liter NSI-QCI-069-33,400 $80.85 72.80
4 x 1 Liter NSI-QCI-069-33,400C $258.30 232.50
Recovery Standards
Oil and Grease Method 1664 Recovery Standards (0.4%)
0.4%(w/v) n-Hexadecane and 0.4% (w/v) Stearic acid in acetone
25 mL NSI-QC-003 $21.00 18.90
10 x 25 mL NSI-QC-003TP $157.50 141.75
Oil and Grease Method 1664 Recovery Standards (0.2%)
0.2%(w/v) n-Hexadecane and 0.2% (w/v) Stearic acid in acetone
25 mL NSI-QC-0003L $21.00 18.90
10 x 25 mL NSI-QC-003LTP $157.50 141.75
All products are supplied with certificates of traceability listing certified concentrations and acceptance limits.
NSI Solutions is ISO 9001 registered and an A2LA accredited PT provider. (certificate #: 2432.01)
Ordering Information
Please Note: Products from NSI Solutions cannot be ordered through the same processes used to order standard Sigma-Aldrich products. Instead, you must use our special Sigma-Aldrich Vendor Portal Order Form below. Also, at this time, shipments are limited to locations within the US only.
Click to download a PDF order form
Sigma-Aldrich Vendor Portal Order Form (click to view/open PDF)
The above order form allows direct submission by E-mail, or you may print and submit by Fax (to: 1-888-286-7740). 
For Customer Service, to make a payment by credit card, or to place your order by Phone, please contact: 1-800-811-5352.