Sigma-Aldrich
Analytical / Chromatography > Fluka & Riedel-de Haën - Analytical > Fine Chemicals Bulk > High Quality Water
Fine Chemicals Bulk
High Quality Water
Water is indispensable in our life system. Especially the use of high quality water is essential in a broad range of chemical applications, such as in the pharma sector, the electronic-, cosmetic- or food industry. Fluka Chemie GmbH, part of the Sigma-Aldrich family, is able to respond to these demands by offering a broad range of high quality water products, both on small as well as on medium-large scale. The quality of the water that enters the purification system determines how many purification cycles have to be run to achieve a certain water quality. Furthermore a highly developed purification system is needed to achieve the right water quality. Due to Fluka`s strategic location in the mountains, as well as its highly developed purification system, high purity water levels can be achieved.
On the production side Fluka has established a reliable water source coming from a fresh drinking water supply which is purified through various stages including UV-irradiation (half-way the mountain), microfiltration, reverse osmosis, ion-exchange separation, activated carbon treatment and depending on the different impurities, specialized purification steps (at Buchs itself). There is a continuous batch production, with a modular and expandable purification array.
water-source
Water source in the mountains
Turbidity_control_system water-purification
Turbidity control system and UV-irradiation system, as pretreatment before the water purification stage
On the analytical side highly sensitive on-line-assays are used to control the system to achieve a first pass in the sensitive final assays, which compete favourably with other vendors.
HPLC-gradient-analysis   Trace-anion-analysis-by-IC
Trace-metal-analysis-by-ICP   Trace-silicium-analysis-by-
Trace-enzyme-analysis
From left to right: HPLC, gradient analysis, Trace anion analysis by IC, Trace metal analysis by ICP-MS, Trace silicium analysis by AAS, Trace enzyme analysis and various bioanalytical analysis by luminescence
On the packaging side technologies have been adapted to the water quality, as it is essential not only to reach the right quality at the time of production, but also to maintain that same high quality over an extended period of time. The quality of the packaging material ranges from simple high-density polyethylenecontainers (without binders and catalysts) to hydrolytic class 1 white-glass bottles with specialized caps.
Our range of small packages:
Partly-assortiment
Total-assortiment
Fluka can offer the following broad range of high-purity water products:
Picture
Product Number
Product Name
Package Size
Note
Total amount per palette
100 ml picture
95285
water for endotoxin analysis, ultrafiltred and autoclaved
100 ml
1638 x 100 ml
100 ml picture
95289
water for cell biology, ultrafiltrered and autoclaved
100 ml
1638 x 100 ml
Total set picture
95284
water for molecular biology, DEPC-treated and autoclaved
100 ml, 1 l, 50 l
375 x 1 l
100 ml picture
95286
water for organic trace analysis
100 ml
2070 x 100 ml
100 ml picture
95287
water for luminescence
100 ml
3978 x 100 ml
Total set picture
95276
water oxygen- and carbondioxide-free
100 ml
2070 x 100 ml
Total set picture
95283
water for HPCE
100 ml, 250 ml, 1 l
375 x 1 l
Total set picture
95305
Water, puriss p.a., ACS, for inorganic trace analysis
100 ml, 1 l, 2.5 l
375 x 1 l
162 x 2.5 l
Total set picture
95303
water for UV-spectroscopy
1 l, 2.5 l
162 x 2.5 l
375 x 1 l
1 lt picture
95270
water for HPLC, for gradient elution
1 l, 5 l, 10 l
375 x 1 l
Total set picture
95304
water for HPLC
1 l, 2.5 l
162 x 2.5 l
275 x 1 l
10 lt picture
95280
Purified water Ph. Eur.
1 l
375 x 1 l
10 lt picture
17749
water for analytical applications
10 l, 25 l
45 x 10 l
22 x 25 l
Note: these products can not only be delivered palette-wise, but also as bulkpackage (see below)
All these products can be delivered both in small packages as well as on palettes. For example 95305 (water, puriss p.a., ACS, for inorganic trace analysis) can be delivered per palette of 162 x 2.5 l. A typical batchsize is a series of 12 palettes of 95304 (water for HPLC) (i.e. 5 tons). Beside palette-shipments of these small-packages, all the above high quality water products can be delivered in special-made bulkpackages (of for example 50l, 100 l, 500 l up to 2400 l) on request.
Our more then 12 different high-purity water products have been responsible for many of our success stories. Can you afford to work without the best quality water products? Rely on our high-purity Fluka water products as a base of your success!
For detailed information concerning our production and analysis of Fluka`s high quality water products or our type of bulkpackages please contact us (at Fluka) at mweber2@sial.com. For obtaining a quote, please contact your local Sigma-Aldrich Office.
Back to Top
Back to Fine Chemicals/Bulk home