Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Fluka Quality
Packaging
User- and Environment- Friendly Packaging
New VOLPAC®
Fluka Packaging
Our popular 10 lt-containers are now also available as 5 lt-containers for volumetric solutions ready for use
A 5 lt or 10 lt-VOLPAC®-container consists of a flexible polyethylene bag with an outlet tap and a cubic cardboard.Due to the concentration of the bag no volume compensation by access of air is necessary during the emptying process and VOLPAC®-containers can be drained to the dregs without contamination!
Advantages of VOLPAC®-containers:
  • high quality of content and packing
  • easy handling
  • no contamination upon emptying
  • reduced storage place
  • reduced packing material
  • easy disposal of empty containers
For a detailed product list, please click here
Fluka Packaging
Fluka is dedicated to deliver the desired product not only in the right quality, but also to deliver it in the correct package material. Whether 100 mg or 200 l of a chemical product, we make sure that the product is packed accordingly.
The choice of package material is aimed to protect product, people and environment and to maintain the high product quality by avoiding any contamination. Following strict safety compliance rules a safe transport and storage can take place, as well as safe and contaminant-free use of the product.
To meet all requirements we continuously improve our package material.
The materials used are in general glass, polyethylene, metal and cardboard. All these materials are disposable or can be recycled.
If you are interested in a specific type of package material, "custom-made" packaging or have got any questions regarding our package material, please contact:
Flukatec@sial.com
fl_package1
fl_package2
We also have developed a number of environmentally friendly, safe returnable stainless steel containers for laboratory chemicals and high purity solvents – ranging from the 7 L bottle to the 185 L drum. As well as contributing to resource conservation through multiple use, these containers help to preserve the quality of the high grade contents. The program is rounded off by dispensing systems that are safe and easy to handle.
Back to top
Back to Quality overview