Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Analytical / Chromatography > Food Safety > Emerging Contaminants
Food Safety
Emerging Contaminants
Acrylamides
Perchlorate (ClO4-)
Benzene
Perfluoroctanoic acid (PFOA)
Phytoestrogens
Acrylamides back to top
Acrylamides are found in foods processed at high temperatures such as potato chips, French fries, breakfast cereals, and crisp bread. These compounds have been shown to cause tumors in laboratory animals. Fluka offers an Acrylamide kit  to make determination faster and easier with all the necessary standards and solvents for extraction and quantification.
Applications
Acrylamide in Fried Potato Chips
Analysis of Acrylamide and Acrylic Acid on Discovery® HS F5
Sigma-Aldrich Acrylamide kit method (pdf)
Official Methods
FDA Detection and Quantitation of Acrylamide in Foods
Literature & References
European Food Safety Authority - Acrylamide
Canada Food Safety - Health Canada - Acrylamide
WHO Food Safety - Acrylamide FAQ
Related Products
Ready-to-use Kit for Acrylamide Determination (72615)
SPE Tubes
  Discovery DSC-18, 1g/6mL (52606-U)
  Discovery DSC-MCAX, 300 mg/3mL (52784-U)
HPLC Columns
  Discovery HS F5, 15 cm × 4.6 mm I.D., 5 µm particles (567516-U)
Perchlorate (ClO4-) back to top
Perchlorate is an anion that originates as a contaminant in ground water and surface waters from the dissolution of ammonium, potassium, magnesium, or sodium salts. It occurs naturally in the environment and is also manufactured as an industrial chemical for use in rocket fuel, fertilizers, fireworks, flares and other explosive products. It has been found to cause problems associated with the thyroid gland as a result of interference with iodide uptake. In 2004, the FDA found the chemical in milk and lettuce from 15 states, including some apparently uncontaminated areas, showing that human exposure may come from more sources than expected.
On August 19, 2009, EPA published a Supplemental Request for Comments Federal Register notice seeking input on additional ways to analyze data related to the regulatory determination of perchlorate. The EPA has set a provisional action level for drinking water of 32 µg/L (ppb) and more states have also established provisional levels, including California, Texas, Nevada and Arizona, with those of California and Nevada currently the most stringent, at 18 µg/L (ppb).
Applications
Highly Sensitive Detection of Organic and Inorganic Anions with Di- or Tricationic LC/MS Additives (Analytix Issue 2, 2009, pg 12-T409022) (pdf)
Official Methods
FDA - Rapid Determination of Perchlorate Anion in Foods by Ion Chromatography-Tandem Mass Spectrometry
Literature & References
EPA Perchlorate in Drinking Water
EPA CLU-IN Contaminated Site Clean-Up Information - Perchlorate
FDA Perchlorate Questions
Related Products
SPE
  Supelclean ENVI-Carb SPE Cartridge, 500 mg/6 mL (57094)
  Visiprep 24-port SPE Vacuum Manifold (57250-U)
Analytical Standards
  Sodium Perchlorate 18O4 (631981)
  Sodium Perchlorate Monohydrate, ACS Reagent (71853)
Benzene back to top
Benzene is a volatile organic carbon (VOC) and a known carcinogenic compound with a notorious history for causing cancers in industrial manufacturing applications. In 1990, the FDA found it in carbonated beverages, whereupon further investigation was warranted. It was determined that sodium or potassium benzoate and ascorbic acid in certain conditions (heat and light) produce benzene through a basic chemical reaction in which carbon dioxide is removed (decarboxylation) from the benzoate. The EPA regulates benzene in drinking water but there is no mandate for sodas or other beverages.
Several country regulatory agencies have set limits on benzene content in drinking water.
United States: 5 ppb
*Special State Limits: California, Connecticut, New Jersey, and Florida: 1 ppb
** The EPA and California have set public health goals for benzene of 0 ppb and 0.15 ppb, respectively  
Canada: 5 ppb
European Union: 1 ppb
Republic of Korea (South Korea): 10 ppb
World Health Organization (WHO): 10 ppb (benzene should be avoided whenever technically feasible.)
Official Methods
FDA Method-Determination of Benzene in Soft Drinks and Other Beverages
Literature & References
FDA Data on Benzene in Soft Drinks and Other Beverages
International Council of Beverages Association (ICBA) Guidance Document to Mitigate the Potential for Benzene Formation in Beverages (pdf)
Related Products
GC Columns
  SPB-624 Capillary Column, 30 m x 0.25 mm I.D, 1.40 µm (24255)
  Equity-1 Capillary Column, 60 m x 0.25 mm I.D., 0.25 µm (28047-U)
  SLB-5ms Capillary Column, 30 m x 0.25 mm I.D., 0.25 µm (28471-U)
Sample Preparation
  Purge Trap J - BTEX
Analytical Standards
  Benzene, puriss. p.a. standard for GC, >99.9% (12540)
  OEKANAL Benzene Standard Purity >99% (46186)
  Benzene-d6 Purity >99.6% (175870)
Perfluoro-octanoic acid (PFOA) back to top
Perfluoro-octanoic acid is also known as C8 or perfluorooctanoate and does not occur naturally in the environment. It is used to make fluoropolymers for use in products such as non-stick coatings for cookware and weather-proof clothing. PFOAs are persistent organic pollutants, toxic and carcinogenic to animals. It is most often found in human blood rather than in fatty tissues. Previous studies have indicated that PFOA was metabolically inert in mammals, but recent metabolism studies with related fluorochemicals suggested that PFOA might form a glucuronide conjugate. PFOAs have been detected in industrial waste, stain resistant carpets, carpet cleaning liquids, house dust, microwave popcorn bags, water, food, and PTFE. There is uncertainty in how people are exposed to PFOAs. Non-stick cookware coating has been suspected in the past. However, the leading theory is that the most likely route to human exposure is through agricultural means where the land is spread with biosolids (treated sewage sludge).
The US EPA does not have an estabilished limit of exposure for this compound however, a provisional health advisory of 0.4 ppb has been drafted for PFOAs in soil. At this time, manufacturing companies are not mandated to monitor this contaminant.
Literature & References
EPA Perfluorooctanoic Acid (PFOA) and Fluorinated Telomers
EPA Related Programs and Studies (includes country links)
EU Food Safety Perfluorooctanate sulfonate (PFOS), Perfluorooctanoic acid (PFOA) and their salts Scientific Opinion of the Panel on Contaminants in the Food chain
3M Human Health and the Environment – PFOAs
Related Products
Analytical Standards
  OEKANAL Perfluoroctanoic Acid (33824)
HPLC Columns
  Ascentis RP-Amide, 15 cm x 2.1 mm I.D., 3 µm particles (565302-U)
  Ascentis RP-Amide Guard Cartridge Kit (565373-U)
Phytoestrogens back to top
It is well established, that plants contain substances with estrogenic effects, called phytoestrogens. These substances have been found in numerous plant food products including carrots, peas, lentil, rice, cabbage and soy. They are structurally similar to the mammalian estrogen or estradiol, and show estrogenic properties. However, their estrogenic activity is generally much less than that of human estrogens (estrogenic activity ranges from 1/500 to 1/1000 of the activity of oestradiol). Hence phytoestrogens can act as anti -estrogenic agents by blocking the estrogen receptors and exerting a much weaker estrogenic effect compared with the hormone. As a consequence it has been suggested that they might partly suppress or inhibit normal estrogenic activity in estrogen-responsive tissues such as breast tissue and may reduce the risk of breast cancer. Fluka offers a large number of selected pure flavones, flavonones and isoflavones for the examination of phytoestrogenic effects and analysis of phytoestrogene contents.
Related Products
Analytical Standards
  Food Residue Analysis - Natural Compounds