Accessories
FocusLiner™
Achieve 0.2% RSDs on your GC analyses
FocusLiner inlet liners feature a unique design that prevents the wool plug from shifting during repeated injections or sudden inlet pressure changes. They provide RSD values consistently in the 0.2% range; up to 50-times lower than traditional inlet liners with wool plugs.
NEW! Product Information Sheet (PDF, 368 kb)
We have expanded our line of inlet liners to include the FocusLiner™ series and other configurations previously not available.
Features & Benefits
• High temperature deactivation
• Enhanced sample vaporization
• Precision, accuracy, and reliability
• Maximum sensitivity/detection levels
• Guaranteed to fit
• Competitively priced
FocusLiners incorporate a unique design to prevent shifting of the quartz wool during repeated injections or sudden inlet pressure changes. Tapers inside the FocusLiner hold the glass wool plug in an optimum position.
  tapers
focusliner-tapers
This positioning results in wool plug penetration by the needle tip during an injection. The wool provides a large surface area for sample vaporization, and the wiping action removes residual liquid sample from the needle tip, thus preventing droplet formation. This improves reproducibility as much as 10 fold, reduces solvent tailing, and lowers mass discrimination during split injections without any cost disadvantage compared to alternative liners.
FocusLiners are available for all major G.C. instruments, a variety of injection techniques, and for 0.10 mm I.D. Fast Capillary columns. For technical assistance or help chosing an inlet liner, contact Supelco Technical Service at (800) 359-3041 or e-mail us at techservice@sial.com.
Instrument Manufacturers:
Agilent / Finnigan
PerkinElmer
Shimadzu
Thermo
Varian
Agilent / Finnigan back to top
Split / Splitless
  Style
Dimensions Length x O.D. x I.D. (mm) Mfr Part No.
Application 2879901XU
Cat. No.
Pkg Size
  FocusLiner™ with single taper
28799-liner
78.5 x 6.3 x 4.0 5183-4711
Dirty samples, improved reproducibility, active compounds 2879901-U
2879905-U
2879925-U
 1 ea
 5 ea
25 ea
  FocusLiner™
28798-liner
78.5 x 6.3 x 4.0  
Dirty samples,improved reproducibility 2879801-U
2879805-U
2879825-U
 1 ea
 5 ea
25 ea
  with recessed gooseneck
28797-liner
78.5 x 6.3 x 4.0  
Dirty samples 2879701-U
2879705-U
2879725-U
 1 ea
 5 ea
25 ea
  FAST FocusLiner™
28796-liner
78.5 x 6.3 x 2.3  
For use with 100 µm I.D. columns 2879601-U
2879605-U
2879625-U
 1 ea
 5 ea
25 ea
  FAST FocusLiner™ with single taper
28795-liner
78.5 x 6.3 x 2.3  
For use with 100 µm I.D. columns, dirty samples, active compounds 2879501-U
2879505-U
2879525-U
 1 ea
 5 ea
25 ea
Split
  Style
Dimensions Length x O.D. x I.D. (mm) Mfr Part No.
Application Cat. No. Pkg Size
  straight through
28794-liner
78.5 x 6.3 x 4.0 210-3003
General purpose 2879401-U
2879405-U
2879425-U
 1 ea
 5 ea
25 ea
Splitless
  with recessed gooseneck
28793-liner
78.5 x 6.3 x 2.0  
Active compounds 2879301-U
2879305-U
 1 ea
 5 ea
PerkinElmer back to top
Split / Splitless
  Style
Dimensions Length x O.D. x I.D. (mm) Mfr Part No.
Application Cat. No. Pkg Size
  for AutoSystem and Clarus 500
28792-liner
92 x 6.2 x 4.0  
Dirty samples, improved reproducibility 2879201-U
2879205-U
 1 ea
 5 ea
  for AutoSystem and Clarus 500
28791-liner
92 x 6.2 x 4.0  
Dirty samples, improved reproducibility,
active compounds
2879101-U
2879105-U
 1 ea
 5 ea
  FocusLiner™ for PSS Injector
28789-liner
86.2 x 4.0 x 2.0  
Dirty samples, improved reproducibility 2878901-U
2878905-U
 1 ea
 5 ea
Split
  Straight through for AutoSystem and Clarus 500
28787-liner
92 x 6.2 x 4.0  
General purpose 2878701-U
2878705-U
 1 ea
 5 ea
PTV Liner
  28788-liner
88 x 2.0 x 1.0 N600-2017
  2878801-U
2878805-U
 1 ea
 5 ea
Shimadzu back to top
Split / Splitless
  Style
Dimensions Length x O.D. x I.D. (mm) Mfr Part No.
Application Cat. No. Pkg Size
  FocusLiner™ for 17A with SP-17
28786-liner
95 x 5.0 x 3.4  
Dirty samples, improved reproducibility 2878601-U
2878605-U
 1 ea
 5 ea
  FocusLiner™ for 17A with SP-17, single taper
28784-liner
95 x 5.0 x 3.4  
Dirty samples, improved reproducibility, active compounds 2878401-U
2878405-U
 1 ea
 5 ea
  for 17A with SP-17, with middle gooseneck
28783-liner
95 x 5.0 x 3.4  
General purpose 2878301-U
2878305-U
 1 ea
 5 ea
  FocusLiner™ for 14/15A/16 with SPL-14
28781-liner
99 x 5.0 x 3.4  
Dirty samples, improved reproducibility 2878101-U
2878105-U
 1 ea
 5 ea
  for 17A with SP-17, with recessed gooseneck
28779-liner
99 x 5.0 x 3.4  
Dirty samples 2877901-U
2877905-U
 1 ea
 5 ea
  for 14/15A/16 with SPL-14, with middle gooseneck
28777-liner
99 x 5.0 x 3.4  
General purpose 2877701-U
2877705-U
 1 ea
 5 ea
  FocusLiner™ for 14/15A/16 with SPL-14, single taper
28778-liner
99 x 5.0 x 3.4  
Dirty samples, improved reproducibility, active compounds 2877801-U
2877805-U
 1 ea
 5 ea
  FocusLiner™ for GC-2010 system
28776-liner
99 x 5.0 x 3.4  
Dirty samples, improved reproducibility 2877601-U
2877605-U
 1 ea
 5 ea
Splitless
  for 9A and 16A
28782-liner
139 x 5.0 x 3.4  
Active compounds 2878201-U
2878205-U
 1 ea
 5 ea
Thermo back to top
Split / Splitless
  Style
Dimensions Length x O.D. x I.D. (mm) Mfr Part No.
Application Cat. No. Pkg Size
  FocusLiner™ for TQ
28781-liner
105 x 8.0 x 5  
Dirty samples, improved reproducibility 2877501-U
2877505-U
 1 ea
 5 ea
  for TQ Mega Series 4000, 5000, and 6000 with recessed gooseneck
28769-liner
79.5 x 5.0 x 3.0  
Dirty samples 2876901-U
2876905-U
 1 ea
 5 ea
Split
  for ThermoQuest 8000 and TRACE, straight through
28771-liner
105 x 8 x 5  
Active compounds 2877101-U
2877105-U
 1 ea
 5 ea
  FocusLiner™ for ThermoQuest 80-00 and TRACE
28770-liner
105 x 8 x 5  
for use with 50 mm needle 2877001-U
2877005-U
 1 ea
 5 ea
  for ThermoQuest 8000 and TRACE, straight through
28767-liner
105 x 8 x 3.0  
General purpose 2876701-U
2876705-U
 1 ea
 5 ea
  28762-liner
120 x 2.75 x 1.75  
  2876201-U
2876205-U
 1 ea
 5 ea
  for TQ Mega Series 4000, 5000, and 6000, straight through
28761-liner
79.5 x 5.0 x 3.0  
General purpose 2876101-U
2876105-U
 1 ea
 5 ea
Splitless
  for ThermoQuest 8000 and TRACE, single taper
28774-liner
105 x 8 x 3  
Active compounds 2877401-U
2877405-U
 1 ea
 5 ea
  for ThermoQuest 8000 and TRACE, single taper
28773-liner
105 x 8 x 5  
Active compounds 2877301-U
2877305-U
 1 ea
 5 ea
  FocusLiner™ for ThermoQuest 8000 and TRACE
28772-liner
105 x 8 x 5  
for use with 70 mm needle 2877201-U
2877205-U
 1 ea
 5 ea
  for TQ Mega Series 4000, 5000, and 6000, straight through
28758-liner
79.5 x 5.0 x 2.0  
General purpose 2875801-U
2875805-U
 1 ea
 5 ea
SPME
  for ThermoQuest 8000 and TRACE, straight through
28766-liner
105 x 8 x 0.8  
SPME 2876601-U
2876605-U
 1 ea
 5 ea
PTV Liner
  PTV liner for TRACE 2000
28759-liner
120 x 2.75 x 2.0  
  2875901-U
2875905-U
 1 ea
 5 ea
Varian back to top
Split / Splitless
  Style
Dimensions Length x O.D. x I.D. (mm) Mfr Part No.
Application Cat. No. Pkg Size
  FocusLiner™ with single taper, for CP-1177 injector
28799-liner
78.5 x 6.3 x 4.0 5183-4711
Dirty samples, improved reproducibility, active compounds 2879901-U
2879905-U
2879925-U
 1 ea
 5 ea
25 ea
  FocusLiner™, for CP-1177 injector
28798-liner
78.5 x 6.3 x 4.0  
Dirty samples, improved reproducibility 2879801-U
2879805-U
2879825-U
 1 ea
 5 ea
25 ea
  with recessed gooseneck, for CP-1177 injector
28797-liner
78.5 x 6.3 x 4.0  
Dirty samples 2879701-U
2879705-U
2879725-U
 1 ea
 5 ea
25 ea
  FAST FocusLiner™, for CP-1177 injector
28796-liner
78.5 x 6.3 x 2.3  
For use with 100 µm I.D. columns 2879601-U
2879605-U
2879625-U
 1 ea
 5 ea
25 ea
  FAST FocusLiner™ with single taper, for CP-1177 injector
28795-liner
78.5 x 6.3 x 2.3  
For use with 100 µm I.D. columns, dirty samples, active compounds 2879501-U
2879505-U
2879525-U
 1 ea
 5 ea
25 ea
  FocusLiner™ for 1078/1079 injector, single taper
28757-liner
54 x 5.0 x 3.4  
Dirty samples, improved reproducibility, active compounds 2875701-U
2875705-U
 1 ea
 5 ea
  FocusLiner™ for 1078/1079 injector, dual taper
28755-liner
54 x 5.0 x 3.4  
Dirty samples, improved reproducibility, active compounds 2875501-U
2875505-U
 1 ea
 5 ea
Split
  Style
Dimensions Length x O.D. x I.D. (mm) Mfr Part No.
Application Cat. No. Pkg Size
  straight through, for CP-1177 injector
28794-liner
78.5 x 6.3 x 4.0 210-3003
General purpose 2879401-U
2879405-U
2879425-U
 1 ea
 5 ea
25 ea
  with taper for 1075/1077 injector
28756-liner
72 x 6.3 x 4.0  
Dirty samples, active compounds 2875601-U
2875605-U
 1 ea
 5 ea
  FocusLiner™ for 1075/1077 injector
28754-liner
78.5 x 6.3 x 4.0  
Dirty samples, improved reproducibility 2875401-U
2875405-U
 1 ea
 5 ea
  FocusLiner™ for 1075/1077 injector
28748-liner
78.5 x 6.3 x 4.0  
Dirty samples, improved reproducibility, active compounds 2874801-U
2874805-U
 1 ea
 5 ea
  FAST FocusLiner™ with taper for 1075/1077 injector
28747-liner
72 x 6.3 x 2.3  
For use with 100 µm columns 2874701-U
2874705-U
 1 ea
 5 ea
  FocusLiner™ with top-end restriction for 1075/1077 injector
28749-liner
72 x 6.3 x 4.0  
Dirty samples, improved reproducibility, active compounds 2874901-U
2874905-U
 1 ea
 5 ea
Splitless
  Style
Dimensions Length x O.D. x I.D. (mm) Mfr Part No.
Application Cat. No. Pkg Size
  with recessed gooseneck, for CP-1177 injector
28793-liner
78.5 x 6.3 x 2.0  
Active compounds 2879301-U
2879305-U
 1 ea
 5 ea
  FocusLiner™ with top-end restriction for 1075/1077 injector
28746-liner
74 x 6.3 x 4.0  
Dirty samples, improved reproducibility, active compounds 2874601-U
2874605-U
 1 ea
 5 ea
  straight through for 1078/1079 injector
28752-liner
54 x 5.0 x 0.5  
  2875201-U
2875205-U
 1 ea
 5 ea
TRADEMARKS
FocusLiner is a trademark of SGE.