|
Gas Chromatography
CHIRALDEX BP
|
|
|
Astec Products | Technical Resources | Chiral Services Ordering Information: G-BP
Astec CHIRALDEX G-BP capillary GC columns incorporate a phase consisting of a 2,6-di-O-pentyl-3-butyryl derivative of γ-cyclodextrin. This phase exhibits high selectivity for amino acids, amines, and furans.
| Type / Name |
Derivative |
γ-Phase |
Max. Temp.(°C) Isothermal |
Max. Temp.(°C) Programmed |
| BP / Butyryl |
2,6-di-O-pentyl-3-butyryl |
G-BP |
200 |
220 |
Features
- High selectivity for amino acids, amines, and furans
- Thermally stable
Mechanism Observations
- Alkyl chain on analyte contributes to chiral recognition
- High sample capacity, therefore, primary surface interactions
Size Selectivity
The influence of the inclusion mechanism on selectivity is much reduced and capacities are, therefore, generally higher.
Specific Classes of Compounds Separated
Use this table as a reference to help determine the proper column based on the analyte(s) of interest.
| Type |
Compound Class |
| G-BP |
Amino acids, certain primary amines, and furans. |