Sigma-Aldrich
Analytical / Chromatography > Gas Chromatography > Capillary GC Columns > Astec CHIRALDEX > CHIRALDEX DP
Gas Chromatography
CHIRALDEX DP
Astec Products | Technical Resources  | Chiral Services
Ordering Information: B-DP | G-DP
Astec CHIRALDEX B-DP and G-DP capillary GC columns incorporate a phase consisting of a 2,3-di-O-propionyl-6-t-butyl silyl derivative of either β- or γ-cyclodextrin. These phases exhibit good hydrolytic stability, broad chiral selectivity, and are excellent for aliphatic and aromatic amines. They are also good for many aliphatic and some aromatic esters as well as exhibiting high efficiency and resolution at low retention times for polar racemates.
Type / Name Derivative β-Phase γ-Phase Max. Temp.(°C)
Isothermal
Max. Temp.(°C)
Programmed
DP / Dipropionyl 2,3-di-O-propionyl-6-t-butyl silyl B-DP G-DP 200 220
Features
  • Broad chiral selectivity
  • Good hydrolytic stability
  • Excellent for aliphatic and aromatic amines
  • Good for many aliphatic and some aromatic esters
  • High efficiency and resolution at low retention times for polar racemates
Mechanism Observations
  • Mostly surface interactions
  • Fused ring structures exhibit better selectivity on the G-DP
  • Acids have better selectivity as methyl esters rather than ethyl esters
Size Selectivity
Both speed and sample capacity indicate a surface type mechanism for very polar racemates. Large bulky molecules still require a larger surface area than the B-DP provides, therefore, an increase in selectivity is seen on the G-DP for fused ring structures. The B-DP covered the largest range of molecular sizes. However, the choice between the B-DP and the G-DP is compound-dependent.
Specific Classes of Compounds Separated
Use this table as a reference to help determine the proper column based on the analyte(s) of interest.
Type Compound Class
B-DP Acid esters, amines, lactones, alcohols, and some diols.
G-DP Acid esters (long chain), fused ring/bulky amines, and lactones.