Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Gas Chromatography
CHIRALDEX PM
Astec Products | Technical Resources  | Chiral Services
Ordering Information: B-PM
Astec CHIRALDEX B-PM and Bonded B-PM capillary GC columns incorporate a phase consisting of a 2,3,6-tri-O-methyl derivative of β-cyclodextrin. This phase exhibits broad chiral selectivity and is good for terpenes and tertiary amines.
Type / Name Derivative β-Phase Max. Temp.(°C)
Isothermal
Max. Temp.(°C)
Programmed
PM / Permethyl 2,3,6-tri-O-methyl B-PM 200 220
Features
  • Broad chiral selectivity
  • Strong inclusion/size selectivity
Mechanism Observations
  • Inclusion is the dominant mechanism
  • Along with the DM phases, has the highest temperature stability of the Astec CHIRALDEX line
Size Selectivity
Some size selectivity is evident with this phase. Covers a broad range of molecule sizes.
Specific Classes of Compounds Separated
Use this table as a reference to help determine the proper column based on the analyte(s) of interest.
Type Compound Class
B-PM Acids, alcohols, barbitals, diols, epoxides, esters, hydrocarbons, ketones, lactones, and terpenes.