|
All columns in this line will incorporate a combination of unique advances in polymer synthesis, proprietary surface deactivation chemistry, and innovative manufacturing processes. These columns are characterized by low bleed, inertness, durability, and consistency.
SLB-5ms
The low phenyl content provides essentially a boiling point elution order with a slight increase in selectivity, especially for aromatic compounds. The SLB-5ms is the new column of choice for performing GC-MS analyses of semivolatiles. By addressing the need for low detection limits, easy mass spectral identification, less instrument downtime, great resolution, short analysis times, and long column life, SLB-5ms provides the necessary confidence to make the move from your current column.
USP Code: This column meets USP G27 and G36 requirements.
Phase: Bonded and highly crosslinked; silphenylene polymer virtually equivalent in polarity to poly(5% diphenyl/95% methyl siloxane)
| Temperature Limits: |
0.10 to 0.32 mm I.D.: |
-60 °C to 340 °C (isothermal) |
| |
|
-60 °C to 360 °C (programmable) |
| |
0.53 mm I.D.: |
-60 °C to 330 °C (isothermal) |
| |
|
-60 °C to 340 °C (programmable) |
Operating Conditions: Chemically compatible with water and other injection solvents. Sensitive to strong inorganic acids and bases, stable to low levels of HCl in non-aqueous samples. Not damaged by organic acids or bases. Columns can be rinsed.
|