|
FAME
Animal Source Omegawax 100
|
|
|
| Animal Source FAMEs on the Omegawax 100 |
| Conditions |
| Column: |
Omegawax 100, 15 m x 0.10 mm I.D., 0.10 µm (23399-U) |
| Oven: |
140 °C, 40 °C/min. to 280 °C (2 min.) |
| Inj.: |
250 °C |
| Det.: |
FID, 280 °C |
| Carrier Gas: |
hydrogen, 50 cm/sec constant |
| Injection: |
0.2 µL, 200:1 split |
| Liner: |
4 mm I.D., split, cup design |
| Sample: |
PUFA No. II – Animal Source (47015-U), diluted to 50 mg/mL in methylene chloride |
| Peak IDs |
| 1. |
C16:0 |
| 2. |
C18:0 |
| 3. |
C18:1n9 |
| 4. |
C18:1n7 |
| 5. |
C18:2n6 |
| 6. |
C18:3n6 |
| 7. |
C20:0 |
| 8. |
C20:1n9 |
| 9. |
C20:2n9 |
| 10. |
C20:3n6 |
| 11. |
C20:4n6 |
| 12. |
C20:5n3 |
| 13. |
C22:5n6 |
| 14. |
C22:5n3 |
| 15. |
C22:6n3 |