|
| Bacterial Acid Methyl Esters (BAMEs) on the Equity-1 |
| Conditions |
| Column: |
Equity-1, 15 m x 0.10 mm I.D., 0.10 µm (28039-U) |
| Oven: |
175 °C, 30 °C/min. to 275 °C (1 min.) |
| Inj.: |
280 °C |
| Det.: |
FID, 280 °C |
| Carrier Gas: |
hydrogen, 45 cm/sec constant |
| Injection: |
0.5 µL, 200:1 split |
| Liner: |
4 mm I.D., split, cup design |
| Sample: |
Bacterial Acid Methyl Ester (BAME) Mix (47080-U), methyl ester derivatives of a naturally occurring mix of bacterial fatty acids, total concentration of 10 mg/mL in methyl caproate |
| Peak IDs |
| 1. |
Methyl 2-hydroxydecanoate (2-OH-C10:0) |
| 2. |
Methyl undecanoate (C11:0) |
| 3. |
Methyl dodecanoate (C12:0) |
| 4. |
Methyl 2-hydroxydodecanoate (2-OH-C12:0) |
| 5. |
Methyl 3-hydroxydodecanoate (3-OH-C12:0) |
| 6. |
Methyl tridecanoate (C13:0) |
| 7. |
Methyl tetradecanoate (C14:0) |
| 8. |
Methyl 2-hydroxytetradecanoate (2-OH-C14:0) |
| 9. |
Methyl 3-hydroxytetradecanoate (3-OH-C14:0) |
| 10. |
Methyl pentadecanoate (C15:0) |
| 11. |
Methyl 13-methyltetradecanoate (i-C15:0) |
| 12. |
Methyl 12-methyltetradecanoate (α-C15:0) |
| 13. |
Methyl hexadecanoate (C16:0) |
| 14. |
Methyl 14-methylpentadecanoate (i-C16:0) |
| 15. |
Methyl-2-hydroxyhexadeanoate (2-OH-C16:0) |
| 16. |
Methyl cis-9-hexadecenoate (C16:19) |
| 17. |
Methyl heptadecanoate (C17:0) |
| 18. |
Methyl 15-methylhexadecanoate (i-C17:0) |
| 19. |
Methyl cis-9,10-methylenehexadecanoate (C17:0Δ) |
| 20. |
Methyl octadecanoate (C18:0) |
| 21. |
Methyl cis-9-octadecenoate (C18:19) |
| 22. |
Methyl trans-9-octadecenoate (C18:19) and cis-11-octadecenoate (C18:111) |
| 23. |
Methyl cis-9,12-octadecadienoate (C18:29,12) |
| 24. |
Methyl nonadecanoate (C19:0) |
| 25. |
Methyl cis-9,10-methyleneoctadecanoate (C19:0Δ) |
| 26. |
Methyl eicosanoate (C20:0) |