Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
FAME
FAME Isomer 75m SP-2560
Detailed Analysis of C18 FAME Isomers on the 75 m SP-2560
Conditions
Column: SP-2560, 75 m x 0.18 mm I.D., 0.14 µm (23348-U)
Oven: 180 °C
Inj.: 220 °C
Det.: FID, 220 °C
Carrier Gas: hydrogen, 25 cm/sec @ 180 °C
Injection: 0.5 µL, 100:1 split
Liner: 4 mm I.D., split, cup design
Sample: Mixture of C18:1, C18:2, and C18:3 FAMEs in methylene chloride
g003460
Peak IDs
1. C18:1 Δ 7t and C18:1 Δ 6t
2. C18:1 Δ 9t
3. C18:1 Δ 11t
4. C18:1 Δ 12t, C18:1 Δ 6c, C18:1 Δ 7c and C18:1 Δ 13t
5. C18:1 Δ 9c
6. C18:1 Δ 11c
7. C18:1 Δ 12c
8. C18:1 Δ 13c
9. C18:2 Δ 9t, 12t
10. C18:2 Δ 9c, 12t
11. C18:2 Δ 9t, 12c
12. C18:2 Δ 9c, 12c
13. C18:3 Δ 9t, 12t, 15t
14. C18:3 Δ 9t, 12t, 15c
15. C18:3 Δ 9t, 12c, 15t
16. C18:3 Δ 9c, 12t, 15t and C18:3 Δ 9c, 12c, 15t
17. C18:3 Δ 9c, 12t, 15c
18. C18:3 Δ 9t, 12c, 15c
19. C18:3 Δ 9c, 12c, 15c