| Peak IDs |
| 1. |
Butyric Acid Methyl Ester (C4:0) at 4 wt % |
| 2. |
Caproic Acid Methyl Ester (C6:0) at 4 wt % |
| 3. |
Caprylic Acid Methyl Ester (C8:0) at 4 wt % |
| 4. |
Capric Acid Methyl Ester (C10:0) at 4 wt % |
| 5. |
Undecanoic Acid Methyl Ester (C11:0) at 2 wt % |
| 6. |
Lauric Acid Methyl Ester (C12:0) at 4 wt % |
| 7. |
Tridecanoic Acid Methyl Ester (C13:0) at 2 wt % |
| 8. |
Myristic Acid Methyl Ester (C14:0) at 4 wt % |
| 9. |
Myristoleic Acid Methyl Ester (C14:1) at 2 wt % |
| 10. |
Pentadecanoic Acid Methyl Ester (C15:0) at 2 wt % |
| 11. |
cis-10-Pentadecenoic Acid Methyl Ester (C15:1) at 2 wt % |
| 12. |
Palmitic Acid Methyl Ester (C16:0) at 6 wt % |
| 13. |
Palmitoleic Acid Methyl Ester (C16:1) at 2 wt % |
| 14. |
Heptadecanoic Acid Methyl Ester (C17:0) at 2 wt % |
| 15. |
cis-10-Heptadecenoic Acid Methyl Ester (C17:1) at 2 wt % |
| 16. |
Stearic Acid Methyl Ester (C18:0) at 4 wt % |
| 17. |
Oleic Acid Methyl Ester (C18:1n9c) at 4 wt % |
| 18. |
Elaidic Acid Methyl Ester (C18:1n9t) at 2 wt % |
| 19. |
Linoleic Acid Methyl Ester (C18:2n6c) at 2 wt % |
| 20. |
Linolelaidic Acid Methyl Ester (C18:2n6t) at 2 wt % |
| 21. |
γ-Linolenic Acid Methyl Ester (C18:3n6) at 2 wt % |
| 22. |
α-Linolenic Acid Methyl Ester (C18:3n3) at 2 wt % |
| 23. |
Arachidic Acid Methyl Ester (C20:0) at 4 wt % |
| 24. |
cis-11-Eicosenoic Acid Methyl Ester (C20:1n9) at 2 wt % |
| 25. |
cis-11,14-Eicosadienoic Acid Methyl Ester (C20:2) at 2 wt % |
| 26. |
cis-8,11,14-Eicosatrienoic Acid Methyl Ester (C20:3n6) at 2 wt % |
| 27. |
cis-11,14,17-Eicosatrienoic Acid Methyl Ester (C20:3n3) at 2 wt % |
| 28. |
Arachidonic Acid Methyl Ester (C20:4n6) at 2 wt % |
| 29. |
cis-5,8,11,14,17-Eicosapentaenoic Acid Methyl Ester (C20:5n3) at 2 wt % |
| 30. |
Heneicosanoic Acid Methyl Ester (C21:0) at 2 wt % |
| 31. |
Behenic Acid Methyl Ester (C22:0) at 4 wt % |
| 32. |
Erucic Acid Methyl Ester (C22:1n9) at 2 wt % |
| 33. |
cis-13,16-Docosadienoic Acid Methyl Ester (C22:2) at 2 wt % |
| 34. |
cis-4,7,10,13,16,19-Docosahexaenoic Acid Methyl Ester (C22:6n3) at 2 wt % |
| 35. |
Tricosanoic Acid Methyl Ester (C23:0) at 2 wt % |
| 36. |
Lignoceric Acid Methyl Ester (C24:0) at 4 wt % |
| 37. |
Nervonic Acid Methyl Ester (C24:1n9) at 2 wt % |