|
| Short Chain Free Fatty Acids on the Nukol |
| Conditions |
| Column: |
Nukol, 30 m x 0.25 mm I.D., 0.25 µm (24107) |
| Oven: |
185 °C |
| Det.: |
FID |
| Carrier Gas: |
helium, 20 cm/sec |
| Injection: |
1 µL, 100:1 split |
| Sample: |
Volatile Free Acid Mix (46975-U), each analyte at 10 mM in deionized water |
| Peak IDs |
| 1. |
Acetic acid |
| 2. |
Propionic acid |
| 3. |
Isobutyric acid |
| 4. |
Butyric acid |
| 5. |
Isovaleric acid |
| 6. |
Valeric acid |
| 7. |
Isocaproic acid |
| 8. |
Caproic acid |
| 9. |
Heptanoic acid |