|
| Short and Long Chain Free Fatty Acids on the Nukol |
| Conditions |
| Column: |
Nukol, 15 m x 0.53 mm I.D., 0.50 µm (25326) |
| Oven: |
100 °C, 10 °C/min. to 220 °C |
| Det.: |
FID |
| Carrier Gas: |
helium, 30 mL/min. |
| Injection: |
0.5 µL, direct injection |
| Sample: |
16 analytes, at various concentrations from 50 to 800 µg/mL |
| Peak IDs |
| 1. |
Acetic acid |
| 2. |
Propionic acid |
| 3. |
Isobutyric acid |
| 4. |
Butyric acid |
| 5. |
Isovaleric acid |
| 6. |
Valeric acid |
| 7. |
Isocaproic acid |
| 8. |
Caproic acid |
| 9. |
Heptanoic acid |
| 10. |
Octanoic acid |
| 11. |
Decanoic acid |
| 12. |
Dodecanoic acid |
| 13. |
Tetradecanoic acid |
| 14. |
Hexadecanoic acid |
| 15. |
Octadecanoic acid |
| 16. |
Eicosanoic acid |