|
FAME
Test Mix Omegawax 320
|
|
|
| Omegawax Test Mix on the Omegawax 320 |
| Conditions |
| Column: |
Omegawax 320, 30 m x 0.32 mm I.D., 0.25 µm (24152) |
| Oven: |
200 °C |
| Inj.: |
250 °C |
| Det.: |
FID, 260 °C |
| Carrier Gas: |
helium, 25 cm/sec @ 200 °C |
| Injection: |
1 µL, 100:1 split |
| Sample: |
Omegawax Column Test Mix (48476), methyl ester derivatives of a naturally occurring mix of fatty acids, total concentration of 50 mg/mL in hexane |
| Peak IDs |
| 1. |
C14:0 |
| 2. |
C16:0 |
| 3. |
C18:0 |
| 4. |
C18:1n9 |
| 5. |
C18:2n6 |
| 6. |
C18:4n3 |
| 7. |
C20:0 |
| 8. |
C20:1n9 |
| 9. |
C20:5n3 |
| 10. |
C21:5n3 |
| 11. |
C22:0 |
| 12. |
C22:5n3 |
| 13. |
C22:6n3 |
| 14. |
C23:0 |
| 15. |
C24:0 |
| 16. |
C24:1 |