|
FAME
FAME Solid Phase Extraction
|
|
|
| Discovery Ag-Ion (silver-ion) SPE |
When silver ions are loaded and immobilized on to an SCX phase as counter-ions, they have the ability to form polar complexes with unsaturated fatty acid double bonds under normal-phase conditions. Discovery Ag-Ion SPE was developed for the fractionation cis/trans isomers, and can also resolve FAMEs by degree of unsaturation in which retention strength increases with increasing number of double bonds. As a result, Discovery Ag-Ion SPE allows users to fractionate FAME samples prior to GC analysis thereby simplifying analytical chromatography and improving method accuracy.
Discovery Ag-Ion SPE phases are loaded with silver using a proprietary procedure to offer optimal resolution, performance, and capacity. Each lot is tested and quality controlled for cis/trans FAME resolution.
|
- Specifically developed for the fractionation of FAMEs based on degree of unsaturation, and the resolution of cis- and trans- isomers
- Phase is loaded with silver using a proprietary procedure to offer optimal resolution, performance, and capacity
- Each lot is tested and quality controlled for cis- and trans- FAME resolution
- 750mg bed weight has a high capacity of 1mg FAMEs
|
| |
| Description |
Quantity |
Cat. No. |
| Discovery Ag-Ion SPE |
|
|
| 750 mg / 6 mL SPE Tube |
30 |
54225-U |
| 750 mg / 1 mL Rezorian™ cartridge |
10 |
54226-U |
back to FAME main page