Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
FAME
FAME Solvents & Reagents
  Blood fatty acid assessment
  CHROMASOLV® Solvents
  Esterification and Transesterification Reagents
Blood fatty acid assessment    
Kits for convenient collection of blood drops, their storage, shipment, and processing the samples for fatty acid analysis ( (3-n and 6-n polyunsaturated fatty acids) via gas chromatography.
These kits allow efficient sample collection and processing for quick collection of analytical information on the fatty acid content in the blood samples. Subsequently, it helps the care providers in development and application of adequate preventive dietary strategies for an individual.
Description Pkg. Cat. No.
Blood Collection Kit 1 Kit 11312
Derivatization Kit 1 Kit 05904
CHROMASOLV® Solvents
Description Pkg. Sizes Cat. No.
CHLOROFORM, for HPLC, >=99.8%, stabilized with amylenes 100 ml
6 x 1L
4 x 4L
34854
CHLOROFORM, for HPLC, >=99.9%, contains 0.5-1.0% ethanol 100 ml
6 x 1L
4 x 4L
366927
HEPTANE, for HPLC, >=99% 100 ml
6 x 1L
4 x 4L
34873
DICHLOROMETHANE, for HPLC, >=99.8% 100 ml
6 x 1L
4 x 4L
34856
Related products:    
Click here to see our complete line of CHROMASOLV solvents    
Esterification and Transesterification Reagents    
Esterification is the most popular alkylation method. Alkyl esters offer excellent stability, and provide quick and quantitative samples for GC analysis.
Esterification is the reaction of an acid with an alcohol in the presence of a catalyst to form an ester. The process involves the condensation of the carboxyl group of the acid and the hydroxyl group of the alcohol, with the elimination of water. Esterification is best done in the presence of a catalyst (e.g., hydrogen chloride), which is removed with the water.
Transesterification is the displacement of the alcohol from an ester by another alcohol. This has been widely used for making esters of higher alcohols from those of lower alcohols. Transesterification can be performed with an acidic or basic catalyst using methanol to react with fats and oils.
Description Pkg. Cat. No.
BF3 - Butanol 10 % (w/w) 10 × 5 mL 33126-U
BF3 - Butanol 10 % (w/w) 100 mL 33125-U
BF3 - Methanol 10 % (w/w) 20 × 1 mL 33356
BF3 - Methanol 10 % (w/w) 19 × 2 mL 33020-U
BF3 - Methanol 10 % (w/w) 10 × 5 mL 33040-U
BF3 - Methanol 10 % (w/w) 400 mL 33021
Methanolic Base 0.5 N 30 mL 33352
Methanolic Base 0.5 N 100 mL 33080
Methanolic HCl 0.5 N 20 × 1 mL 33354
Methanolic HCl 0.5 N 10 × 5 mL 33095
Methanolic HCl 3 N 20 × 1 mL 33355
Methanolic HCl 3 N 10 × 3 mL 33051
Methanolic HCl 3 N 400 mL 33050-U
Methanolic H2SO4 10 % (v/v) in methanol 6 × 5 mL 506516
View all Derivatization Reagents in the online catalog.
 Back to FAME main page