Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Analytical / Chromatography > Gas Chromatography > Fatty Acid/FAME Analysis > FAME Vials, Syringes & Labware
FAME
FAME Vials, Syringes & Labware
  Micro Reaction Vessel
  Syringes
  Block heater
Micro Reaction Vessel       back to top
micro_reaction
These vials are made of heavy wall borosilicate glass. They have a cone shape interior for easy removal of small samples. They are supplied with Teflon-faced rubber septum and cap. These products can be autoclaved or centrifuged. We also offer Mininert valves, replacement septa, and the septa inserter.
Volume (mL) O.D. (mm) Height (mm) Depth (mm) Cap Pkg Cat. No.
Screw top, clear glass, graduated with hole cap (phenolic black)
Dimensions: Height with cap, depth from septum to bottom of cone.
3.0 20 46 43 20-400 12 ea 33297
5.0 20 61 58 20-400 12 ea 33299
Screw top, amber glass with hole cap (phenolic black)
Dimensions: Height without cap.
3.0 20 47 45 20-400 12 ea 27477-U
5.0 20 62 60 20-400 12 ea 27478-U
Related products:        
Click here to see our complete line of Vials        
Syringes         Back to top
Volume (µL) Model Needle Needle Length (mm) Pkg Cat. No.
Hamilton® Microliter™ Syringes for Agilent 7673 & 7683 (fixed needle)
5  75 ASN 23s ga (cone tip) 43 6 ea 21315
5  75 ASN 23s-26s ga (cone tip) 43 6 ea 24571
10 701 ASN 23s ga (cone tip) 43 6 ea 21317
10 701 ASN 26s ga (cone tip) 43 6 ea 21316
10 701 ASN 23s-26s ga (cone tip) 43 6 ea 24574
Hamilton 700 Series, N (cemented needle)
10 701N 26s ga (bevel tip) 51 6 ea 20779
Hamilton 700 Series, RN (removable needle)
10 701RN 26 ga (bevel tip) 51 6 ea 20793
Hamilton 800 Series, N (cemented needle)
5 85N 26s ga (bevel tip) 51 1 ea 20835-U
10 801N 26s ga (bevel tip) 51 1 ea 20795-U
Hamilton 800 Series, RN (removable needle)
5 85RN 26s ga (bevel tip) 51 1 ea 26201
10 801RN 26s ga (bevel tip) 51 1 ea 20797
Related products:
Click here to see our complete line of Syringes
Block heater Back to top
Constant temperatures for derivatization and other reactions. The block heater features:
• Two temperature ranges, ambient to 60 °C, 50 °C to 130 °C
• Three-position power switch and two adjustable thermostats
• Pilot light shows heat cycling operation
Two models: 120 volts (100 watts, 0.83 amps, 50/60 Hz) and 240 volts (100 watts, 0.42 amps, 50/60 Hz).
Order blocks separately. Block 1 has eight holes (21 mm wide and 30 mm deep), and accepts our 3 mL or 5 mL micro reaction vessels. Block 2 has twelve holes (17 mm wide and 24 mm deep), and accepts our 0.1 mL to 2 mL micro reaction vessels.
block-heater
Block heater
(8-hole block not included)
8-hole-block
8-hole block for heater
Description Cat. No.
Block Heater AC input 110 V (block not included) 33315
Block Heater AC input 240 V CE compliant (block not included) 33318-U
Block #1 33316
Block #2 33317-U
Thermometer length 5 in. temperature range -10 °C to 110 °C 57332
Back to FAME main page