|
Gas Chromatography
Solvents for Headspace GC
|
Ideal for USP and Ph.Eur. methods for residual solvent determination by headspace GC Analysis of residual solvents using headspace GC techniques is an important quality control procedure in pharmaceutical and food-related laboratories. With the aim of further ensuring minimal solvent exposure in people taking medications, effective July 1, 2008 USP 467 “Organic Volatile Impurities (OVI)” will change to USP 467 “Residual Solvents.” Sigma-Aldrich is pleased to offer Fluka-brand solvents that have been specifically designed and tested for this important application.
Features and Benefits of Headspace-Grade Solvents from Sigma-Aldrich
- Includes most popular GC-HS solvents for wide applicability - Very high purity to maximize detector sensitivity - Virtually no background due to volatile impurities on both polar and non-polar GC capillary columns
Product range